Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 148955129-148955671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UST
Entrez Gene ID
10090
Gene Name
uronyl 2-sulfotransferase
Gene Aliases
2OST
UniGene
Hs.657370
Gene Chromosome Location
Chr.6: 148745891-149076990 on Build GRCh38
Transcripts
NM_005715.2 XM_011535378.2 XM_017010152.1 NR_038408.1

Gene Symbol
UST-AS1
Entrez Gene ID
100128176
Gene Name
UST antisense RNA 1
Gene Aliases
-
UniGene
Hs.557541
Gene Chromosome Location
Chr.6: 148954259-148964684 on Build GRCh38
Transcripts
NM_005715.2 XM_011535378.2 XM_017010152.1 NR_038408.1

SNP Detail

SNP ID Chromosome Location
rs755168183 Chr.6: 148955198-148955198 on Build GRCh38
rs764331905 Chr.6: 148955576-148955576 on Build GRCh38
rs773081489 Chr.6: 148955360-148955360 on Build GRCh38
rs1002787918 Chr.6: 148955578-148955578 on Build GRCh38
rs953618683 Chr.6: 148955284-148955284 on Build GRCh38
rs565096393 Chr.6: 148955296-148955296 on Build GRCh38
rs898238852 Chr.6: 148955491-148955491 on Build GRCh38
rs74978443 Chr.6: 148955331-148955331 on Build GRCh38
rs927139929 Chr.6: 148955268-148955268 on Build GRCh38
rs907863566 Chr.6: 148955640-148955640 on Build GRCh38
rs912165005 Chr.6: 148955326-148955326 on Build GRCh38
rs986370959 Chr.6: 148955316-148955316 on Build GRCh38
rs556252573 Chr.6: 148955191-148955191 on Build GRCh38
rs531267146 Chr.6: 148955431-148955431 on Build GRCh38
rs971298977 Chr.6: 148955215-148955215 on Build GRCh38
rs532999622 Chr.6: 148955407-148955407 on Build GRCh38
rs576102344 Chr.6: 148955202-148955202 on Build GRCh38
rs767193517 Chr.6: 148955337-148955337 on Build GRCh38
rs971240679 Chr.6: 148955180-148955180 on Build GRCh38
rs957017422 Chr.6: 148955154-148955154 on Build GRCh38
rs990882948 Chr.6: 148955320-148955320 on Build GRCh38
rs571152728 Chr.6: 148955493-148955493 on Build GRCh38
rs148134600 Chr.6: 148955149-148955149 on Build GRCh38
rs962782678 Chr.6: 148955250-148955250 on Build GRCh38
rs944944656 Chr.6: 148955338-148955338 on Build GRCh38
rs9485341 Chr.6: 148955632-148955632 on Build GRCh38
rs1009828533 Chr.6: 148955173-148955173 on Build GRCh38
rs983461076 Chr.6: 148955636-148955636 on Build GRCh38
rs577911791 Chr.6: 148955324-148955324 on Build GRCh38
rs545390596 Chr.6: 148955255-148955255 on Build GRCh38
rs973870863 Chr.6: 148955613-148955613 on Build GRCh38
rs892863608 Chr.6: 148955432-148955432 on Build GRCh38
rs1004470090 Chr.6: 148955187-148955187 on Build GRCh38
rs749226674 Chr.6: 148955469-148955469 on Build GRCh38
rs995695676 Chr.6: 148955527-148955527 on Build GRCh38
rs981689451 Chr.6: 148955265-148955265 on Build GRCh38
rs569264451 Chr.6: 148955615-148955615 on Build GRCh38
rs966698513 Chr.6: 148955623-148955623 on Build GRCh38
rs920965720 Chr.6: 148955281-148955281 on Build GRCh38
rs971753986 Chr.6: 148955507-148955507 on Build GRCh38
rs112265624 Chr.6: 148955375-148955375 on Build GRCh38
rs908125160 Chr.6: 148955335-148955335 on Build GRCh38
rs76857025 Chr.6: 148955386-148955386 on Build GRCh38
rs562573113 Chr.6: 148955396-148955396 on Build GRCh38
rs768214397 Chr.6: 148955171-148955171 on Build GRCh38
rs1040978744 Chr.6: 148955340-148955340 on Build GRCh38
rs140715350 Chr.6: 148955586-148955586 on Build GRCh38
rs560861323 Chr.6: 148955608-148955608 on Build GRCh38
rs959758261 Chr.6: 148955290-148955290 on Build GRCh38
rs920697999 Chr.6: 148955619-148955619 on Build GRCh38
rs763308534 Chr.6: 148955343-148955344 on Build GRCh38
rs986443007 Chr.6: 148955593-148955593 on Build GRCh38
rs1009859654 Chr.6: 148955451-148955451 on Build GRCh38
rs534533126 Chr.6: 148955574-148955574 on Build GRCh38
rs185898372 Chr.6: 148955286-148955286 on Build GRCh38
rs973764136 Chr.6: 148955271-148955271 on Build GRCh38
rs900666881 Chr.6: 148955368-148955368 on Build GRCh38
rs7758814 Chr.6: 148955328-148955328 on Build GRCh38
rs35325556 Chr.6: 148955490-148955491 on Build GRCh38
rs76561242 Chr.6: 148955468-148955468 on Build GRCh38
rs144577005 Chr.6: 148955596-148955596 on Build GRCh38

View Full Product Details