Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 165940119-165940623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00473
Entrez Gene ID
90632
Gene Name
long intergenic non-protein coding RNA 473
Gene Aliases
C6orf176, bA142J11.1
UniGene
Hs.31917
Gene Chromosome Location
Chr.6: 165924048-165988039 on Build GRCh38
Transcripts
NR_026860.1 NR_026861.1 XM_011535387.2 XM_017010194.1 XM_017010197.1

Gene Symbol
PDE10A
Entrez Gene ID
10846
Gene Name
phosphodiesterase 10A
Gene Aliases
ADSD2, HSPDE10A, IOLOD, LINC00473, PDE10A19
UniGene
Hs.348762
Gene Chromosome Location
Chr.6: 165327287-165988121 on Build GRCh38
Transcripts
NR_026860.1 NR_026861.1 XM_011535387.2 XM_017010194.1 XM_017010197.1

SNP Detail

SNP ID Chromosome Location
rs1026907807 Chr.6: 165940386-165940386 on Build GRCh38
rs559928856 Chr.6: 165940326-165940326 on Build GRCh38
rs1017042705 Chr.6: 165940412-165940412 on Build GRCh38
rs932328922 Chr.6: 165940482-165940482 on Build GRCh38
rs943691841 Chr.6: 165940163-165940163 on Build GRCh38
rs542358842 Chr.6: 165940354-165940354 on Build GRCh38
rs1022993019 Chr.6: 165940325-165940325 on Build GRCh38
rs914810494 Chr.6: 165940265-165940265 on Build GRCh38
rs1041137898 Chr.6: 165940169-165940169 on Build GRCh38
rs908308675 Chr.6: 165940151-165940151 on Build GRCh38
rs553541851 Chr.6: 165940299-165940299 on Build GRCh38
rs12208047 Chr.6: 165940559-165940559 on Build GRCh38
rs895901862 Chr.6: 165940307-165940307 on Build GRCh38
rs971169233 Chr.6: 165940466-165940466 on Build GRCh38
rs914528124 Chr.6: 165940552-165940552 on Build GRCh38
rs1050825279 Chr.6: 165940363-165940363 on Build GRCh38
rs1002154076 Chr.6: 165940364-165940364 on Build GRCh38
rs943646576 Chr.6: 165940282-165940282 on Build GRCh38
rs908275489 Chr.6: 165940454-165940454 on Build GRCh38
rs924694555 Chr.6: 165940495-165940495 on Build GRCh38
rs1031732069 Chr.6: 165940285-165940285 on Build GRCh38
rs184420710 Chr.6: 165940314-165940314 on Build GRCh38
rs574957164 Chr.6: 165940318-165940318 on Build GRCh38
rs565914228 Chr.6: 165940469-165940469 on Build GRCh38
rs141154954 Chr.6: 165940570-165940571 on Build GRCh38
rs1044407102 Chr.6: 165940582-165940582 on Build GRCh38
rs956147315 Chr.6: 165940508-165940508 on Build GRCh38
rs549369286 Chr.6: 165940445-165940445 on Build GRCh38
rs386708416 Chr.6: 165940571-165940572 on Build GRCh38
rs537417092 Chr.6: 165940599-165940599 on Build GRCh38
rs572022193 Chr.6: 165940337-165940337 on Build GRCh38
rs67996003 Chr.6: 165940455-165940455 on Build GRCh38
rs757034282 Chr.6: 165940208-165940208 on Build GRCh38
rs1043248653 Chr.6: 165940498-165940498 on Build GRCh38
rs934816243 Chr.6: 165940538-165940538 on Build GRCh38
rs547254534 Chr.6: 165940465-165940465 on Build GRCh38
rs866275303 Chr.6: 165940380-165940380 on Build GRCh38
rs904664290 Chr.6: 165940244-165940244 on Build GRCh38
rs952502591 Chr.6: 165940415-165940415 on Build GRCh38
rs187666144 Chr.6: 165940463-165940463 on Build GRCh38
rs548662548 Chr.6: 165940572-165940572 on Build GRCh38
rs1053725070 Chr.6: 165940539-165940539 on Build GRCh38
rs189306083 Chr.6: 165940408-165940408 on Build GRCh38
rs1054423055 Chr.6: 165940206-165940206 on Build GRCh38
rs934652398 Chr.6: 165940184-165940184 on Build GRCh38
rs749992715 Chr.6: 165940436-165940436 on Build GRCh38
rs926225507 Chr.6: 165940536-165940536 on Build GRCh38
rs553127224 Chr.6: 165940245-165940245 on Build GRCh38
rs56386360 Chr.6: 165940146-165940146 on Build GRCh38
rs41269579 Chr.6: 165940587-165940587 on Build GRCh38
rs182141006 Chr.6: 165940443-165940443 on Build GRCh38
rs990096227 Chr.6: 165940524-165940524 on Build GRCh38
rs1039299472 Chr.6: 165940300-165940300 on Build GRCh38

View Full Product Details