Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 39404487-39404994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POU6F2
Entrez Gene ID
11281
Gene Name
POU class 6 homeobox 2
Gene Aliases
RPF-1, WT5, WTSL
UniGene
Hs.137106
Gene Chromosome Location
Chr.7: 38977878-39468601 on Build GRCh38
Transcripts
NM_001166018.1 NM_007252.3 XM_011515113.2 NR_046711.1

Gene Symbol
POU6F2-AS1
Entrez Gene ID
100861520
Gene Name
POU6F2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.571292
Gene Chromosome Location
Chr.7: 39404598-39406346 on Build GRCh38
Transcripts
NM_001166018.1 NM_007252.3 XM_011515113.2 NR_046711.1

SNP Detail

SNP ID Chromosome Location
rs979172348 Chr.7: 39404620-39404620 on Build GRCh38
rs909984462 Chr.7: 39404742-39404742 on Build GRCh38
rs1037007838 Chr.7: 39404540-39404540 on Build GRCh38
rs1008279032 Chr.7: 39404805-39404806 on Build GRCh38
rs941342580 Chr.7: 39404515-39404515 on Build GRCh38
rs778556267 Chr.7: 39404684-39404684 on Build GRCh38
rs973213225 Chr.7: 39404823-39404823 on Build GRCh38
rs75000638 Chr.7: 39404644-39404644 on Build GRCh38
rs534464315 Chr.7: 39404579-39404579 on Build GRCh38
rs887168181 Chr.7: 39404550-39404550 on Build GRCh38
rs1041108939 Chr.7: 39404833-39404833 on Build GRCh38
rs1046385612 Chr.7: 39404568-39404568 on Build GRCh38
rs887199163 Chr.7: 39404753-39404753 on Build GRCh38
rs147727338 Chr.7: 39404658-39404658 on Build GRCh38
rs550790405 Chr.7: 39404597-39404597 on Build GRCh38
rs557041411 Chr.7: 39404794-39404795 on Build GRCh38
rs750194552 Chr.7: 39404585-39404585 on Build GRCh38
rs1028889801 Chr.7: 39404888-39404888 on Build GRCh38
rs778855450 Chr.7: 39404758-39404758 on Build GRCh38
rs576765992 Chr.7: 39404699-39404699 on Build GRCh38
rs950356671 Chr.7: 39404567-39404567 on Build GRCh38
rs557809261 Chr.7: 39404591-39404591 on Build GRCh38
rs186852999 Chr.7: 39404551-39404551 on Build GRCh38
rs937513614 Chr.7: 39404958-39404958 on Build GRCh38
rs569068007 Chr.7: 39404652-39404652 on Build GRCh38
rs897411892 Chr.7: 39404557-39404557 on Build GRCh38
rs923709393 Chr.7: 39404642-39404642 on Build GRCh38
rs1046015465 Chr.7: 39404943-39404943 on Build GRCh38
rs1019921289 Chr.7: 39404649-39404649 on Build GRCh38
rs142062717 Chr.7: 39404673-39404673 on Build GRCh38
rs966361869 Chr.7: 39404576-39404576 on Build GRCh38
rs761366162 Chr.7: 39404578-39404578 on Build GRCh38
rs950090383 Chr.7: 39404926-39404926 on Build GRCh38
rs1032731009 Chr.7: 39404606-39404606 on Build GRCh38
rs918618779 Chr.7: 39404858-39404858 on Build GRCh38
rs991794238 Chr.7: 39404645-39404645 on Build GRCh38
rs555519508 Chr.7: 39404746-39404746 on Build GRCh38
rs996527107 Chr.7: 39404852-39404852 on Build GRCh38
rs920113255 Chr.7: 39404687-39404687 on Build GRCh38
rs572401716 Chr.7: 39404851-39404851 on Build GRCh38
rs10259786 Chr.7: 39404952-39404952 on Build GRCh38
rs966086428 Chr.7: 39404651-39404651 on Build GRCh38
rs1029323940 Chr.7: 39404654-39404654 on Build GRCh38
rs112436803 Chr.7: 39404795-39404796 on Build GRCh38
rs948342712 Chr.7: 39404672-39404672 on Build GRCh38
rs774054071 Chr.7: 39404562-39404562 on Build GRCh38
rs542217523 Chr.7: 39404725-39404725 on Build GRCh38
rs892848781 Chr.7: 39404631-39404631 on Build GRCh38

View Full Product Details