Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 41776811-41777357 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INHBA-AS1
Entrez Gene ID
285954
Gene Name
INHBA antisense RNA 1
Gene Aliases
-
UniGene
Hs.656869
Gene Chromosome Location
Chr.7: 41693916-41779378 on Build GRCh38
Transcripts
NR_027118.1

SNP Detail

SNP ID Chromosome Location
rs760325690 Chr.7: 41776937-41776937 on Build GRCh38
rs139826697 Chr.7: 41777245-41777245 on Build GRCh38
rs191224152 Chr.7: 41776840-41776840 on Build GRCh38
rs1043898086 Chr.7: 41777271-41777271 on Build GRCh38
rs1034029936 Chr.7: 41776962-41776962 on Build GRCh38
rs951834346 Chr.7: 41777113-41777113 on Build GRCh38
rs1040639368 Chr.7: 41776870-41776870 on Build GRCh38
rs986228415 Chr.7: 41777177-41777177 on Build GRCh38
rs979228706 Chr.7: 41776993-41776993 on Build GRCh38
rs913816160 Chr.7: 41777078-41777078 on Build GRCh38
rs904050012 Chr.7: 41777294-41777294 on Build GRCh38
rs1040884699 Chr.7: 41777092-41777092 on Build GRCh38
rs573909685 Chr.7: 41777205-41777205 on Build GRCh38
rs746983095 Chr.7: 41777143-41777143 on Build GRCh38
rs962262525 Chr.7: 41777001-41777001 on Build GRCh38
rs976486798 Chr.7: 41777215-41777215 on Build GRCh38
rs116174321 Chr.7: 41776938-41776938 on Build GRCh38
rs888209640 Chr.7: 41777210-41777210 on Build GRCh38
rs572708129 Chr.7: 41776977-41776977 on Build GRCh38
rs146452436 Chr.7: 41776920-41776920 on Build GRCh38
rs372040501 Chr.7: 41776946-41776946 on Build GRCh38
rs944676857 Chr.7: 41776859-41776859 on Build GRCh38
rs368345854 Chr.7: 41776880-41776880 on Build GRCh38
rs944727165 Chr.7: 41777201-41777201 on Build GRCh38
rs970103676 Chr.7: 41776979-41776979 on Build GRCh38
rs557889573 Chr.7: 41776976-41776976 on Build GRCh38
rs60570739 Chr.7: 41776957-41776957 on Build GRCh38
rs892674793 Chr.7: 41776912-41776912 on Build GRCh38
rs1015253102 Chr.7: 41777221-41777221 on Build GRCh38
rs575682693 Chr.7: 41777296-41777296 on Build GRCh38
rs934619853 Chr.7: 41777040-41777040 on Build GRCh38
rs187624830 Chr.7: 41777032-41777032 on Build GRCh38
rs183342687 Chr.7: 41776858-41776858 on Build GRCh38
rs567989431 Chr.7: 41776919-41776919 on Build GRCh38
rs1048579212 Chr.7: 41777127-41777127 on Build GRCh38
rs770302749 Chr.7: 41777281-41777281 on Build GRCh38
rs533823643 Chr.7: 41777007-41777007 on Build GRCh38
rs143144244 Chr.7: 41777327-41777327 on Build GRCh38
rs1014296050 Chr.7: 41776984-41776984 on Build GRCh38
rs985089799 Chr.7: 41776965-41776976 on Build GRCh38
rs190541496 Chr.7: 41777097-41777097 on Build GRCh38
rs539510773 Chr.7: 41776974-41776974 on Build GRCh38
rs910593335 Chr.7: 41777200-41777200 on Build GRCh38
rs1004279182 Chr.7: 41777216-41777216 on Build GRCh38
rs943957846 Chr.7: 41777081-41777081 on Build GRCh38
rs868286221 Chr.7: 41777305-41777305 on Build GRCh38
rs112012838 Chr.7: 41777317-41777317 on Build GRCh38
rs777786703 Chr.7: 41777114-41777114 on Build GRCh38
rs896053356 Chr.7: 41777258-41777258 on Build GRCh38
rs988837956 Chr.7: 41777047-41777047 on Build GRCh38

View Full Product Details