Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 41778336-41778855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INHBA-AS1
Entrez Gene ID
285954
Gene Name
INHBA antisense RNA 1
Gene Aliases
-
UniGene
Hs.656869
Gene Chromosome Location
Chr.7: 41693916-41779378 on Build GRCh38
Transcripts
NR_027118.1

SNP Detail

SNP ID Chromosome Location
rs887867281 Chr.7: 41778760-41778760 on Build GRCh38
rs1012208935 Chr.7: 41778536-41778536 on Build GRCh38
rs150436951 Chr.7: 41778403-41778403 on Build GRCh38
rs537458444 Chr.7: 41778527-41778527 on Build GRCh38
rs1040552771 Chr.7: 41778511-41778511 on Build GRCh38
rs566877863 Chr.7: 41778519-41778519 on Build GRCh38
rs994441534 Chr.7: 41778806-41778806 on Build GRCh38
rs397889084 Chr.7: 41778793-41778793 on Build GRCh38
rs1025688346 Chr.7: 41778539-41778539 on Build GRCh38
rs1053801147 Chr.7: 41778753-41778753 on Build GRCh38
rs942135241 Chr.7: 41778763-41778763 on Build GRCh38
rs912929869 Chr.7: 41778726-41778726 on Build GRCh38
rs1048785658 Chr.7: 41778432-41778432 on Build GRCh38
rs923520401 Chr.7: 41778739-41778739 on Build GRCh38
rs764993810 Chr.7: 41778547-41778547 on Build GRCh38
rs974426494 Chr.7: 41778609-41778609 on Build GRCh38
rs1016820351 Chr.7: 41778370-41778370 on Build GRCh38
rs538434049 Chr.7: 41778581-41778581 on Build GRCh38
rs572871208 Chr.7: 41778388-41778388 on Build GRCh38
rs565616164 Chr.7: 41778434-41778434 on Build GRCh38
rs1006216568 Chr.7: 41778576-41778576 on Build GRCh38
rs1042261772 Chr.7: 41778689-41778689 on Build GRCh38
rs184488383 Chr.7: 41778600-41778600 on Build GRCh38
rs138379676 Chr.7: 41778489-41778489 on Build GRCh38
rs979795560 Chr.7: 41778468-41778468 on Build GRCh38
rs769294208 Chr.7: 41778621-41778621 on Build GRCh38
rs542002937 Chr.7: 41778822-41778822 on Build GRCh38
rs192557380 Chr.7: 41778494-41778494 on Build GRCh38
rs959372957 Chr.7: 41778534-41778534 on Build GRCh38
rs989812780 Chr.7: 41778723-41778723 on Build GRCh38
rs548828781 Chr.7: 41778513-41778513 on Build GRCh38
rs574647352 Chr.7: 41778651-41778651 on Build GRCh38
rs188313525 Chr.7: 41778490-41778490 on Build GRCh38
rs1015379878 Chr.7: 41778591-41778591 on Build GRCh38
rs35119490 Chr.7: 41778665-41778666 on Build GRCh38
rs559181089 Chr.7: 41778550-41778550 on Build GRCh38
rs143895685 Chr.7: 41778563-41778563 on Build GRCh38
rs980348743 Chr.7: 41778664-41778664 on Build GRCh38
rs1036459339 Chr.7: 41778772-41778772 on Build GRCh38
rs1051068994 Chr.7: 41778759-41778759 on Build GRCh38
rs1034094153 Chr.7: 41778688-41778688 on Build GRCh38
rs976308371 Chr.7: 41778737-41778737 on Build GRCh38
rs146411874 Chr.7: 41778645-41778645 on Build GRCh38
rs971215328 Chr.7: 41778463-41778463 on Build GRCh38
rs946302918 Chr.7: 41778504-41778504 on Build GRCh38
rs566427428 Chr.7: 41778501-41778501 on Build GRCh38
rs945708982 Chr.7: 41778736-41778736 on Build GRCh38
rs971307580 Chr.7: 41778363-41778363 on Build GRCh38
rs901984511 Chr.7: 41778523-41778523 on Build GRCh38
rs906802643 Chr.7: 41778775-41778775 on Build GRCh38
rs542049184 Chr.7: 41778427-41778428 on Build GRCh38
rs929027495 Chr.7: 41778428-41778428 on Build GRCh38
rs932105496 Chr.7: 41778757-41778757 on Build GRCh38
rs972539181 Chr.7: 41778433-41778433 on Build GRCh38
rs1006639709 Chr.7: 41778367-41778367 on Build GRCh38
rs115948595 Chr.7: 41778503-41778503 on Build GRCh38
rs59293524 Chr.7: 41778594-41778594 on Build GRCh38
rs936473409 Chr.7: 41778741-41778741 on Build GRCh38
rs901755782 Chr.7: 41778473-41778473 on Build GRCh38
rs957387767 Chr.7: 41778695-41778695 on Build GRCh38

View Full Product Details