Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 71307580-71308124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR3914-1
Entrez Gene ID
100500836
Gene Name
microRNA 3914-1
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.7: 71307672-71307770 on Build GRCh38
Transcripts
NR_037477.1 NR_037479.1 NM_022479.2 XM_011516467.2 XM_011516469.2 XM_017012521.1

Gene Symbol
MIR3914-2
Entrez Gene ID
100500920
Gene Name
microRNA 3914-2
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.7: 71307674-71307768 on Build GRCh38
Transcripts
NR_037477.1 NR_037479.1 NM_022479.2 XM_011516467.2 XM_011516469.2 XM_017012521.1

Gene Symbol
WBSCR17
Entrez Gene ID
64409
Gene Name
Williams-Beuren syndrome chromosome region 17
Gene Aliases
GALNACT17, GALNT16, GALNT20, GALNTL3, GalNAc-T5L
UniGene
Hs.488591
Gene Chromosome Location
Chr.7: 71132414-71713601 on Build GRCh38
Transcripts
NR_037477.1 NR_037479.1 NM_022479.2 XM_011516467.2 XM_011516469.2 XM_017012521.1

SNP Detail

SNP ID Chromosome Location
rs936273948 Chr.7: 71307969-71307970 on Build GRCh38
rs558487816 Chr.7: 71307761-71307761 on Build GRCh38
rs1034615407 Chr.7: 71307704-71307704 on Build GRCh38
rs117732342 Chr.7: 71308062-71308062 on Build GRCh38
rs951195916 Chr.7: 71307800-71307800 on Build GRCh38
rs557475159 Chr.7: 71307617-71307617 on Build GRCh38
rs750269005 Chr.7: 71307650-71307650 on Build GRCh38
rs914641025 Chr.7: 71307776-71307776 on Build GRCh38
rs779218916 Chr.7: 71307941-71307941 on Build GRCh38
rs117009497 Chr.7: 71307646-71307646 on Build GRCh38
rs923808479 Chr.7: 71307830-71307830 on Build GRCh38
rs187134230 Chr.7: 71307834-71307834 on Build GRCh38
rs146957024 Chr.7: 71307989-71307989 on Build GRCh38
rs962515301 Chr.7: 71308069-71308069 on Build GRCh38
rs887521608 Chr.7: 71308043-71308043 on Build GRCh38
rs900427991 Chr.7: 71307988-71307988 on Build GRCh38
rs750978784 Chr.7: 71307645-71307645 on Build GRCh38
rs1013960081 Chr.7: 71307723-71307723 on Build GRCh38
rs529630849 Chr.7: 71307877-71307877 on Build GRCh38
rs182719960 Chr.7: 71308074-71308074 on Build GRCh38
rs990185838 Chr.7: 71307774-71307774 on Build GRCh38
rs568121348 Chr.7: 71307977-71307977 on Build GRCh38
rs543575625 Chr.7: 71307712-71307712 on Build GRCh38
rs1017160292 Chr.7: 71307824-71307824 on Build GRCh38
rs537045089 Chr.7: 71308041-71308041 on Build GRCh38
rs779821140 Chr.7: 71307903-71307903 on Build GRCh38
rs577138151 Chr.7: 71308063-71308063 on Build GRCh38
rs1004996313 Chr.7: 71307629-71307629 on Build GRCh38
rs962648585 Chr.7: 71307825-71307825 on Build GRCh38
rs566050010 Chr.7: 71307835-71307835 on Build GRCh38
rs554694480 Chr.7: 71307887-71307887 on Build GRCh38
rs866134623 Chr.7: 71307828-71307828 on Build GRCh38
rs753994333 Chr.7: 71307836-71307836 on Build GRCh38
rs944556870 Chr.7: 71307843-71307843 on Build GRCh38
rs1033987249 Chr.7: 71307667-71307667 on Build GRCh38
rs1013908246 Chr.7: 71308011-71308011 on Build GRCh38
rs982062606 Chr.7: 71307697-71307697 on Build GRCh38
rs912016084 Chr.7: 71307889-71307889 on Build GRCh38
rs4520056 Chr.7: 71307815-71307815 on Build GRCh38
rs541455394 Chr.7: 71307947-71307947 on Build GRCh38
rs1034992522 Chr.7: 71308092-71308092 on Build GRCh38
rs1003056900 Chr.7: 71308085-71308085 on Build GRCh38
rs191566245 Chr.7: 71307868-71307868 on Build GRCh38
rs946123595 Chr.7: 71307926-71307926 on Build GRCh38
rs879479861 Chr.7: 71307845-71307845 on Build GRCh38
rs971620934 Chr.7: 71307682-71307682 on Build GRCh38
rs894120558 Chr.7: 71307694-71307694 on Build GRCh38
rs977911064 Chr.7: 71307818-71307818 on Build GRCh38

View Full Product Details