Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 29725951-29726475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00589
Entrez Gene ID
619351
Gene Name
long intergenic non-protein coding RNA 589
Gene Aliases
C8orf75
UniGene
Hs.566911
Gene Chromosome Location
Chr.8: 29721259-29748109 on Build GRCh38
Transcripts
NR_026765.1

SNP Detail

SNP ID Chromosome Location
rs751799162 Chr.8: 29726378-29726378 on Build GRCh38
rs186418442 Chr.8: 29726257-29726257 on Build GRCh38
rs1030942515 Chr.8: 29725983-29725983 on Build GRCh38
rs28439654 Chr.8: 29726438-29726438 on Build GRCh38
rs374531555 Chr.8: 29726198-29726198 on Build GRCh38
rs1024065425 Chr.8: 29726382-29726382 on Build GRCh38
rs776270495 Chr.8: 29726020-29726020 on Build GRCh38
rs138446040 Chr.8: 29726374-29726374 on Build GRCh38
rs188873563 Chr.8: 29726144-29726144 on Build GRCh38
rs981891945 Chr.8: 29726049-29726049 on Build GRCh38
rs1018924514 Chr.8: 29726094-29726094 on Build GRCh38
rs919179428 Chr.8: 29726428-29726428 on Build GRCh38
rs544206503 Chr.8: 29726332-29726332 on Build GRCh38
rs777912079 Chr.8: 29726320-29726320 on Build GRCh38
rs761064224 Chr.8: 29726064-29726064 on Build GRCh38
rs28415347 Chr.8: 29726284-29726284 on Build GRCh38
rs969093369 Chr.8: 29726317-29726317 on Build GRCh38
rs754193024 Chr.8: 29726231-29726231 on Build GRCh38
rs150123816 Chr.8: 29726297-29726297 on Build GRCh38
rs984347192 Chr.8: 29726377-29726377 on Build GRCh38
rs1040449152 Chr.8: 29726289-29726289 on Build GRCh38
rs755714996 Chr.8: 29726353-29726354 on Build GRCh38
rs918796915 Chr.8: 29726333-29726333 on Build GRCh38
rs564822273 Chr.8: 29726315-29726315 on Build GRCh38
rs181762409 Chr.8: 29726239-29726239 on Build GRCh38
rs117335302 Chr.8: 29726234-29726234 on Build GRCh38
rs962128128 Chr.8: 29726330-29726330 on Build GRCh38
rs918823550 Chr.8: 29726082-29726082 on Build GRCh38
rs971019018 Chr.8: 29726388-29726388 on Build GRCh38
rs189607927 Chr.8: 29726324-29726324 on Build GRCh38
rs546748273 Chr.8: 29725974-29725974 on Build GRCh38
rs889179582 Chr.8: 29726279-29726279 on Build GRCh38
rs566564810 Chr.8: 29725977-29725977 on Build GRCh38
rs897636747 Chr.8: 29726034-29726034 on Build GRCh38
rs765720492 Chr.8: 29726263-29726263 on Build GRCh38
rs942736694 Chr.8: 29726386-29726386 on Build GRCh38
rs78879480 Chr.8: 29726009-29726009 on Build GRCh38
rs530065306 Chr.8: 29726107-29726107 on Build GRCh38
rs149251281 Chr.8: 29725995-29725995 on Build GRCh38
rs530932797 Chr.8: 29726165-29726166 on Build GRCh38
rs752018040 Chr.8: 29726349-29726352 on Build GRCh38
rs1021642886 Chr.8: 29726110-29726110 on Build GRCh38
rs180791442 Chr.8: 29726338-29726338 on Build GRCh38
rs991141733 Chr.8: 29726196-29726196 on Build GRCh38
rs114201556 Chr.8: 29726038-29726038 on Build GRCh38
rs1007513241 Chr.8: 29726069-29726069 on Build GRCh38
rs1037183230 Chr.8: 29726065-29726065 on Build GRCh38
rs1036296443 Chr.8: 29726027-29726027 on Build GRCh38
rs972174587 Chr.8: 29726422-29726422 on Build GRCh38
rs1041149676 Chr.8: 29726225-29726225 on Build GRCh38
rs928765552 Chr.8: 29726061-29726061 on Build GRCh38
rs1048381590 Chr.8: 29726119-29726119 on Build GRCh38
rs957965264 Chr.8: 29726004-29726004 on Build GRCh38
rs537038285 Chr.8: 29726194-29726194 on Build GRCh38
rs766796455 Chr.8: 29726170-29726170 on Build GRCh38
rs968203153 Chr.8: 29726141-29726141 on Build GRCh38
rs902090295 Chr.8: 29726105-29726105 on Build GRCh38
rs930486893 Chr.8: 29726255-29726255 on Build GRCh38
rs758473705 Chr.8: 29726298-29726298 on Build GRCh38
rs916288518 Chr.8: 29726026-29726026 on Build GRCh38

View Full Product Details