Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 64575330-64575835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC401463
Entrez Gene ID
401463
Gene Name
uncharacterized LOC401463
Gene Aliases
-
UniGene
Hs.651960
Gene Chromosome Location
Chr.8: 64574308-64577263 on Build GRCh38
Transcripts
NR_015374.1

SNP Detail

SNP ID Chromosome Location
rs1015404785 Chr.8: 64575707-64575707 on Build GRCh38
rs547730802 Chr.8: 64575655-64575655 on Build GRCh38
rs567540324 Chr.8: 64575372-64575372 on Build GRCh38
rs527787897 Chr.8: 64575503-64575503 on Build GRCh38
rs912328186 Chr.8: 64575599-64575599 on Build GRCh38
rs973955335 Chr.8: 64575786-64575786 on Build GRCh38
rs537101109 Chr.8: 64575801-64575801 on Build GRCh38
rs956452786 Chr.8: 64575363-64575363 on Build GRCh38
rs968132607 Chr.8: 64575380-64575380 on Build GRCh38
rs886985307 Chr.8: 64575683-64575683 on Build GRCh38
rs540826740 Chr.8: 64575629-64575629 on Build GRCh38
rs182842433 Chr.8: 64575558-64575558 on Build GRCh38
rs907311644 Chr.8: 64575730-64575730 on Build GRCh38
rs948709987 Chr.8: 64575510-64575510 on Build GRCh38
rs907456662 Chr.8: 64575577-64575577 on Build GRCh38
rs1028028598 Chr.8: 64575667-64575667 on Build GRCh38
rs760685495 Chr.8: 64575366-64575366 on Build GRCh38
rs1009274319 Chr.8: 64575364-64575364 on Build GRCh38
rs780275543 Chr.8: 64575802-64575802 on Build GRCh38
rs750587861 Chr.8: 64575513-64575513 on Build GRCh38
rs375430427 Chr.8: 64575460-64575460 on Build GRCh38
rs570796421 Chr.8: 64575752-64575752 on Build GRCh38
rs537783743 Chr.8: 64575422-64575422 on Build GRCh38
rs946204592 Chr.8: 64575511-64575511 on Build GRCh38
rs1034748644 Chr.8: 64575438-64575438 on Build GRCh38
rs1022483137 Chr.8: 64575634-64575634 on Build GRCh38
rs903626800 Chr.8: 64575641-64575641 on Build GRCh38
rs899482763 Chr.8: 64575628-64575628 on Build GRCh38
rs1014971748 Chr.8: 64575430-64575430 on Build GRCh38
rs1039394931 Chr.8: 64575610-64575610 on Build GRCh38
rs1024732159 Chr.8: 64575713-64575713 on Build GRCh38
rs753902301 Chr.8: 64575437-64575437 on Build GRCh38
rs996904580 Chr.8: 64575662-64575662 on Build GRCh38
rs886941154 Chr.8: 64575409-64575409 on Build GRCh38
rs919913384 Chr.8: 64575508-64575508 on Build GRCh38
rs1004007719 Chr.8: 64575688-64575688 on Build GRCh38
rs1053178166 Chr.8: 64575600-64575600 on Build GRCh38
rs1009290940 Chr.8: 64575627-64575627 on Build GRCh38
rs553862784 Chr.8: 64575587-64575587 on Build GRCh38
rs891971571 Chr.8: 64575604-64575604 on Build GRCh38
rs990803381 Chr.8: 64575498-64575498 on Build GRCh38
rs943762074 Chr.8: 64575609-64575609 on Build GRCh38
rs62519833 Chr.8: 64575432-64575432 on Build GRCh38
rs977546455 Chr.8: 64575563-64575563 on Build GRCh38
rs149249490 Chr.8: 64575353-64575353 on Build GRCh38
rs751736548 Chr.8: 64575476-64575478 on Build GRCh38
rs1044429994 Chr.8: 64575561-64575561 on Build GRCh38
rs959008777 Chr.8: 64575447-64575447 on Build GRCh38
rs757312659 Chr.8: 64575453-64575453 on Build GRCh38
rs572175610 Chr.8: 64575590-64575590 on Build GRCh38
rs370427166 Chr.8: 64575676-64575676 on Build GRCh38

View Full Product Details