Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 64577098-64577600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC401463
Entrez Gene ID
401463
Gene Name
uncharacterized LOC401463
Gene Aliases
-
UniGene
Hs.651960
Gene Chromosome Location
Chr.8: 64574308-64577263 on Build GRCh38
Transcripts
NR_015374.1

SNP Detail

SNP ID Chromosome Location
rs140077315 Chr.8: 64577164-64577164 on Build GRCh38
rs532428373 Chr.8: 64577576-64577576 on Build GRCh38
rs543243509 Chr.8: 64577521-64577521 on Build GRCh38
rs941598703 Chr.8: 64577190-64577190 on Build GRCh38
rs554065566 Chr.8: 64577335-64577335 on Build GRCh38
rs1056219283 Chr.8: 64577324-64577324 on Build GRCh38
rs945992804 Chr.8: 64577344-64577344 on Build GRCh38
rs992480138 Chr.8: 64577201-64577201 on Build GRCh38
rs1034046181 Chr.8: 64577552-64577552 on Build GRCh38
rs910154001 Chr.8: 64577189-64577189 on Build GRCh38
rs918794497 Chr.8: 64577577-64577577 on Build GRCh38
rs758533917 Chr.8: 64577376-64577376 on Build GRCh38
rs1037243587 Chr.8: 64577531-64577531 on Build GRCh38
rs1050518467 Chr.8: 64577384-64577384 on Build GRCh38
rs538050344 Chr.8: 64577178-64577178 on Build GRCh38
rs74935600 Chr.8: 64577448-64577448 on Build GRCh38
rs1021943126 Chr.8: 64577382-64577382 on Build GRCh38
rs909874496 Chr.8: 64577526-64577526 on Build GRCh38
rs565111238 Chr.8: 64577540-64577540 on Build GRCh38
rs934767794 Chr.8: 64577137-64577137 on Build GRCh38
rs768459772 Chr.8: 64577124-64577124 on Build GRCh38
rs897322679 Chr.8: 64577199-64577199 on Build GRCh38
rs941650893 Chr.8: 64577528-64577528 on Build GRCh38
rs889227918 Chr.8: 64577401-64577401 on Build GRCh38
rs528441910 Chr.8: 64577171-64577171 on Build GRCh38
rs576528199 Chr.8: 64577518-64577518 on Build GRCh38
rs965910499 Chr.8: 64577491-64577491 on Build GRCh38
rs1037191147 Chr.8: 64577192-64577192 on Build GRCh38
rs999427974 Chr.8: 64577447-64577447 on Build GRCh38
rs976288556 Chr.8: 64577514-64577514 on Build GRCh38
rs574046663 Chr.8: 64577446-64577446 on Build GRCh38
rs750580686 Chr.8: 64577295-64577295 on Build GRCh38
rs556754456 Chr.8: 64577206-64577206 on Build GRCh38
rs923749492 Chr.8: 64577135-64577135 on Build GRCh38
rs538974614 Chr.8: 64577211-64577211 on Build GRCh38
rs188297105 Chr.8: 64577158-64577158 on Build GRCh38
rs577898271 Chr.8: 64577210-64577210 on Build GRCh38
rs1019190024 Chr.8: 64577145-64577145 on Build GRCh38
rs75714505 Chr.8: 64577420-64577420 on Build GRCh38
rs972251195 Chr.8: 64577183-64577183 on Build GRCh38
rs1041511323 Chr.8: 64577356-64577356 on Build GRCh38
rs985450462 Chr.8: 64577524-64577524 on Build GRCh38
rs547946115 Chr.8: 64577128-64577128 on Build GRCh38
rs1050164036 Chr.8: 64577187-64577187 on Build GRCh38
rs1839878 Chr.8: 64577515-64577515 on Build GRCh38
rs1011927361 Chr.8: 64577350-64577350 on Build GRCh38
rs932678887 Chr.8: 64577185-64577185 on Build GRCh38
rs34651409 Chr.8: 64577426-64577426 on Build GRCh38
rs547689350 Chr.8: 64577578-64577578 on Build GRCh38
rs976852440 Chr.8: 64577147-64577147 on Build GRCh38

View Full Product Details