Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128244609-128245100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNM1
Entrez Gene ID
1759
Gene Name
dynamin 1
Gene Aliases
DNM, EIEE31
UniGene
Hs.522413
Gene Chromosome Location
Chr.9: 128203355-128255248 on Build GRCh38
Transcripts
NM_001005336.2 NM_001288737.1 NM_001288738.1 NM_001288739.1 NM_004408.3 XM_005251763.2 XM_005251768.2 XM_005251769.2 XM_006716992.2 XM_011518335.2 XM_011518336.2 XM_017014369.1 XM_017014370.1 XM_017014371.1 XM_017014373.1 NR_029619.1 NR_036110.1

Gene Symbol
MIR199B
Entrez Gene ID
406978
Gene Name
microRNA 199b
Gene Aliases
MIRN199B, mir-199b
UniGene
Gene Chromosome Location
Chr.9: 128244721-128244830 on Build GRCh38
Transcripts
NM_001005336.2 NM_001288737.1 NM_001288738.1 NM_001288739.1 NM_004408.3 XM_005251763.2 XM_005251768.2 XM_005251769.2 XM_006716992.2 XM_011518335.2 XM_011518336.2 XM_017014369.1 XM_017014370.1 XM_017014371.1 XM_017014373.1 NR_029619.1 NR_036110.1

Gene Symbol
MIR3154
Entrez Gene ID
100422893
Gene Name
microRNA 3154
Gene Aliases
mir-3154
UniGene
Gene Chromosome Location
Chr.9: 128244947-128245030 on Build GRCh38
Transcripts
NM_001005336.2 NM_001288737.1 NM_001288738.1 NM_001288739.1 NM_004408.3 XM_005251763.2 XM_005251768.2 XM_005251769.2 XM_006716992.2 XM_011518335.2 XM_011518336.2 XM_017014369.1 XM_017014370.1 XM_017014371.1 XM_017014373.1 NR_029619.1 NR_036110.1

SNP Detail

SNP ID Chromosome Location
rs750371544 Chr.9: 128244897-128244898 on Build GRCh38
rs957378714 Chr.9: 128245013-128245013 on Build GRCh38
rs567313721 Chr.9: 128244892-128244892 on Build GRCh38
rs760941457 Chr.9: 128244716-128244716 on Build GRCh38
rs570941210 Chr.9: 128244858-128244858 on Build GRCh38
rs540586378 Chr.9: 128245056-128245056 on Build GRCh38
rs1024465078 Chr.9: 128244786-128244786 on Build GRCh38
rs944624161 Chr.9: 128244891-128244891 on Build GRCh38
rs766974849 Chr.9: 128244833-128244833 on Build GRCh38
rs759856282 Chr.9: 128244718-128244718 on Build GRCh38
rs770940322 Chr.9: 128244867-128244867 on Build GRCh38
rs72631835 Chr.9: 128244830-128244830 on Build GRCh38
rs929073576 Chr.9: 128244901-128244901 on Build GRCh38
rs560606389 Chr.9: 128244725-128244725 on Build GRCh38
rs765755768 Chr.9: 128244719-128244719 on Build GRCh38
rs772118665 Chr.9: 128244713-128244713 on Build GRCh38
rs769225381 Chr.9: 128244905-128244905 on Build GRCh38
rs375288713 Chr.9: 128244814-128244814 on Build GRCh38
rs531926516 Chr.9: 128244834-128244834 on Build GRCh38
rs776787360 Chr.9: 128244810-128244810 on Build GRCh38
rs754882632 Chr.9: 128244859-128244859 on Build GRCh38
rs867251736 Chr.9: 128244740-128244740 on Build GRCh38
rs966395496 Chr.9: 128245065-128245065 on Build GRCh38
rs751386840 Chr.9: 128245047-128245047 on Build GRCh38
rs773377969 Chr.9: 128244714-128244714 on Build GRCh38
rs1024936275 Chr.9: 128244974-128244974 on Build GRCh38
rs780925686 Chr.9: 128244836-128244836 on Build GRCh38
rs774660548 Chr.9: 128244908-128244908 on Build GRCh38
rs758327834 Chr.9: 128244864-128244864 on Build GRCh38
rs1052574238 Chr.9: 128244788-128244788 on Build GRCh38
rs529476261 Chr.9: 128244773-128244773 on Build GRCh38
rs564511926 Chr.9: 128244658-128244658 on Build GRCh38
rs993242253 Chr.9: 128244675-128244675 on Build GRCh38
rs1056064977 Chr.9: 128244950-128244950 on Build GRCh38
rs919092839 Chr.9: 128245072-128245072 on Build GRCh38
rs777889185 Chr.9: 128244784-128244784 on Build GRCh38
rs768742034 Chr.9: 128244687-128244687 on Build GRCh38
rs779810582 Chr.9: 128244743-128244743 on Build GRCh38
rs370675238 Chr.9: 128245048-128245048 on Build GRCh38
rs770192040 Chr.9: 128244898-128244898 on Build GRCh38
rs773954082 Chr.9: 128244652-128244652 on Build GRCh38
rs113655888 Chr.9: 128244660-128244660 on Build GRCh38
rs755759262 Chr.9: 128244735-128244735 on Build GRCh38
rs1042106985 Chr.9: 128245064-128245064 on Build GRCh38
rs878943628 Chr.9: 128244856-128244856 on Build GRCh38
rs536061513 Chr.9: 128244897-128244897 on Build GRCh38
rs767215036 Chr.9: 128244723-128244723 on Build GRCh38
rs776488161 Chr.9: 128244945-128244945 on Build GRCh38
rs759245856 Chr.9: 128244952-128244952 on Build GRCh38
rs747136162 Chr.9: 128244866-128244866 on Build GRCh38
rs776951533 Chr.9: 128244717-128244717 on Build GRCh38
rs558869242 Chr.9: 128245004-128245004 on Build GRCh38
rs182137771 Chr.9: 128244860-128244860 on Build GRCh38
rs758081834 Chr.9: 128244988-128244988 on Build GRCh38
rs887445206 Chr.9: 128244943-128244943 on Build GRCh38
rs551643583 Chr.9: 128244839-128244839 on Build GRCh38
rs576296873 Chr.9: 128244910-128244910 on Build GRCh38
rs146892675 Chr.9: 128244722-128244722 on Build GRCh38
rs545354223 Chr.9: 128244966-128244966 on Build GRCh38
rs577976262 Chr.9: 128245053-128245053 on Build GRCh38
rs765978439 Chr.9: 128244944-128244944 on Build GRCh38
rs774379099 Chr.9: 128244693-128244693 on Build GRCh38
rs781398039 Chr.9: 128244875-128244875 on Build GRCh38
rs549122310 Chr.9: 128244808-128244808 on Build GRCh38
rs766140622 Chr.9: 128244852-128244852 on Build GRCh38
rs929084890 Chr.9: 128245075-128245075 on Build GRCh38
rs1053045749 Chr.9: 128244911-128244911 on Build GRCh38
rs752571525 Chr.9: 128244985-128244985 on Build GRCh38
rs763185585 Chr.9: 128244900-128244900 on Build GRCh38
rs756562959 Chr.9: 128244744-128244744 on Build GRCh38
rs373272982 Chr.9: 128244881-128244881 on Build GRCh38
rs771218946 Chr.9: 128244809-128244809 on Build GRCh38
rs768626729 Chr.9: 128244781-128244781 on Build GRCh38
rs560389192 Chr.9: 128245070-128245070 on Build GRCh38
rs537212527 Chr.9: 128244670-128244670 on Build GRCh38
rs896319242 Chr.9: 128244728-128244728 on Build GRCh38
rs754759597 Chr.9: 128244772-128244772 on Build GRCh38
rs892551318 Chr.9: 128244803-128244803 on Build GRCh38
rs763494265 Chr.9: 128244831-128244831 on Build GRCh38
rs368873525 Chr.9: 128244695-128244695 on Build GRCh38
rs894750031 Chr.9: 128244963-128244963 on Build GRCh38
rs556444770 Chr.9: 128244904-128244904 on Build GRCh38
rs753705566 Chr.9: 128244754-128244754 on Build GRCh38
rs529339788 Chr.9: 128245078-128245078 on Build GRCh38
rs749275916 Chr.9: 128244672-128244672 on Build GRCh38

View Full Product Details