Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130578953-130579446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100272217
Entrez Gene ID
100272217
Gene Name
uncharacterized LOC100272217
Gene Aliases
-
UniGene
Hs.601255
Gene Chromosome Location
Chr.9: 130577350-130579494 on Build GRCh38
Transcripts
NR_027440.1

SNP Detail

SNP ID Chromosome Location
rs541465228 Chr.9: 130579275-130579275 on Build GRCh38
rs909341447 Chr.9: 130579360-130579360 on Build GRCh38
rs913248313 Chr.9: 130579312-130579313 on Build GRCh38
rs923895755 Chr.9: 130579414-130579414 on Build GRCh38
rs764331343 Chr.9: 130579181-130579181 on Build GRCh38
rs1054036024 Chr.9: 130579423-130579423 on Build GRCh38
rs992091497 Chr.9: 130579079-130579079 on Build GRCh38
rs922185197 Chr.9: 130579363-130579363 on Build GRCh38
rs893611929 Chr.9: 130579424-130579424 on Build GRCh38
rs983625218 Chr.9: 130578993-130578993 on Build GRCh38
rs999801984 Chr.9: 130579155-130579155 on Build GRCh38
rs997452629 Chr.9: 130579272-130579272 on Build GRCh38
rs56154996 Chr.9: 130579197-130579197 on Build GRCh38
rs916267047 Chr.9: 130579397-130579397 on Build GRCh38
rs1031029291 Chr.9: 130579310-130579310 on Build GRCh38
rs964659453 Chr.9: 130579265-130579265 on Build GRCh38
rs902416656 Chr.9: 130579147-130579147 on Build GRCh38
rs1011635925 Chr.9: 130579427-130579427 on Build GRCh38
rs758583357 Chr.9: 130579141-130579141 on Build GRCh38
rs932176169 Chr.9: 130579392-130579392 on Build GRCh38
rs770575565 Chr.9: 130578990-130578990 on Build GRCh38
rs949062878 Chr.9: 130579401-130579401 on Build GRCh38
rs1005006828 Chr.9: 130579025-130579025 on Build GRCh38
rs1011583284 Chr.9: 130579232-130579232 on Build GRCh38
rs925071923 Chr.9: 130579161-130579161 on Build GRCh38
rs941756928 Chr.9: 130579014-130579014 on Build GRCh38
rs1017572896 Chr.9: 130579236-130579236 on Build GRCh38
rs916213372 Chr.9: 130579051-130579051 on Build GRCh38
rs1027857934 Chr.9: 130578990-130578990 on Build GRCh38
rs990538031 Chr.9: 130579383-130579383 on Build GRCh38
rs149315717 Chr.9: 130579062-130579063 on Build GRCh38
rs752819642 Chr.9: 130579128-130579128 on Build GRCh38
rs766855495 Chr.9: 130579031-130579031 on Build GRCh38
rs893215575 Chr.9: 130579186-130579186 on Build GRCh38
rs1046059925 Chr.9: 130579413-130579413 on Build GRCh38
rs984064681 Chr.9: 130579333-130579333 on Build GRCh38
rs373064175 Chr.9: 130579400-130579400 on Build GRCh38
rs777011352 Chr.9: 130579032-130579032 on Build GRCh38
rs369506774 Chr.9: 130579274-130579274 on Build GRCh38
rs908989404 Chr.9: 130578998-130578998 on Build GRCh38
rs963129544 Chr.9: 130579366-130579366 on Build GRCh38
rs950989089 Chr.9: 130579323-130579323 on Build GRCh38
rs944696647 Chr.9: 130579316-130579316 on Build GRCh38
rs542893728 Chr.9: 130579058-130579058 on Build GRCh38
rs752519624 Chr.9: 130579418-130579418 on Build GRCh38
rs4740351 Chr.9: 130579102-130579102 on Build GRCh38

View Full Product Details