Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 111520155-111520684 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00890
Entrez Gene ID
401613
Gene Name
long intergenic non-protein coding RNA 890
Gene Aliases
DKFZp686D0853
UniGene
Hs.98581
Gene Chromosome Location
Chr.X: 111511662-111522399 on Build GRCh38
Transcripts
NR_033974.1

SNP Detail

SNP ID Chromosome Location
rs976390760 Chr.X: 111520361-111520361 on Build GRCh38
rs1003232662 Chr.X: 111520491-111520491 on Build GRCh38
rs773489738 Chr.X: 111520417-111520417 on Build GRCh38
rs774073080 Chr.X: 111520398-111520398 on Build GRCh38
rs779440890 Chr.X: 111520181-111520182 on Build GRCh38
rs768510076 Chr.X: 111520278-111520278 on Build GRCh38
rs1010703041 Chr.X: 111520395-111520395 on Build GRCh38
rs989212800 Chr.X: 111520379-111520379 on Build GRCh38
rs747990368 Chr.X: 111520392-111520392 on Build GRCh38
rs964982325 Chr.X: 111520351-111520351 on Build GRCh38
rs906450583 Chr.X: 111520457-111520457 on Build GRCh38
rs950939027 Chr.X: 111520223-111520223 on Build GRCh38
rs1049185012 Chr.X: 111520651-111520651 on Build GRCh38
rs922008319 Chr.X: 111520184-111520184 on Build GRCh38
rs1055143679 Chr.X: 111520556-111520556 on Build GRCh38
rs1042663191 Chr.X: 111520437-111520437 on Build GRCh38
rs749196795 Chr.X: 111520268-111520268 on Build GRCh38
rs768606061 Chr.X: 111520328-111520328 on Build GRCh38
rs939311873 Chr.X: 111520499-111520499 on Build GRCh38
rs932019919 Chr.X: 111520193-111520193 on Build GRCh38
rs994906872 Chr.X: 111520642-111520642 on Build GRCh38
rs778741480 Chr.X: 111520371-111520371 on Build GRCh38
rs772272776 Chr.X: 111520397-111520397 on Build GRCh38
rs182330276 Chr.X: 111520424-111520424 on Build GRCh38
rs895179912 Chr.X: 111520586-111520586 on Build GRCh38
rs935261570 Chr.X: 111520355-111520355 on Build GRCh38
rs1051714286 Chr.X: 111520216-111520216 on Build GRCh38
rs920983718 Chr.X: 111520367-111520367 on Build GRCh38

View Full Product Details