Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 111522364-111522680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00890
Entrez Gene ID
401613
Gene Name
long intergenic non-protein coding RNA 890
Gene Aliases
DKFZp686D0853
UniGene
Hs.98581
Gene Chromosome Location
Chr.X: 111511662-111522399 on Build GRCh38
Transcripts
NR_033974.1

SNP Detail

SNP ID Chromosome Location
rs960596770 Chr.X: 111522607-111522607 on Build GRCh38
rs746853952 Chr.X: 111522611-111522611 on Build GRCh38
rs753378985 Chr.X: 111522403-111522403 on Build GRCh38
rs1041941500 Chr.X: 111522628-111522628 on Build GRCh38
rs765999727 Chr.X: 111522402-111522402 on Build GRCh38
rs952010505 Chr.X: 111522469-111522469 on Build GRCh38
rs747928561 Chr.X: 111522468-111522468 on Build GRCh38
rs1033635893 Chr.X: 111522406-111522406 on Build GRCh38
rs190527054 Chr.X: 111522453-111522453 on Build GRCh38
rs986204167 Chr.X: 111522591-111522591 on Build GRCh38
rs770706793 Chr.X: 111522616-111522616 on Build GRCh38
rs932423 Chr.X: 111522582-111522582 on Build GRCh38
rs758354948 Chr.X: 111522521-111522521 on Build GRCh38
rs371099489 Chr.X: 111522454-111522454 on Build GRCh38
rs976071418 Chr.X: 111522505-111522505 on Build GRCh38
rs752574181 Chr.X: 111522500-111522500 on Build GRCh38
rs1056155833 Chr.X: 111522646-111522646 on Build GRCh38
rs910982477 Chr.X: 111522416-111522416 on Build GRCh38
rs924860629 Chr.X: 111522637-111522637 on Build GRCh38
rs781335336 Chr.X: 111522649-111522649 on Build GRCh38
rs914745056 Chr.X: 111522603-111522603 on Build GRCh38
rs145500581 Chr.X: 111522643-111522643 on Build GRCh38
rs999605613 Chr.X: 111522397-111522397 on Build GRCh38
rs1015263605 Chr.X: 111522490-111522490 on Build GRCh38
rs962029625 Chr.X: 111522496-111522496 on Build GRCh38
rs181645718 Chr.X: 111522487-111522487 on Build GRCh38

View Full Product Details