Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 12379057-12379601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM66A
Entrez Gene ID
100133172
Gene Name
family with sequence similarity 66 member A
Gene Aliases
-
UniGene
Hs.721640
Gene Chromosome Location
Chr.8: 12362019-12411001 on Build GRCh38
Transcripts
NR_026789.1 NR_046415.1

Gene Symbol
LOC649352
Entrez Gene ID
649352
Gene Name
ubiquitin carboxyl-terminal hydrolase 17-like protein 2-like
Gene Aliases
-
UniGene
Hs.742474
Gene Chromosome Location
Chr.8: 12379267-12379789 on Build GRCh38
Transcripts
NR_026789.1 NR_046415.1

SNP Detail

SNP ID Chromosome Location
rs2739961 Chr.8: 12379535-12379535 on Build GRCh38
rs546219921 Chr.8: 12379370-12379370 on Build GRCh38
rs117580162 Chr.8: 12379243-12379243 on Build GRCh38
rs535866804 Chr.8: 12379286-12379286 on Build GRCh38
rs199994439 Chr.8: 12379077-12379077 on Build GRCh38
rs202203027 Chr.8: 12379271-12379271 on Build GRCh38
rs201889255 Chr.8: 12379230-12379230 on Build GRCh38
rs569501390 Chr.8: 12379272-12379272 on Build GRCh38
rs1007810951 Chr.8: 12379486-12379486 on Build GRCh38
rs781491187 Chr.8: 12379412-12379412 on Build GRCh38
rs9694631 Chr.8: 12379143-12379143 on Build GRCh38
rs369606820 Chr.8: 12379164-12379164 on Build GRCh38
rs530448202 Chr.8: 12379198-12379198 on Build GRCh38
rs201717135 Chr.8: 12379565-12379565 on Build GRCh38
rs534674499 Chr.8: 12379574-12379574 on Build GRCh38
rs560089059 Chr.8: 12379189-12379189 on Build GRCh38
rs563501799 Chr.8: 12379204-12379204 on Build GRCh38
rs1042408505 Chr.8: 12379468-12379468 on Build GRCh38
rs929076745 Chr.8: 12379148-12379148 on Build GRCh38
rs112536584 Chr.8: 12379467-12379467 on Build GRCh38
rs189809861 Chr.8: 12379082-12379082 on Build GRCh38
rs199538762 Chr.8: 12379579-12379579 on Build GRCh38
rs558621041 Chr.8: 12379087-12379087 on Build GRCh38
rs9693028 Chr.8: 12379346-12379346 on Build GRCh38
rs918920870 Chr.8: 12379112-12379112 on Build GRCh38
rs552343123 Chr.8: 12379206-12379206 on Build GRCh38
rs201837560 Chr.8: 12379483-12379483 on Build GRCh38
rs377704278 Chr.8: 12379395-12379395 on Build GRCh38
rs537057709 Chr.8: 12379273-12379273 on Build GRCh38
rs570441022 Chr.8: 12379561-12379561 on Build GRCh38
rs998248034 Chr.8: 12379471-12379471 on Build GRCh38
rs184917892 Chr.8: 12379079-12379079 on Build GRCh38
rs201925797 Chr.8: 12379320-12379320 on Build GRCh38
rs535211199 Chr.8: 12379307-12379307 on Build GRCh38
rs200694502 Chr.8: 12379083-12379083 on Build GRCh38
rs144311455 Chr.8: 12379322-12379322 on Build GRCh38
rs199796996 Chr.8: 12379287-12379287 on Build GRCh38
rs1017396777 Chr.8: 12379497-12379497 on Build GRCh38
rs71521627 Chr.8: 12379520-12379520 on Build GRCh38
rs529840468 Chr.8: 12379443-12379443 on Build GRCh38
rs181070339 Chr.8: 12379208-12379208 on Build GRCh38
rs576920484 Chr.8: 12379290-12379290 on Build GRCh38
rs13273613 Chr.8: 12379426-12379426 on Build GRCh38
rs200889251 Chr.8: 12379269-12379269 on Build GRCh38
rs963327398 Chr.8: 12379513-12379513 on Build GRCh38
rs77190750 Chr.8: 12379465-12379465 on Build GRCh38
rs530958185 Chr.8: 12379205-12379205 on Build GRCh38
rs9694401 Chr.8: 12379317-12379317 on Build GRCh38
rs564401242 Chr.8: 12379383-12379383 on Build GRCh38
rs35728551 Chr.8: 12379518-12379518 on Build GRCh38
rs377075699 Chr.8: 12379328-12379328 on Build GRCh38
rs542231589 Chr.8: 12379199-12379199 on Build GRCh38
rs200083121 Chr.8: 12379274-12379274 on Build GRCh38
rs894057736 Chr.8: 12379330-12379330 on Build GRCh38
rs78090203 Chr.8: 12379161-12379161 on Build GRCh38
rs546704821 Chr.8: 12379423-12379423 on Build GRCh38
rs973310598 Chr.8: 12379519-12379519 on Build GRCh38
rs756582497 Chr.8: 12379479-12379479 on Build GRCh38
rs62496734 Chr.8: 12379209-12379209 on Build GRCh38
rs13280704 Chr.8: 12379520-12379520 on Build GRCh38
rs373710762 Chr.8: 12379430-12379430 on Build GRCh38
rs189565892 Chr.8: 12379224-12379224 on Build GRCh38
rs528596394 Chr.8: 12379404-12379404 on Build GRCh38
rs918956823 Chr.8: 12379532-12379532 on Build GRCh38
rs1046561729 Chr.8: 12379239-12379239 on Build GRCh38
rs186106089 Chr.8: 12379217-12379217 on Build GRCh38
rs928179756 Chr.8: 12379251-12379251 on Build GRCh38
rs181926183 Chr.8: 12379323-12379323 on Build GRCh38
rs536105958 Chr.8: 12379237-12379237 on Build GRCh38
rs541684231 Chr.8: 12379172-12379172 on Build GRCh38
rs368686203 Chr.8: 12379336-12379336 on Build GRCh38
rs373861475 Chr.8: 12379482-12379482 on Build GRCh38
rs200383517 Chr.8: 12379499-12379499 on Build GRCh38

View Full Product Details