Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 39522641-39523186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM3A
Entrez Gene ID
1587
Gene Name
ADAM metallopeptidase domain 3A (pseudogene)
Gene Aliases
ADAM3, CYRN1, tMDCI
UniGene
Hs.98848
Gene Chromosome Location
Chr.8: 39451045-39522989 on Build GRCh38
Transcripts
NR_001569.3 NR_024106.2 NR_024107.2 NR_073423.1

SNP Detail

SNP ID Chromosome Location
rs969011290 Chr.8: 39522865-39522865 on Build GRCh38
rs534029007 Chr.8: 39522835-39522835 on Build GRCh38
rs1021915141 Chr.8: 39522849-39522849 on Build GRCh38
rs116900755 Chr.8: 39523157-39523157 on Build GRCh38
rs746184751 Chr.8: 39523011-39523011 on Build GRCh38
rs571267073 Chr.8: 39522809-39522809 on Build GRCh38
rs148018918 Chr.8: 39523147-39523147 on Build GRCh38
rs7341671 Chr.8: 39522877-39522877 on Build GRCh38
rs1006286022 Chr.8: 39522932-39522932 on Build GRCh38
rs543044836 Chr.8: 39522686-39522686 on Build GRCh38
rs1014105407 Chr.8: 39522960-39522960 on Build GRCh38
rs964535865 Chr.8: 39523106-39523106 on Build GRCh38
rs886250491 Chr.8: 39522866-39522866 on Build GRCh38
rs910631954 Chr.8: 39523072-39523072 on Build GRCh38
rs970122171 Chr.8: 39522998-39522998 on Build GRCh38
rs1035661234 Chr.8: 39523013-39523013 on Build GRCh38
rs977436654 Chr.8: 39522724-39522724 on Build GRCh38
rs551498955 Chr.8: 39522795-39522795 on Build GRCh38
rs969148027 Chr.8: 39522721-39522721 on Build GRCh38
rs866899441 Chr.8: 39522920-39522920 on Build GRCh38
rs1047403130 Chr.8: 39522817-39522817 on Build GRCh38
rs74562221 Chr.8: 39523136-39523136 on Build GRCh38
rs1013073016 Chr.8: 39522844-39522844 on Build GRCh38
rs1054875119 Chr.8: 39522839-39522839 on Build GRCh38
rs1031674632 Chr.8: 39523033-39523033 on Build GRCh38
rs1016290605 Chr.8: 39522967-39522967 on Build GRCh38
rs760145841 Chr.8: 39522743-39522747 on Build GRCh38
rs973194861 Chr.8: 39523144-39523144 on Build GRCh38
rs73674587 Chr.8: 39522738-39522738 on Build GRCh38
rs538114283 Chr.8: 39522794-39522794 on Build GRCh38
rs894687992 Chr.8: 39522843-39522843 on Build GRCh38
rs910392643 Chr.8: 39522774-39522774 on Build GRCh38
rs556697257 Chr.8: 39522952-39522952 on Build GRCh38
rs894349285 Chr.8: 39522933-39522933 on Build GRCh38
rs924999420 Chr.8: 39522714-39522714 on Build GRCh38
rs536481755 Chr.8: 39522924-39522924 on Build GRCh38
rs1038992157 Chr.8: 39522751-39522751 on Build GRCh38
rs547123667 Chr.8: 39522971-39522971 on Build GRCh38
rs147777660 Chr.8: 39522735-39522735 on Build GRCh38
rs954837246 Chr.8: 39523054-39523054 on Build GRCh38
rs1021882625 Chr.8: 39522701-39522701 on Build GRCh38
rs906154821 Chr.8: 39522823-39522823 on Build GRCh38
rs141156403 Chr.8: 39522789-39522789 on Build GRCh38
rs549153582 Chr.8: 39522760-39522760 on Build GRCh38
rs771097953 Chr.8: 39522780-39522780 on Build GRCh38
rs180776500 Chr.8: 39522922-39522922 on Build GRCh38
rs901761378 Chr.8: 39522752-39522752 on Build GRCh38
rs143438796 Chr.8: 39523057-39523057 on Build GRCh38
rs868625662 Chr.8: 39522969-39522969 on Build GRCh38

View Full Product Details