Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 33509061-33509629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUGT1P1
Entrez Gene ID
441394
Gene Name
SGT1 homolog, MIS12 kinetochore complex assembly cochaperone pseudogene 1
Gene Aliases
SGT1P, SUGT1P, bA255A11.1
UniGene
Hs.521441
Gene Chromosome Location
Chr.9: 33500756-33511166 on Build GRCh38
Transcripts
NR_003667.3

SNP Detail

SNP ID Chromosome Location
rs1008593893 Chr.9: 33509431-33509431 on Build GRCh38
rs543764908 Chr.9: 33509115-33509115 on Build GRCh38
rs1017274210 Chr.9: 33509443-33509443 on Build GRCh38
rs764896107 Chr.9: 33509250-33509250 on Build GRCh38
rs551971934 Chr.9: 33509332-33509332 on Build GRCh38
rs563619705 Chr.9: 33509577-33509577 on Build GRCh38
rs193131906 Chr.9: 33509247-33509247 on Build GRCh38
rs537234750 Chr.9: 33509305-33509305 on Build GRCh38
rs949333218 Chr.9: 33509570-33509573 on Build GRCh38
rs140586446 Chr.9: 33509405-33509405 on Build GRCh38
rs768985819 Chr.9: 33509134-33509134 on Build GRCh38
rs563558719 Chr.9: 33509118-33509118 on Build GRCh38
rs192154991 Chr.9: 33509422-33509422 on Build GRCh38
rs575433371 Chr.9: 33509530-33509530 on Build GRCh38
rs908539261 Chr.9: 33509539-33509539 on Build GRCh38
rs764981446 Chr.9: 33509423-33509423 on Build GRCh38
rs139897874 Chr.9: 33509503-33509503 on Build GRCh38
rs1046299051 Chr.9: 33509342-33509342 on Build GRCh38
rs556971033 Chr.9: 33509517-33509517 on Build GRCh38
rs986953533 Chr.9: 33509276-33509276 on Build GRCh38
rs528937050 Chr.9: 33509241-33509241 on Build GRCh38
rs912728114 Chr.9: 33509284-33509284 on Build GRCh38
rs1047474536 Chr.9: 33509323-33509332 on Build GRCh38
rs761771732 Chr.9: 33509507-33509508 on Build GRCh38
rs1002121530 Chr.9: 33509129-33509129 on Build GRCh38
rs752358051 Chr.9: 33509573-33509573 on Build GRCh38
rs988190736 Chr.9: 33509226-33509226 on Build GRCh38
rs528141475 Chr.9: 33509263-33509263 on Build GRCh38
rs376105632 Chr.9: 33509147-33509147 on Build GRCh38
rs570350874 Chr.9: 33509380-33509380 on Build GRCh38
rs567619019 Chr.9: 33509379-33509379 on Build GRCh38
rs957731620 Chr.9: 33509190-33509190 on Build GRCh38
rs761403805 Chr.9: 33509173-33509173 on Build GRCh38
rs760547247 Chr.9: 33509378-33509378 on Build GRCh38
rs550906362 Chr.9: 33509307-33509307 on Build GRCh38
rs905131539 Chr.9: 33509120-33509123 on Build GRCh38
rs548697027 Chr.9: 33509246-33509246 on Build GRCh38
rs187636866 Chr.9: 33509419-33509419 on Build GRCh38
rs7027139 Chr.9: 33509494-33509494 on Build GRCh38
rs975963075 Chr.9: 33509500-33509500 on Build GRCh38
rs538726533 Chr.9: 33509427-33509427 on Build GRCh38
rs920131524 Chr.9: 33509301-33509301 on Build GRCh38
rs184610566 Chr.9: 33509412-33509412 on Build GRCh38
rs1020674410 Chr.9: 33509237-33509237 on Build GRCh38
rs536819663 Chr.9: 33509300-33509300 on Build GRCh38
rs926672192 Chr.9: 33509572-33509572 on Build GRCh38
rs779780536 Chr.9: 33509364-33509364 on Build GRCh38
rs1046522598 Chr.9: 33509571-33509571 on Build GRCh38
rs551973655 Chr.9: 33509293-33509293 on Build GRCh38
rs949428026 Chr.9: 33509341-33509342 on Build GRCh38

View Full Product Details