Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 33674407-33674968 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTENP1
Entrez Gene ID
11191
Gene Name
phosphatase and tensin homolog pseudogene 1
Gene Aliases
PTEN-rs, PTEN2, PTENpg1, PTH2, psiPTEN
UniGene
Hs.493716
Gene Chromosome Location
Chr.9: 33673504-33677420 on Build GRCh38
Transcripts
NR_023917.1

SNP Detail

SNP ID Chromosome Location
rs10814026 Chr.9: 33674791-33674791 on Build GRCh38
rs765503744 Chr.9: 33674507-33674507 on Build GRCh38
rs553715608 Chr.9: 33674790-33674790 on Build GRCh38
rs10971638 Chr.9: 33674679-33674679 on Build GRCh38
rs988324808 Chr.9: 33674570-33674570 on Build GRCh38
rs548868205 Chr.9: 33674565-33674565 on Build GRCh38
rs576050221 Chr.9: 33674768-33674768 on Build GRCh38
rs913549709 Chr.9: 33674597-33674597 on Build GRCh38
rs980590551 Chr.9: 33674557-33674558 on Build GRCh38
rs10814025 Chr.9: 33674779-33674779 on Build GRCh38
rs531074377 Chr.9: 33674823-33674823 on Build GRCh38
rs535596882 Chr.9: 33674558-33674558 on Build GRCh38
rs565274606 Chr.9: 33674799-33674799 on Build GRCh38
rs887210452 Chr.9: 33674789-33674789 on Build GRCh38
rs544387659 Chr.9: 33674888-33674888 on Build GRCh38
rs536740044 Chr.9: 33674722-33674722 on Build GRCh38
rs190060084 Chr.9: 33674941-33674941 on Build GRCh38
rs201109094 Chr.9: 33674455-33674456 on Build GRCh38
rs969308379 Chr.9: 33674556-33674556 on Build GRCh38
rs958132295 Chr.9: 33674885-33674885 on Build GRCh38
rs1002564227 Chr.9: 33674876-33674876 on Build GRCh38
rs114679179 Chr.9: 33674503-33674503 on Build GRCh38
rs771071074 Chr.9: 33674636-33674636 on Build GRCh38
rs776300811 Chr.9: 33674658-33674658 on Build GRCh38
rs1024981898 Chr.9: 33674824-33674824 on Build GRCh38
rs1047202430 Chr.9: 33674779-33674780 on Build GRCh38
rs150668519 Chr.9: 33674668-33674668 on Build GRCh38
rs192450748 Chr.9: 33674587-33674587 on Build GRCh38
rs556979595 Chr.9: 33674723-33674723 on Build GRCh38
rs184811851 Chr.9: 33674796-33674796 on Build GRCh38
rs12004082 Chr.9: 33674752-33674752 on Build GRCh38
rs569739613 Chr.9: 33674518-33674518 on Build GRCh38
rs1024874483 Chr.9: 33674513-33674513 on Build GRCh38
rs905105880 Chr.9: 33674832-33674832 on Build GRCh38
rs1020630241 Chr.9: 33674934-33674934 on Build GRCh38
rs987950937 Chr.9: 33674914-33674914 on Build GRCh38
rs188031847 Chr.9: 33674471-33674471 on Build GRCh38
rs764102518 Chr.9: 33674780-33674780 on Build GRCh38
rs534350597 Chr.9: 33674607-33674607 on Build GRCh38
rs912782371 Chr.9: 33674640-33674640 on Build GRCh38
rs1032162987 Chr.9: 33674883-33674883 on Build GRCh38

View Full Product Details