Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 85840126-85840697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC389765
Entrez Gene ID
389765
Gene Name
kinesin family member 27 pseudogene
Gene Aliases
-
UniGene
Hs.715986
Gene Chromosome Location
Chr.9: 85806002-85842879 on Build GRCh38
Transcripts
NR_029410.1

SNP Detail

SNP ID Chromosome Location
rs752519622 Chr.9: 85840183-85840183 on Build GRCh38
rs199634922 Chr.9: 85840275-85840275 on Build GRCh38
rs776008398 Chr.9: 85840510-85840510 on Build GRCh38
rs577506111 Chr.9: 85840310-85840310 on Build GRCh38
rs889164473 Chr.9: 85840481-85840481 on Build GRCh38
rs907708633 Chr.9: 85840589-85840589 on Build GRCh38
rs575224137 Chr.9: 85840348-85840348 on Build GRCh38
rs758476081 Chr.9: 85840199-85840199 on Build GRCh38
rs115339478 Chr.9: 85840561-85840561 on Build GRCh38
rs930064996 Chr.9: 85840566-85840566 on Build GRCh38
rs869292054 Chr.9: 85840510-85840511 on Build GRCh38
rs7868282 Chr.9: 85840397-85840397 on Build GRCh38
rs942720330 Chr.9: 85840165-85840165 on Build GRCh38
rs1025114883 Chr.9: 85840359-85840359 on Build GRCh38
rs796395723 Chr.9: 85840659-85840659 on Build GRCh38
rs540125371 Chr.9: 85840480-85840480 on Build GRCh38
rs115239041 Chr.9: 85840553-85840553 on Build GRCh38
rs906299995 Chr.9: 85840376-85840376 on Build GRCh38
rs982866595 Chr.9: 85840585-85840585 on Build GRCh38
rs1006084743 Chr.9: 85840291-85840291 on Build GRCh38
rs62566540 Chr.9: 85840527-85840527 on Build GRCh38
rs922619658 Chr.9: 85840562-85840562 on Build GRCh38
rs886304808 Chr.9: 85840226-85840226 on Build GRCh38
rs200478280 Chr.9: 85840648-85840648 on Build GRCh38
rs1037618932 Chr.9: 85840318-85840318 on Build GRCh38
rs958232145 Chr.9: 85840419-85840419 on Build GRCh38
rs534471287 Chr.9: 85840184-85840184 on Build GRCh38
rs376066330 Chr.9: 85840528-85840528 on Build GRCh38
rs933127158 Chr.9: 85840205-85840205 on Build GRCh38
rs561877286 Chr.9: 85840524-85840525 on Build GRCh38
rs6559883 Chr.9: 85840322-85840322 on Build GRCh38
rs922724884 Chr.9: 85840187-85840187 on Build GRCh38
rs555368649 Chr.9: 85840653-85840653 on Build GRCh38
rs941868923 Chr.9: 85840593-85840593 on Build GRCh38
rs989692915 Chr.9: 85840497-85840497 on Build GRCh38
rs7854337 Chr.9: 85840320-85840320 on Build GRCh38
rs1047538579 Chr.9: 85840208-85840208 on Build GRCh38
rs895074899 Chr.9: 85840627-85840627 on Build GRCh38
rs190361151 Chr.9: 85840407-85840407 on Build GRCh38
rs7868234 Chr.9: 85840608-85840608 on Build GRCh38
rs548252269 Chr.9: 85840162-85840162 on Build GRCh38
rs562718776 Chr.9: 85840591-85840591 on Build GRCh38
rs574125372 Chr.9: 85840434-85840434 on Build GRCh38
rs1007954787 Chr.9: 85840536-85840536 on Build GRCh38
rs538358392 Chr.9: 85840151-85840151 on Build GRCh38
rs989759536 Chr.9: 85840157-85840157 on Build GRCh38
rs186655484 Chr.9: 85840294-85840294 on Build GRCh38
rs559972403 Chr.9: 85840550-85840550 on Build GRCh38
rs1018546324 Chr.9: 85840510-85840510 on Build GRCh38
rs568499149 Chr.9: 85840180-85840180 on Build GRCh38
rs1046047242 Chr.9: 85840654-85840655 on Build GRCh38

View Full Product Details