Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 92837194-92837748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD19P
Entrez Gene ID
138649
Gene Name
ankyrin repeat domain 19, pseudogene
Gene Aliases
ANKRD19, bA526D8.2
UniGene
Hs.643597
Gene Chromosome Location
Chr.9: 92809611-92838457 on Build GRCh38
Transcripts
NR_026868.1

SNP Detail

SNP ID Chromosome Location
rs933098914 Chr.9: 92837466-92837466 on Build GRCh38
rs1023758422 Chr.9: 92837518-92837518 on Build GRCh38
rs960837284 Chr.9: 92837261-92837261 on Build GRCh38
rs971723561 Chr.9: 92837612-92837612 on Build GRCh38
rs1036178469 Chr.9: 92837238-92837238 on Build GRCh38
rs878926704 Chr.9: 92837437-92837437 on Build GRCh38
rs758049521 Chr.9: 92837301-92837301 on Build GRCh38
rs994255622 Chr.9: 92837239-92837239 on Build GRCh38
rs879055865 Chr.9: 92837273-92837273 on Build GRCh38
rs997363427 Chr.9: 92837267-92837267 on Build GRCh38
rs543017562 Chr.9: 92837717-92837717 on Build GRCh38
rs901706217 Chr.9: 92837260-92837260 on Build GRCh38
rs868769924 Chr.9: 92837564-92837564 on Build GRCh38
rs772858385 Chr.9: 92837693-92837693 on Build GRCh38
rs771657621 Chr.9: 92837648-92837648 on Build GRCh38
rs189660466 Chr.9: 92837590-92837590 on Build GRCh38
rs1032564405 Chr.9: 92837711-92837711 on Build GRCh38
rs879101486 Chr.9: 92837452-92837452 on Build GRCh38
rs549430408 Chr.9: 92837295-92837295 on Build GRCh38
rs745590892 Chr.9: 92837508-92837508 on Build GRCh38
rs879031566 Chr.9: 92837275-92837275 on Build GRCh38
rs748852745 Chr.9: 92837546-92837546 on Build GRCh38
rs879215717 Chr.9: 92837384-92837384 on Build GRCh38
rs1001456714 Chr.9: 92837708-92837708 on Build GRCh38
rs1025687014 Chr.9: 92837244-92837244 on Build GRCh38
rs746517649 Chr.9: 92837485-92837485 on Build GRCh38
rs755871001 Chr.9: 92837526-92837526 on Build GRCh38
rs940421199 Chr.9: 92837444-92837444 on Build GRCh38
rs986586338 Chr.9: 92837581-92837581 on Build GRCh38
rs878890815 Chr.9: 92837383-92837383 on Build GRCh38
rs971517614 Chr.9: 92837410-92837410 on Build GRCh38
rs750826095 Chr.9: 92837425-92837425 on Build GRCh38
rs879194962 Chr.9: 92837416-92837416 on Build GRCh38
rs892696088 Chr.9: 92837721-92837721 on Build GRCh38
rs927516618 Chr.9: 92837421-92837421 on Build GRCh38
rs905323416 Chr.9: 92837702-92837702 on Build GRCh38
rs878910364 Chr.9: 92837316-92837316 on Build GRCh38
rs1020340697 Chr.9: 92837403-92837403 on Build GRCh38
rs1034565011 Chr.9: 92837565-92837565 on Build GRCh38
rs969651106 Chr.9: 92837303-92837303 on Build GRCh38
rs957035225 Chr.9: 92837553-92837553 on Build GRCh38
rs575590778 Chr.9: 92837639-92837639 on Build GRCh38
rs879141021 Chr.9: 92837390-92837390 on Build GRCh38
rs1038266666 Chr.9: 92837636-92837636 on Build GRCh38
rs1028888639 Chr.9: 92837300-92837300 on Build GRCh38
rs563159873 Chr.9: 92837312-92837312 on Build GRCh38
rs1036085273 Chr.9: 92837455-92837455 on Build GRCh38
rs879089499 Chr.9: 92837358-92837358 on Build GRCh38
rs878964326 Chr.9: 92837381-92837381 on Build GRCh38
rs898456644 Chr.9: 92837456-92837456 on Build GRCh38
rs879142786 Chr.9: 92837579-92837579 on Build GRCh38
rs764300572 Chr.9: 92837387-92837387 on Build GRCh38
rs61743154 Chr.9: 92837557-92837557 on Build GRCh38
rs560863747 Chr.9: 92837611-92837611 on Build GRCh38
rs1050901340 Chr.9: 92837420-92837420 on Build GRCh38
rs765801717 Chr.9: 92837424-92837424 on Build GRCh38
rs985089119 Chr.9: 92837571-92837571 on Build GRCh38
rs778358623 Chr.9: 92837629-92837629 on Build GRCh38
rs747789633 Chr.9: 92837645-92837645 on Build GRCh38
rs879121368 Chr.9: 92837417-92837417 on Build GRCh38
rs3824428 Chr.9: 92837566-92837566 on Build GRCh38
rs141804236 Chr.9: 92837577-92837577 on Build GRCh38
rs879059561 Chr.9: 92837435-92837435 on Build GRCh38
rs771986683 Chr.9: 92837583-92837583 on Build GRCh38
rs1010158855 Chr.9: 92837722-92837722 on Build GRCh38
rs547965230 Chr.9: 92837432-92837432 on Build GRCh38
rs908899918 Chr.9: 92837428-92837428 on Build GRCh38
rs753994224 Chr.9: 92837412-92837412 on Build GRCh38
rs892382815 Chr.9: 92837716-92837716 on Build GRCh38
rs144558728 Chr.9: 92837516-92837516 on Build GRCh38
rs532550195 Chr.9: 92837330-92837330 on Build GRCh38
rs879175925 Chr.9: 92837246-92837246 on Build GRCh38
rs942535973 Chr.9: 92837615-92837615 on Build GRCh38
rs536927928 Chr.9: 92837502-92837502 on Build GRCh38
rs1050256966 Chr.9: 92837488-92837488 on Build GRCh38
rs878897956 Chr.9: 92837519-92837519 on Build GRCh38
rs111957413 Chr.9: 92837237-92837237 on Build GRCh38
rs7045305 Chr.9: 92837543-92837543 on Build GRCh38
rs893109996 Chr.9: 92837523-92837523 on Build GRCh38
rs988941676 Chr.9: 92837385-92837385 on Build GRCh38
rs933088919 Chr.9: 92837652-92837652 on Build GRCh38
rs10821026 Chr.9: 92837302-92837302 on Build GRCh38
rs148475371 Chr.9: 92837527-92837527 on Build GRCh38
rs878906591 Chr.9: 92837483-92837483 on Build GRCh38
rs896531678 Chr.9: 92837649-92837649 on Build GRCh38
rs542492108 Chr.9: 92837609-92837609 on Build GRCh38
rs957669263 Chr.9: 92837378-92837378 on Build GRCh38
rs879222820 Chr.9: 92837544-92837544 on Build GRCh38
rs12006097 Chr.9: 92837723-92837723 on Build GRCh38
rs1053688583 Chr.9: 92837713-92837713 on Build GRCh38
rs919851496 Chr.9: 92837637-92837637 on Build GRCh38
rs879167222 Chr.9: 92837567-92837567 on Build GRCh38
rs913001851 Chr.9: 92837414-92837414 on Build GRCh38
rs751967762 Chr.9: 92837520-92837520 on Build GRCh38
rs866833751 Chr.9: 92837709-92837709 on Build GRCh38
rs878980697 Chr.9: 92837478-92837478 on Build GRCh38
rs949465608 Chr.9: 92837687-92837687 on Build GRCh38
rs566180462 Chr.9: 92837487-92837487 on Build GRCh38
rs1038726619 Chr.9: 92837443-92837443 on Build GRCh38
rs878887243 Chr.9: 92837552-92837552 on Build GRCh38
rs761746274 Chr.9: 92837415-92837415 on Build GRCh38
rs878869836 Chr.9: 92837407-92837407 on Build GRCh38

View Full Product Details