Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 52628018-52628609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SSX8
Entrez Gene ID
280659
Gene Name
SSX family member 8
Gene Aliases
-
UniGene
Hs.694472
Gene Chromosome Location
Chr.X: 52622935-52633948 on Build GRCh38
Transcripts
NR_027250.2

SNP Detail

SNP ID Chromosome Location
rs782236876 Chr.X: 52628111-52628111 on Build GRCh38
rs370233690 Chr.X: 52628451-52628451 on Build GRCh38
rs782583439 Chr.X: 52628113-52628113 on Build GRCh38
rs782252755 Chr.X: 52628293-52628293 on Build GRCh38
rs781958887 Chr.X: 52628129-52628129 on Build GRCh38
rs181959060 Chr.X: 52628586-52628586 on Build GRCh38
rs782168989 Chr.X: 52628114-52628114 on Build GRCh38
rs145831782 Chr.X: 52628331-52628331 on Build GRCh38
rs373397165 Chr.X: 52628277-52628277 on Build GRCh38
rs782316991 Chr.X: 52628428-52628428 on Build GRCh38
rs782472500 Chr.X: 52628108-52628108 on Build GRCh38
rs781935847 Chr.X: 52628363-52628363 on Build GRCh38
rs782631674 Chr.X: 52628079-52628079 on Build GRCh38
rs377100456 Chr.X: 52628294-52628294 on Build GRCh38
rs190664824 Chr.X: 52628437-52628437 on Build GRCh38
rs782098710 Chr.X: 52628478-52628478 on Build GRCh38
rs140778822 Chr.X: 52628134-52628134 on Build GRCh38
rs782229394 Chr.X: 52628094-52628094 on Build GRCh38
rs782425995 Chr.X: 52628122-52628122 on Build GRCh38
rs150113654 Chr.X: 52628306-52628306 on Build GRCh38
rs782237534 Chr.X: 52628144-52628144 on Build GRCh38
rs782347706 Chr.X: 52628360-52628360 on Build GRCh38

View Full Product Details