Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 73127772-73128314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAP1L6
Entrez Gene ID
645996
Gene Name
nucleosome assembly protein 1 like 6
Gene Aliases
-
UniGene
Hs.617486
Gene Chromosome Location
Chr.X: 73126037-73128080 on Build GRCh38
Transcripts
NR_027291.1

SNP Detail

SNP ID Chromosome Location
rs1049504574 Chr.X: 73128192-73128192 on Build GRCh38
rs782115033 Chr.X: 73128212-73128212 on Build GRCh38
rs782363931 Chr.X: 73127893-73127893 on Build GRCh38
rs181194657 Chr.X: 73128102-73128102 on Build GRCh38
rs782250833 Chr.X: 73127874-73127874 on Build GRCh38
rs377178047 Chr.X: 73128185-73128185 on Build GRCh38
rs782637663 Chr.X: 73127937-73127937 on Build GRCh38
rs12835457 Chr.X: 73128243-73128243 on Build GRCh38
rs997228093 Chr.X: 73127953-73127953 on Build GRCh38
rs905868412 Chr.X: 73127808-73127808 on Build GRCh38
rs937330370 Chr.X: 73128191-73128191 on Build GRCh38
rs782203763 Chr.X: 73128019-73128019 on Build GRCh38
rs1028787113 Chr.X: 73127978-73127978 on Build GRCh38
rs782621419 Chr.X: 73127932-73127932 on Build GRCh38
rs45629131 Chr.X: 73128229-73128229 on Build GRCh38
rs1036624353 Chr.X: 73128259-73128259 on Build GRCh38
rs1001523143 Chr.X: 73127836-73127836 on Build GRCh38
rs979115167 Chr.X: 73127996-73127996 on Build GRCh38
rs782230166 Chr.X: 73127923-73127924 on Build GRCh38
rs782757339 Chr.X: 73127879-73127879 on Build GRCh38
rs1049304152 Chr.X: 73127845-73127845 on Build GRCh38
rs373025063 Chr.X: 73128167-73128167 on Build GRCh38
rs966245575 Chr.X: 73127951-73127951 on Build GRCh38
rs925208404 Chr.X: 73128008-73128008 on Build GRCh38
rs887802169 Chr.X: 73128208-73128208 on Build GRCh38
rs142112395 Chr.X: 73128081-73128081 on Build GRCh38
rs953021423 Chr.X: 73127984-73127984 on Build GRCh38
rs782405407 Chr.X: 73128142-73128142 on Build GRCh38
rs782295820 Chr.X: 73128146-73128146 on Build GRCh38

View Full Product Details