Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 101892933-101893486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJA1P5
Entrez Gene ID
94236
Gene Name
DnaJ heat shock protein family (Hsp40) member A1 pseudogene 5
Gene Aliases
HEJ1
UniGene
Hs.606193
Gene Chromosome Location
Chr.1: 101872012-101894743 on Build GRCh38
Transcripts
NR_033424.2 NM_001288823.1 NM_058170.3 NR_110210.1 NR_110211.1

Gene Symbol
OLFM3
Entrez Gene ID
118427
Gene Name
olfactomedin 3
Gene Aliases
NOE3, NOELIN3, OPTIMEDIN
UniGene
Hs.484475
Gene Chromosome Location
Chr.1: 101800028-101997234 on Build GRCh38
Transcripts
NR_033424.2 NM_001288823.1 NM_058170.3 NR_110210.1 NR_110211.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5761500 c.194A>C p.*65S Chr.1: 101893308-101893308 on Build GRCh38
5352525 c.137delA p.K49fs*8 Chr.1: 101893251-101893251 on Build GRCh38
220059 c.133G>A p.G45R Chr.1: 101893247-101893247 on Build GRCh38
7307351 c.137A>G p.E46G Chr.1: 101893251-101893251 on Build GRCh38
4216924 c.138A>G p.E46E Chr.1: 101893252-101893252 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1006868085 Chr.1: 101892974-101892974 on Build GRCh38
rs532856345 Chr.1: 101892973-101892973 on Build GRCh38
rs3748653 Chr.1: 101893446-101893446 on Build GRCh38
rs751714119 Chr.1: 101893181-101893181 on Build GRCh38
rs750741441 Chr.1: 101893277-101893277 on Build GRCh38
rs149392954 Chr.1: 101893239-101893239 on Build GRCh38
rs137983447 Chr.1: 101893087-101893087 on Build GRCh38
rs1010083341 Chr.1: 101893425-101893425 on Build GRCh38
rs781427412 Chr.1: 101893263-101893263 on Build GRCh38
rs1039769309 Chr.1: 101893291-101893291 on Build GRCh38
rs1034069201 Chr.1: 101893182-101893182 on Build GRCh38
rs3891557 Chr.1: 101892987-101892987 on Build GRCh38
rs528369964 Chr.1: 101893244-101893244 on Build GRCh38
rs1037065246 Chr.1: 101893048-101893048 on Build GRCh38
rs910175555 Chr.1: 101893019-101893019 on Build GRCh38
rs940309225 Chr.1: 101893264-101893264 on Build GRCh38
rs1048195503 Chr.1: 101893251-101893251 on Build GRCh38
rs111919363 Chr.1: 101893117-101893117 on Build GRCh38
rs776135859 Chr.1: 101893125-101893125 on Build GRCh38
rs1000034291 Chr.1: 101893110-101893110 on Build GRCh38
rs1048493956 Chr.1: 101892960-101892960 on Build GRCh38
rs1023948337 Chr.1: 101893140-101893140 on Build GRCh38
rs973000113 Chr.1: 101893267-101893267 on Build GRCh38
rs116598506 Chr.1: 101893129-101893129 on Build GRCh38
rs144794765 Chr.1: 101893330-101893330 on Build GRCh38
rs112454819 Chr.1: 101893029-101893029 on Build GRCh38
rs538711792 Chr.1: 101893329-101893329 on Build GRCh38
rs901147154 Chr.1: 101892985-101892985 on Build GRCh38
rs1000358713 Chr.1: 101893064-101893064 on Build GRCh38
rs910112333 Chr.1: 101893251-101893251 on Build GRCh38
rs112649894 Chr.1: 101893346-101893346 on Build GRCh38
rs903433746 Chr.1: 101893362-101893362 on Build GRCh38
rs918501085 Chr.1: 101893220-101893220 on Build GRCh38
rs1049803780 Chr.1: 101893292-101893292 on Build GRCh38
rs891686408 Chr.1: 101893405-101893405 on Build GRCh38
rs534630211 Chr.1: 101893230-101893230 on Build GRCh38
rs150335893 Chr.1: 101892983-101892983 on Build GRCh38
rs752854731 Chr.1: 101893037-101893037 on Build GRCh38
rs765767761 Chr.1: 101893172-101893172 on Build GRCh38
rs940008366 Chr.1: 101893024-101893024 on Build GRCh38
rs564348357 Chr.1: 101893260-101893261 on Build GRCh38
rs7417910 Chr.1: 101893310-101893310 on Build GRCh38
rs922569225 Chr.1: 101893275-101893275 on Build GRCh38
rs982442368 Chr.1: 101893157-101893157 on Build GRCh38
rs7417894 Chr.1: 101893238-101893238 on Build GRCh38
rs571390014 Chr.1: 101893321-101893321 on Build GRCh38
rs563092442 Chr.1: 101893082-101893082 on Build GRCh38
rs35873118 Chr.1: 101893103-101893104 on Build GRCh38
rs926768315 Chr.1: 101892964-101892964 on Build GRCh38
rs182469635 Chr.1: 101893351-101893351 on Build GRCh38
rs61808281 Chr.1: 101893249-101893249 on Build GRCh38
rs998248848 Chr.1: 101893057-101893057 on Build GRCh38
rs929849547 Chr.1: 101893250-101893251 on Build GRCh38
rs571441400 Chr.1: 101893245-101893245 on Build GRCh38
rs971054538 Chr.1: 101893163-101893163 on Build GRCh38
rs901240266 Chr.1: 101893054-101893054 on Build GRCh38
rs3898278 Chr.1: 101893099-101893099 on Build GRCh38
rs1049087109 Chr.1: 101893000-101893000 on Build GRCh38

View Full Product Details