Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 202177822-202178365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPRVP
Entrez Gene ID
148713
Gene Name
protein tyrosine phosphatase, receptor type V, pseudogene
Gene Aliases
ESP, OST-PTP, PTPRV
UniGene
Hs.523870
Gene Chromosome Location
Chr.1: 202168051-202189449 on Build GRCh38
Transcripts
NR_002930.2

SNP Detail

SNP ID Chromosome Location
rs1014330630 Chr.1: 202178242-202178242 on Build GRCh38
rs574797759 Chr.1: 202178325-202178325 on Build GRCh38
rs973269881 Chr.1: 202177976-202177976 on Build GRCh38
rs988620224 Chr.1: 202178112-202178112 on Build GRCh38
rs969869680 Chr.1: 202178070-202178070 on Build GRCh38
rs572233573 Chr.1: 202178260-202178260 on Build GRCh38
rs1014648950 Chr.1: 202177912-202177912 on Build GRCh38
rs1044029594 Chr.1: 202178329-202178329 on Build GRCh38
rs756315631 Chr.1: 202178291-202178291 on Build GRCh38
rs1047418621 Chr.1: 202177985-202177985 on Build GRCh38
rs960803160 Chr.1: 202178106-202178106 on Build GRCh38
rs936135298 Chr.1: 202178131-202178131 on Build GRCh38
rs774509558 Chr.1: 202178025-202178025 on Build GRCh38
rs988196834 Chr.1: 202177877-202177877 on Build GRCh38
rs35994064 Chr.1: 202178328-202178328 on Build GRCh38
rs1014816484 Chr.1: 202178292-202178292 on Build GRCh38
rs769261595 Chr.1: 202177862-202177862 on Build GRCh38
rs145848916 Chr.1: 202177959-202177959 on Build GRCh38
rs187578149 Chr.1: 202178006-202178006 on Build GRCh38
rs764470556 Chr.1: 202178013-202178013 on Build GRCh38
rs558412477 Chr.1: 202178212-202178212 on Build GRCh38
rs910744183 Chr.1: 202177958-202177958 on Build GRCh38
rs943594613 Chr.1: 202178259-202178259 on Build GRCh38
rs980822664 Chr.1: 202177863-202177863 on Build GRCh38
rs911544241 Chr.1: 202178037-202178037 on Build GRCh38
rs911679715 Chr.1: 202178344-202178344 on Build GRCh38
rs374009497 Chr.1: 202177974-202177974 on Build GRCh38
rs910773605 Chr.1: 202178195-202178195 on Build GRCh38
rs10800822 Chr.1: 202178117-202178117 on Build GRCh38
rs1046260079 Chr.1: 202178253-202178253 on Build GRCh38
rs534513294 Chr.1: 202178285-202178285 on Build GRCh38
rs923420441 Chr.1: 202178283-202178283 on Build GRCh38
rs960768835 Chr.1: 202177866-202177866 on Build GRCh38
rs149229514 Chr.1: 202178181-202178181 on Build GRCh38
rs762227558 Chr.1: 202178087-202178087 on Build GRCh38
rs987217792 Chr.1: 202178337-202178337 on Build GRCh38
rs549318176 Chr.1: 202178033-202178033 on Build GRCh38
rs1055987478 Chr.1: 202178172-202178172 on Build GRCh38
rs375537712 Chr.1: 202178322-202178322 on Build GRCh38
rs35586904 Chr.1: 202177850-202177850 on Build GRCh38
rs190536700 Chr.1: 202178203-202178203 on Build GRCh38
rs552828086 Chr.1: 202177928-202177928 on Build GRCh38
rs60852204 Chr.1: 202177911-202177911 on Build GRCh38
rs894523979 Chr.1: 202178184-202178184 on Build GRCh38
rs567248129 Chr.1: 202178000-202178000 on Build GRCh38
rs1041947092 Chr.1: 202178071-202178071 on Build GRCh38
rs928015867 Chr.1: 202177865-202177865 on Build GRCh38
rs543441728 Chr.1: 202177847-202177847 on Build GRCh38
rs527769242 Chr.1: 202178026-202178026 on Build GRCh38
rs370809838 Chr.1: 202178182-202178182 on Build GRCh38
rs993652342 Chr.1: 202177984-202177984 on Build GRCh38
rs61757817 Chr.1: 202178315-202178315 on Build GRCh38
rs913875741 Chr.1: 202177893-202177893 on Build GRCh38
rs183390531 Chr.1: 202178326-202178326 on Build GRCh38
rs547497823 Chr.1: 202177975-202177975 on Build GRCh38
rs921636338 Chr.1: 202177978-202177978 on Build GRCh38
rs1034838583 Chr.1: 202177900-202177900 on Build GRCh38
rs762539005 Chr.1: 202177971-202177971 on Build GRCh38
rs138333174 Chr.1: 202177972-202177972 on Build GRCh38
rs750915476 Chr.1: 202178160-202178160 on Build GRCh38
rs776867070 Chr.1: 202178091-202178091 on Build GRCh38
rs1000358290 Chr.1: 202177898-202177898 on Build GRCh38
rs1022810474 Chr.1: 202177859-202177859 on Build GRCh38
rs1023264759 Chr.1: 202178053-202178053 on Build GRCh38

View Full Product Details