Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 220266828-220267375 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AURKAPS1
Entrez Gene ID
6791
Gene Name
aurora kinase A pseudogene 1
Gene Aliases
AurAps1, STK6P
UniGene
Hs.654849
Gene Chromosome Location
Chr.1: 220266179-220267715 on Build GRCh38
Transcripts
NR_001587.1 NM_012414.3

Gene Symbol
RAB3GAP2
Entrez Gene ID
25782
Gene Name
RAB3 GTPase activating non-catalytic protein subunit 2
Gene Aliases
RAB3-GAP150, RAB3GAP150, SPG69, WARBM2, p150
UniGene
Hs.654849
Gene Chromosome Location
Chr.1: 220148268-220272501 on Build GRCh38
Transcripts
NR_001587.1 NM_012414.3

SNP Detail

SNP ID Chromosome Location
rs772836849 Chr.1: 220267310-220267310 on Build GRCh38
rs374849440 Chr.1: 220267219-220267219 on Build GRCh38
rs1003322993 Chr.1: 220266897-220266897 on Build GRCh38
rs945996429 Chr.1: 220266888-220266888 on Build GRCh38
rs938682090 Chr.1: 220267325-220267325 on Build GRCh38
rs538869103 Chr.1: 220267007-220267007 on Build GRCh38
rs150070001 Chr.1: 220267215-220267215 on Build GRCh38
rs769292505 Chr.1: 220267301-220267301 on Build GRCh38
rs568642577 Chr.1: 220267024-220267024 on Build GRCh38
rs566548264 Chr.1: 220266858-220266858 on Build GRCh38
rs955763575 Chr.1: 220266922-220266922 on Build GRCh38
rs963790050 Chr.1: 220267150-220267150 on Build GRCh38
rs1007356249 Chr.1: 220267351-220267351 on Build GRCh38
rs574235001 Chr.1: 220267111-220267111 on Build GRCh38
rs542955377 Chr.1: 220267176-220267176 on Build GRCh38
rs531046539 Chr.1: 220267298-220267298 on Build GRCh38
rs1055826791 Chr.1: 220267330-220267330 on Build GRCh38
rs746774515 Chr.1: 220266983-220266983 on Build GRCh38
rs1007067905 Chr.1: 220266959-220266959 on Build GRCh38
rs999475913 Chr.1: 220267280-220267280 on Build GRCh38
rs879268224 Chr.1: 220267201-220267201 on Build GRCh38
rs1029745183 Chr.1: 220266903-220266903 on Build GRCh38
rs139745899 Chr.1: 220267186-220267186 on Build GRCh38
rs1055879903 Chr.1: 220266901-220266901 on Build GRCh38
rs537422118 Chr.1: 220267032-220267032 on Build GRCh38
rs916387203 Chr.1: 220267074-220267074 on Build GRCh38
rs558216017 Chr.1: 220267008-220267008 on Build GRCh38
rs1042074034 Chr.1: 220266891-220266891 on Build GRCh38
rs748064813 Chr.1: 220267288-220267288 on Build GRCh38
rs553675876 Chr.1: 220267182-220267182 on Build GRCh38
rs999590173 Chr.1: 220266894-220266894 on Build GRCh38
rs946666563 Chr.1: 220267210-220267210 on Build GRCh38
rs914416667 Chr.1: 220267242-220267242 on Build GRCh38
rs41304119 Chr.1: 220267072-220267072 on Build GRCh38
rs927742033 Chr.1: 220267043-220267043 on Build GRCh38
rs928576703 Chr.1: 220267304-220267304 on Build GRCh38
rs527448002 Chr.1: 220267021-220267021 on Build GRCh38
rs780317476 Chr.1: 220266856-220266856 on Build GRCh38
rs974308216 Chr.1: 220267001-220267001 on Build GRCh38
rs939057478 Chr.1: 220267064-220267064 on Build GRCh38
rs367895298 Chr.1: 220267025-220267040 on Build GRCh38
rs1006374995 Chr.1: 220266928-220266928 on Build GRCh38
rs894591472 Chr.1: 220266921-220266921 on Build GRCh38
rs955066110 Chr.1: 220267023-220267024 on Build GRCh38
rs962968865 Chr.1: 220266991-220266991 on Build GRCh38

View Full Product Details