Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 43897234-43897783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALKBH3
Entrez Gene ID
221120
Gene Name
alkB homolog 3, alpha-ketoglutaratedependent dioxygenase
Gene Aliases
ABH3, DEPC-1, DEPC1, PCA1, hABH3
UniGene
Hs.720708
Gene Chromosome Location
Chr.11: 43880807-43920275 on Build GRCh38
Transcripts
NM_139178.3 NR_026952.1

Gene Symbol
SEC14L1P1
Entrez Gene ID
729799
Gene Name
SEC14 like 1 pseudogene 1
Gene Aliases
-
UniGene
Hs.368920
Gene Chromosome Location
Chr.11: 43897303-43899888 on Build GRCh38
Transcripts
NM_139178.3 NR_026952.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5492778 c.407T>G p.V136G Chr.11: 43897234-43897234 on Build GRCh38
5360677 c.449G>A p.R150H Chr.11: 43897276-43897276 on Build GRCh38
7091777 c.429G>A p.P143P Chr.11: 43897256-43897256 on Build GRCh38
6885075 c.509C>T p.P170L Chr.11: 43897336-43897336 on Build GRCh38
4135677 c.474C>T p.G158G Chr.11: 43897301-43897301 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs941144223 Chr.11: 43897738-43897738 on Build GRCh38
rs765220294 Chr.11: 43897627-43897627 on Build GRCh38
rs910017558 Chr.11: 43897553-43897553 on Build GRCh38
rs1130575 Chr.11: 43897542-43897542 on Build GRCh38
rs989368904 Chr.11: 43897378-43897378 on Build GRCh38
rs975981132 Chr.11: 43897749-43897749 on Build GRCh38
rs77207849 Chr.11: 43897707-43897707 on Build GRCh38
rs188912695 Chr.11: 43897279-43897279 on Build GRCh38
rs368918085 Chr.11: 43897275-43897275 on Build GRCh38
rs1030800761 Chr.11: 43897355-43897355 on Build GRCh38
rs771622854 Chr.11: 43897291-43897291 on Build GRCh38
rs1022746574 Chr.11: 43897484-43897484 on Build GRCh38
rs765296653 Chr.11: 43897335-43897335 on Build GRCh38
rs117994943 Chr.11: 43897256-43897256 on Build GRCh38
rs1032277016 Chr.11: 43897556-43897556 on Build GRCh38
rs771327999 Chr.11: 43897308-43897308 on Build GRCh38
rs574191020 Chr.11: 43897362-43897362 on Build GRCh38
rs956179407 Chr.11: 43897557-43897557 on Build GRCh38
rs774804678 Chr.11: 43897282-43897282 on Build GRCh38
rs749348035 Chr.11: 43897280-43897280 on Build GRCh38
rs751692480 Chr.11: 43897260-43897260 on Build GRCh38
rs920566153 Chr.11: 43897632-43897632 on Build GRCh38
rs781290325 Chr.11: 43897267-43897267 on Build GRCh38
rs900071725 Chr.11: 43897576-43897576 on Build GRCh38
rs750335965 Chr.11: 43897342-43897342 on Build GRCh38
rs537949900 Chr.11: 43897428-43897428 on Build GRCh38
rs1011136545 Chr.11: 43897676-43897676 on Build GRCh38
rs987711895 Chr.11: 43897579-43897579 on Build GRCh38
rs943413549 Chr.11: 43897596-43897596 on Build GRCh38
rs758386445 Chr.11: 43897343-43897343 on Build GRCh38
rs895726493 Chr.11: 43897720-43897720 on Build GRCh38
rs909652205 Chr.11: 43897721-43897721 on Build GRCh38
rs551768198 Chr.11: 43897307-43897307 on Build GRCh38
rs566455842 Chr.11: 43897265-43897265 on Build GRCh38
rs62621395 Chr.11: 43897594-43897594 on Build GRCh38
rs778265203 Chr.11: 43897276-43897276 on Build GRCh38
rs893621925 Chr.11: 43897641-43897641 on Build GRCh38
rs749937139 Chr.11: 43897715-43897715 on Build GRCh38
rs558998580 Chr.11: 43897687-43897687 on Build GRCh38
rs753681581 Chr.11: 43897326-43897326 on Build GRCh38
rs1048703524 Chr.11: 43897365-43897365 on Build GRCh38
rs748449972 Chr.11: 43897268-43897268 on Build GRCh38
rs998906496 Chr.11: 43897630-43897630 on Build GRCh38
rs143752599 Chr.11: 43897686-43897686 on Build GRCh38
rs1023992989 Chr.11: 43897411-43897411 on Build GRCh38
rs570309961 Chr.11: 43897422-43897422 on Build GRCh38
rs556031421 Chr.11: 43897460-43897460 on Build GRCh38
rs11600211 Chr.11: 43897372-43897372 on Build GRCh38
rs547295434 Chr.11: 43897453-43897453 on Build GRCh38
rs772934903 Chr.11: 43897561-43897561 on Build GRCh38
rs886922079 Chr.11: 43897416-43897416 on Build GRCh38
rs879056196 Chr.11: 43897653-43897653 on Build GRCh38
rs755069273 Chr.11: 43897264-43897264 on Build GRCh38
rs749083050 Chr.11: 43897570-43897570 on Build GRCh38
rs759071688 Chr.11: 43897283-43897283 on Build GRCh38
rs568136984 Chr.11: 43897506-43897506 on Build GRCh38
rs763917154 Chr.11: 43897451-43897451 on Build GRCh38
rs570375596 Chr.11: 43897590-43897590 on Build GRCh38
rs192564360 Chr.11: 43897619-43897619 on Build GRCh38
rs556058063 Chr.11: 43897651-43897651 on Build GRCh38
rs116511955 Chr.11: 43897703-43897703 on Build GRCh38
rs781330582 Chr.11: 43897405-43897405 on Build GRCh38
rs866723894 Chr.11: 43897640-43897640 on Build GRCh38
rs968879971 Chr.11: 43897541-43897541 on Build GRCh38
rs899243978 Chr.11: 43897299-43897299 on Build GRCh38
rs1012804843 Chr.11: 43897748-43897748 on Build GRCh38
rs745864562 Chr.11: 43897727-43897727 on Build GRCh38
rs774832632 Chr.11: 43897292-43897292 on Build GRCh38
rs761632476 Chr.11: 43897332-43897332 on Build GRCh38
rs778936509 Chr.11: 43897259-43897259 on Build GRCh38
rs753573140 Chr.11: 43897255-43897255 on Build GRCh38
rs1012585248 Chr.11: 43897482-43897482 on Build GRCh38
rs1049424 Chr.11: 43897758-43897758 on Build GRCh38
rs533538038 Chr.11: 43897425-43897425 on Build GRCh38

View Full Product Details