Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 76716287-76716838 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GUCY2EP
Entrez Gene ID
390226
Gene Name
guanylate cyclase 2E, pseudogene
Gene Aliases
GC-E, GCD, GUCY2E
UniGene
Hs.440026
Gene Chromosome Location
Chr.11: 76680166-76721789 on Build GRCh38
Transcripts
NR_024042.1

SNP Detail

SNP ID Chromosome Location
rs533003891 Chr.11: 76716812-76716812 on Build GRCh38
rs1024095783 Chr.11: 76716515-76716515 on Build GRCh38
rs918025724 Chr.11: 76716372-76716372 on Build GRCh38
rs774787931 Chr.11: 76716391-76716391 on Build GRCh38
rs913332442 Chr.11: 76716375-76716375 on Build GRCh38
rs10899265 Chr.11: 76716630-76716630 on Build GRCh38
rs1017443477 Chr.11: 76716336-76716336 on Build GRCh38
rs1038812196 Chr.11: 76716673-76716673 on Build GRCh38
rs559971676 Chr.11: 76716548-76716548 on Build GRCh38
rs780519713 Chr.11: 76716801-76716801 on Build GRCh38
rs1024167753 Chr.11: 76716367-76716368 on Build GRCh38
rs922106061 Chr.11: 76716637-76716637 on Build GRCh38
rs892096061 Chr.11: 76716454-76716454 on Build GRCh38
rs980874881 Chr.11: 76716598-76716598 on Build GRCh38
rs989670310 Chr.11: 76716813-76716813 on Build GRCh38
rs773740567 Chr.11: 76716472-76716472 on Build GRCh38
rs11236900 Chr.11: 76716779-76716779 on Build GRCh38
rs745374631 Chr.11: 76716685-76716685 on Build GRCh38
rs534997422 Chr.11: 76716363-76716363 on Build GRCh38
rs1009203894 Chr.11: 76716469-76716469 on Build GRCh38
rs1037143554 Chr.11: 76716750-76716750 on Build GRCh38
rs137909534 Chr.11: 76716742-76716742 on Build GRCh38
rs1032982722 Chr.11: 76716370-76716370 on Build GRCh38
rs758252275 Chr.11: 76716609-76716609 on Build GRCh38
rs757160643 Chr.11: 76716641-76716641 on Build GRCh38
rs777536261 Chr.11: 76716627-76716627 on Build GRCh38
rs1024960617 Chr.11: 76716743-76716743 on Build GRCh38
rs748834766 Chr.11: 76716545-76716545 on Build GRCh38
rs566687739 Chr.11: 76716313-76716313 on Build GRCh38
rs557913403 Chr.11: 76716393-76716393 on Build GRCh38
rs970745912 Chr.11: 76716753-76716753 on Build GRCh38
rs943105783 Chr.11: 76716657-76716657 on Build GRCh38
rs917957861 Chr.11: 76716570-76716570 on Build GRCh38
rs1045513527 Chr.11: 76716806-76716807 on Build GRCh38
rs957656491 Chr.11: 76716371-76716371 on Build GRCh38
rs372012979 Chr.11: 76716362-76716362 on Build GRCh38
rs1051373209 Chr.11: 76716631-76716631 on Build GRCh38
rs1015818168 Chr.11: 76716328-76716328 on Build GRCh38
rs905754542 Chr.11: 76716516-76716516 on Build GRCh38
rs982836352 Chr.11: 76716367-76716367 on Build GRCh38
rs909481613 Chr.11: 76716733-76716733 on Build GRCh38
rs35016114 Chr.11: 76716673-76716674 on Build GRCh38
rs146922099 Chr.11: 76716725-76716725 on Build GRCh38
rs559983134 Chr.11: 76716453-76716453 on Build GRCh38
rs898966148 Chr.11: 76716686-76716686 on Build GRCh38
rs143478692 Chr.11: 76716758-76716758 on Build GRCh38
rs779910547 Chr.11: 76716724-76716724 on Build GRCh38
rs56336901 Chr.11: 76716476-76716476 on Build GRCh38
rs766437268 Chr.11: 76716563-76716563 on Build GRCh38
rs377354808 Chr.11: 76716470-76716470 on Build GRCh38
rs1007347535 Chr.11: 76716331-76716331 on Build GRCh38
rs542177017 Chr.11: 76716308-76716308 on Build GRCh38
rs1045250569 Chr.11: 76716383-76716383 on Build GRCh38
rs970799793 Chr.11: 76716576-76716576 on Build GRCh38
rs780711304 Chr.11: 76716658-76716658 on Build GRCh38
rs926769709 Chr.11: 76716601-76716601 on Build GRCh38
rs771374091 Chr.11: 76716774-76716774 on Build GRCh38
rs890161578 Chr.11: 76716643-76716643 on Build GRCh38
rs926849358 Chr.11: 76716386-76716386 on Build GRCh38
rs1054421670 Chr.11: 76716611-76716611 on Build GRCh38
rs773836514 Chr.11: 76716463-76716463 on Build GRCh38
rs10751261 Chr.11: 76716687-76716687 on Build GRCh38
rs954519674 Chr.11: 76716320-76716320 on Build GRCh38
rs980032601 Chr.11: 76716368-76716368 on Build GRCh38
rs542883927 Chr.11: 76716488-76716488 on Build GRCh38
rs1017853977 Chr.11: 76716479-76716479 on Build GRCh38
rs1026245199 Chr.11: 76716524-76716524 on Build GRCh38
rs963620303 Chr.11: 76716337-76716337 on Build GRCh38
rs563460718 Chr.11: 76716380-76716380 on Build GRCh38
rs995151248 Chr.11: 76716489-76716489 on Build GRCh38
rs966159037 Chr.11: 76716624-76716624 on Build GRCh38
rs372010351 Chr.11: 76716514-76716514 on Build GRCh38
rs1030049347 Chr.11: 76716309-76716309 on Build GRCh38
rs776449910 Chr.11: 76716344-76716344 on Build GRCh38
rs374670660 Chr.11: 76716602-76716602 on Build GRCh38
rs56136160 Chr.11: 76716427-76716427 on Build GRCh38
rs780639183 Chr.11: 76716408-76716408 on Build GRCh38
rs765452469 Chr.11: 76716557-76716557 on Build GRCh38

View Full Product Details