Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 9424100-9424671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX12P
Entrez Gene ID
440081
Gene Name
DEAD/H-box helicase 12, pseudogene
Gene Aliases
CHLR2, DDX12
UniGene
Hs.447869
Gene Chromosome Location
Chr.12: 9417691-9448172 on Build GRCh38
Transcripts
NR_033399.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3955318 c.1766G>T p.S589I Chr.12: 9424637-9424637 on Build GRCh38
4676750 c.1826T>C p.V609A Chr.12: 9424577-9424577 on Build GRCh38
1470619 c.1862G>A p.G621E Chr.12: 9424541-9424541 on Build GRCh38
6138227 c.1841G>T p.R614L Chr.12: 9424562-9424562 on Build GRCh38
7112204 c.1770C>G p.L590L Chr.12: 9424633-9424633 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752716183 Chr.12: 9424500-9424500 on Build GRCh38
rs923686494 Chr.12: 9424409-9424409 on Build GRCh38
rs376165112 Chr.12: 9424237-9424237 on Build GRCh38
rs781512598 Chr.12: 9424622-9424622 on Build GRCh38
rs760512009 Chr.12: 9424461-9424461 on Build GRCh38
rs964329374 Chr.12: 9424270-9424270 on Build GRCh38
rs1047212044 Chr.12: 9424460-9424460 on Build GRCh38
rs756795648 Chr.12: 9424134-9424137 on Build GRCh38
rs766746117 Chr.12: 9424207-9424207 on Build GRCh38
rs866994597 Chr.12: 9424139-9424139 on Build GRCh38
rs977851474 Chr.12: 9424406-9424406 on Build GRCh38
rs952895926 Chr.12: 9424201-9424201 on Build GRCh38
rs369938321 Chr.12: 9424450-9424450 on Build GRCh38
rs115863102 Chr.12: 9424455-9424455 on Build GRCh38
rs757990238 Chr.12: 9424341-9424341 on Build GRCh38
rs202206491 Chr.12: 9424139-9424140 on Build GRCh38
rs868183707 Chr.12: 9424135-9424135 on Build GRCh38
rs866673170 Chr.12: 9424474-9424474 on Build GRCh38
rs781629543 Chr.12: 9424219-9424219 on Build GRCh38
rs757785369 Chr.12: 9424633-9424633 on Build GRCh38
rs947860163 Chr.12: 9424640-9424640 on Build GRCh38
rs879823301 Chr.12: 9424620-9424620 on Build GRCh38
rs989333371 Chr.12: 9424418-9424418 on Build GRCh38
rs374009957 Chr.12: 9424502-9424502 on Build GRCh38
rs755408037 Chr.12: 9424290-9424290 on Build GRCh38
rs765657014 Chr.12: 9424627-9424627 on Build GRCh38
rs752874436 Chr.12: 9424127-9424127 on Build GRCh38
rs770381241 Chr.12: 9424291-9424291 on Build GRCh38
rs768301032 Chr.12: 9424637-9424637 on Build GRCh38
rs780383346 Chr.12: 9424138-9424138 on Build GRCh38
rs775559891 Chr.12: 9424462-9424462 on Build GRCh38
rs184609117 Chr.12: 9424140-9424140 on Build GRCh38
rs1029981976 Chr.12: 9424280-9424280 on Build GRCh38
rs779416451 Chr.12: 9424292-9424292 on Build GRCh38
rs376686229 Chr.12: 9424584-9424584 on Build GRCh38
rs59215376 Chr.12: 9424141-9424141 on Build GRCh38
rs879123914 Chr.12: 9424504-9424504 on Build GRCh38
rs201553936 Chr.12: 9424562-9424562 on Build GRCh38
rs954413766 Chr.12: 9424573-9424573 on Build GRCh38
rs61914669 Chr.12: 9424417-9424417 on Build GRCh38
rs765180155 Chr.12: 9424468-9424468 on Build GRCh38
rs778037314 Chr.12: 9424163-9424163 on Build GRCh38
rs750858166 Chr.12: 9424626-9424626 on Build GRCh38
rs182585090 Chr.12: 9424169-9424169 on Build GRCh38
rs868579073 Chr.12: 9424475-9424475 on Build GRCh38
rs750195974 Chr.12: 9424529-9424529 on Build GRCh38
rs3825308 Chr.12: 9424431-9424431 on Build GRCh38
rs869093905 Chr.12: 9424134-9424135 on Build GRCh38
rs1025713706 Chr.12: 9424197-9424197 on Build GRCh38
rs977380720 Chr.12: 9424279-9424279 on Build GRCh38
rs948008035 Chr.12: 9424439-9424439 on Build GRCh38
rs745718991 Chr.12: 9424459-9424459 on Build GRCh38
rs994280745 Chr.12: 9424195-9424195 on Build GRCh38
rs1018825079 Chr.12: 9424227-9424227 on Build GRCh38
rs369826628 Chr.12: 9424629-9424629 on Build GRCh38
rs1046729680 Chr.12: 9424442-9424442 on Build GRCh38
rs998472444 Chr.12: 9424540-9424540 on Build GRCh38
rs899986174 Chr.12: 9424528-9424528 on Build GRCh38
rs186442260 Chr.12: 9424602-9424602 on Build GRCh38
rs957832647 Chr.12: 9424614-9424614 on Build GRCh38
rs778666291 Chr.12: 9424648-9424648 on Build GRCh38
rs773366874 Chr.12: 9424546-9424546 on Build GRCh38
rs768352105 Chr.12: 9424401-9424401 on Build GRCh38
rs767624598 Chr.12: 9424147-9424147 on Build GRCh38
rs868007694 Chr.12: 9424134-9424134 on Build GRCh38

View Full Product Details