Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 13003869-13004427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM234B
Entrez Gene ID
57613
Gene Name
family with sequence similarity 234 member B
Gene Aliases
KIAA1467
UniGene
Hs.132660
Gene Chromosome Location
Chr.12: 12927680-13083449 on Build GRCh38
Transcripts
XM_017019706.1 NR_002774.3

Gene Symbol
HTR7P1
Entrez Gene ID
93164
Gene Name
5-hydroxytryptamine receptor 7 pseudogene 1
Gene Aliases
HTR7P
UniGene
Hs.593282
Gene Chromosome Location
Chr.12: 13000442-13004830 on Build GRCh38
Transcripts
XM_017019706.1 NR_002774.3

SNP Detail

SNP ID Chromosome Location
rs778464917 Chr.12: 13004204-13004204 on Build GRCh38
rs950930479 Chr.12: 13003934-13003934 on Build GRCh38
rs190373709 Chr.12: 13003946-13003946 on Build GRCh38
rs541592488 Chr.12: 13004248-13004248 on Build GRCh38
rs981825376 Chr.12: 13003902-13003902 on Build GRCh38
rs953910897 Chr.12: 13004329-13004329 on Build GRCh38
rs1016488479 Chr.12: 13004280-13004280 on Build GRCh38
rs558498680 Chr.12: 13004166-13004166 on Build GRCh38
rs187666370 Chr.12: 13004365-13004365 on Build GRCh38
rs912222232 Chr.12: 13004336-13004336 on Build GRCh38
rs909374898 Chr.12: 13003943-13003943 on Build GRCh38
rs1005127056 Chr.12: 13004271-13004271 on Build GRCh38
rs780127831 Chr.12: 13004349-13004349 on Build GRCh38
rs914449765 Chr.12: 13004373-13004373 on Build GRCh38
rs1002902583 Chr.12: 13004229-13004229 on Build GRCh38
rs367901938 Chr.12: 13003979-13003979 on Build GRCh38
rs554420264 Chr.12: 13003956-13003956 on Build GRCh38
rs983641858 Chr.12: 13003942-13003942 on Build GRCh38
rs529382396 Chr.12: 13004177-13004177 on Build GRCh38
rs367792749 Chr.12: 13004335-13004335 on Build GRCh38
rs372079089 Chr.12: 13004082-13004082 on Build GRCh38
rs572748063 Chr.12: 13003978-13003978 on Build GRCh38
rs1291359 Chr.12: 13004333-13004333 on Build GRCh38
rs947253132 Chr.12: 13004388-13004388 on Build GRCh38
rs1052918538 Chr.12: 13004073-13004073 on Build GRCh38
rs555906119 Chr.12: 13004023-13004023 on Build GRCh38
rs543843140 Chr.12: 13004087-13004087 on Build GRCh38
rs932350008 Chr.12: 13004022-13004022 on Build GRCh38
rs182991102 Chr.12: 13004106-13004106 on Build GRCh38
rs920991269 Chr.12: 13003989-13003989 on Build GRCh38
rs934294920 Chr.12: 13004350-13004350 on Build GRCh38
rs1035956286 Chr.12: 13003929-13003929 on Build GRCh38
rs568208552 Chr.12: 13003940-13003940 on Build GRCh38
rs963598874 Chr.12: 13004306-13004306 on Build GRCh38
rs941140019 Chr.12: 13003959-13003959 on Build GRCh38
rs568127483 Chr.12: 13004399-13004399 on Build GRCh38
rs1035672243 Chr.12: 13004245-13004245 on Build GRCh38
rs141641408 Chr.12: 13004313-13004313 on Build GRCh38
rs552197793 Chr.12: 13004314-13004314 on Build GRCh38
rs372971187 Chr.12: 13004361-13004361 on Build GRCh38
rs891067075 Chr.12: 13004059-13004059 on Build GRCh38
rs150092684 Chr.12: 13004060-13004060 on Build GRCh38
rs138515078 Chr.12: 13004253-13004253 on Build GRCh38

View Full Product Details