Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 45063733-45064276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RACGAP1P
Entrez Gene ID
83956
Gene Name
Rac GTPase activating protein 1 pseudogene
Gene Aliases
FKSG42
UniGene
Hs.650500
Gene Chromosome Location
Chr.12: 45062618-45065411 on Build GRCh38
Transcripts
NR_026583.1

SNP Detail

SNP ID Chromosome Location
rs568863059 Chr.12: 45064082-45064082 on Build GRCh38
rs935630619 Chr.12: 45064254-45064254 on Build GRCh38
rs1000920366 Chr.12: 45064247-45064247 on Build GRCh38
rs1020185679 Chr.12: 45064091-45064091 on Build GRCh38
rs201643869 Chr.12: 45063822-45063822 on Build GRCh38
rs556299666 Chr.12: 45063884-45063884 on Build GRCh38
rs568269721 Chr.12: 45063767-45063767 on Build GRCh38
rs939829726 Chr.12: 45063901-45063901 on Build GRCh38
rs1044134490 Chr.12: 45064226-45064226 on Build GRCh38
rs141588874 Chr.12: 45064170-45064170 on Build GRCh38
rs952679416 Chr.12: 45064143-45064143 on Build GRCh38
rs184933314 Chr.12: 45064137-45064137 on Build GRCh38
rs779550567 Chr.12: 45064127-45064127 on Build GRCh38
rs143713352 Chr.12: 45063773-45063773 on Build GRCh38
rs1055179708 Chr.12: 45064251-45064251 on Build GRCh38
rs189442486 Chr.12: 45064195-45064195 on Build GRCh38
rs1042458869 Chr.12: 45064232-45064232 on Build GRCh38
rs529087468 Chr.12: 45064142-45064142 on Build GRCh38
rs1013792070 Chr.12: 45064107-45064107 on Build GRCh38
rs772492159 Chr.12: 45064233-45064233 on Build GRCh38
rs1048989393 Chr.12: 45063945-45063945 on Build GRCh38
rs139328774 Chr.12: 45063956-45063956 on Build GRCh38
rs766020529 Chr.12: 45063814-45063814 on Build GRCh38
rs192810876 Chr.12: 45064076-45064076 on Build GRCh38
rs923289035 Chr.12: 45064197-45064197 on Build GRCh38
rs116849587 Chr.12: 45063966-45063966 on Build GRCh38
rs1009651500 Chr.12: 45064050-45064050 on Build GRCh38
rs1033923453 Chr.12: 45063803-45063803 on Build GRCh38
rs919086227 Chr.12: 45063887-45063887 on Build GRCh38
rs930501307 Chr.12: 45063900-45063900 on Build GRCh38
rs903914585 Chr.12: 45064235-45064235 on Build GRCh38
rs1009710722 Chr.12: 45064083-45064083 on Build GRCh38
rs916920027 Chr.12: 45063820-45063820 on Build GRCh38
rs752223336 Chr.12: 45064019-45064019 on Build GRCh38
rs991093314 Chr.12: 45063815-45063815 on Build GRCh38
rs565818084 Chr.12: 45063797-45063797 on Build GRCh38
rs1040366649 Chr.12: 45064003-45064003 on Build GRCh38
rs142339076 Chr.12: 45063777-45063777 on Build GRCh38
rs3803173 Chr.12: 45064138-45064138 on Build GRCh38
rs945400042 Chr.12: 45064198-45064198 on Build GRCh38
rs1033828681 Chr.12: 45064087-45064087 on Build GRCh38
rs904182161 Chr.12: 45064007-45064007 on Build GRCh38
rs901844586 Chr.12: 45064016-45064016 on Build GRCh38
rs557435744 Chr.12: 45064201-45064201 on Build GRCh38
rs1042900990 Chr.12: 45064065-45064065 on Build GRCh38
rs557766064 Chr.12: 45063854-45063854 on Build GRCh38
rs531464427 Chr.12: 45064119-45064119 on Build GRCh38
rs188649645 Chr.12: 45063955-45063955 on Build GRCh38
rs777066463 Chr.12: 45063973-45063973 on Build GRCh38
rs983995400 Chr.12: 45063848-45063848 on Build GRCh38
rs943228203 Chr.12: 45064191-45064191 on Build GRCh38
rs1044207319 Chr.12: 45063984-45063984 on Build GRCh38
rs184695924 Chr.12: 45063856-45063856 on Build GRCh38
rs147654499 Chr.12: 45063774-45063774 on Build GRCh38
rs866088194 Chr.12: 45064052-45064052 on Build GRCh38
rs371034350 Chr.12: 45064104-45064104 on Build GRCh38
rs544907705 Chr.12: 45063857-45063857 on Build GRCh38
rs565762611 Chr.12: 45064183-45064183 on Build GRCh38
rs999872570 Chr.12: 45064053-45064053 on Build GRCh38

View Full Product Details