Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 24928397-24928974 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPTE2P1
Entrez Gene ID
646405
Gene Name
transmembrane phosphoinositide 3-phosphatase and tensin homolog 2 pseudogene 1
Gene Aliases
-
UniGene
Hs.620592
Gene Chromosome Location
Chr.13: 24928755-24968469 on Build GRCh38
Transcripts
NR_026730.1

SNP Detail

SNP ID Chromosome Location
rs1016682778 Chr.13: 24928584-24928584 on Build GRCh38
rs915841718 Chr.13: 24928945-24928945 on Build GRCh38
rs539046106 Chr.13: 24928777-24928777 on Build GRCh38
rs535806122 Chr.13: 24928614-24928614 on Build GRCh38
rs780628564 Chr.13: 24928882-24928882 on Build GRCh38
rs574731583 Chr.13: 24928682-24928682 on Build GRCh38
rs547063069 Chr.13: 24928492-24928492 on Build GRCh38
rs146951712 Chr.13: 24928681-24928681 on Build GRCh38
rs531806860 Chr.13: 24928818-24928818 on Build GRCh38
rs987613396 Chr.13: 24928727-24928727 on Build GRCh38
rs541789261 Chr.13: 24928815-24928815 on Build GRCh38
rs926045857 Chr.13: 24928807-24928807 on Build GRCh38
rs575364628 Chr.13: 24928828-24928828 on Build GRCh38
rs759393458 Chr.13: 24928429-24928429 on Build GRCh38
rs567161221 Chr.13: 24928524-24928524 on Build GRCh38
rs2863955 Chr.13: 24928710-24928710 on Build GRCh38
rs1029224860 Chr.13: 24928689-24928689 on Build GRCh38
rs573079508 Chr.13: 24928805-24928805 on Build GRCh38
rs549532053 Chr.13: 24928650-24928650 on Build GRCh38
rs1014111988 Chr.13: 24928523-24928523 on Build GRCh38
rs71431717 Chr.13: 24928809-24928809 on Build GRCh38
rs1041721497 Chr.13: 24928444-24928444 on Build GRCh38
rs561443183 Chr.13: 24928745-24928747 on Build GRCh38
rs767361564 Chr.13: 24928478-24928478 on Build GRCh38
rs189762376 Chr.13: 24928792-24928792 on Build GRCh38
rs962727954 Chr.13: 24928666-24928666 on Build GRCh38
rs561893209 Chr.13: 24928822-24928822 on Build GRCh38
rs576380745 Chr.13: 24928723-24928723 on Build GRCh38
rs796741794 Chr.13: 24928509-24928509 on Build GRCh38
rs2359101 Chr.13: 24928480-24928480 on Build GRCh38
rs541329421 Chr.13: 24928597-24928597 on Build GRCh38
rs946216496 Chr.13: 24928766-24928766 on Build GRCh38
rs550818022 Chr.13: 24928452-24928452 on Build GRCh38
rs533375147 Chr.13: 24928491-24928491 on Build GRCh38
rs777384612 Chr.13: 24928531-24928531 on Build GRCh38
rs111500154 Chr.13: 24928840-24928840 on Build GRCh38
rs1004333030 Chr.13: 24928540-24928540 on Build GRCh38
rs894377096 Chr.13: 24928518-24928518 on Build GRCh38

View Full Product Details