Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 24933007-24933558 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPTE2P1
Entrez Gene ID
646405
Gene Name
transmembrane phosphoinositide 3-phosphatase and tensin homolog 2 pseudogene 1
Gene Aliases
-
UniGene
Hs.620592
Gene Chromosome Location
Chr.13: 24928755-24968469 on Build GRCh38
Transcripts
NR_026730.1

SNP Detail

SNP ID Chromosome Location
rs200967936 Chr.13: 24933360-24933360 on Build GRCh38
rs1033756749 Chr.13: 24933296-24933296 on Build GRCh38
rs1010131051 Chr.13: 24933063-24933063 on Build GRCh38
rs536451098 Chr.13: 24933088-24933088 on Build GRCh38
rs191559226 Chr.13: 24933346-24933346 on Build GRCh38
rs745515481 Chr.13: 24933460-24933460 on Build GRCh38
rs762764295 Chr.13: 24933309-24933309 on Build GRCh38
rs146934719 Chr.13: 24933102-24933102 on Build GRCh38
rs184877254 Chr.13: 24933390-24933390 on Build GRCh38
rs901548945 Chr.13: 24933199-24933199 on Build GRCh38
rs541768809 Chr.13: 24933322-24933322 on Build GRCh38
rs778523286 Chr.13: 24933454-24933454 on Build GRCh38
rs974491182 Chr.13: 24933357-24933357 on Build GRCh38
rs945573122 Chr.13: 24933445-24933445 on Build GRCh38
rs7997578 Chr.13: 24933225-24933225 on Build GRCh38
rs903942156 Chr.13: 24933264-24933264 on Build GRCh38
rs565884649 Chr.13: 24933511-24933511 on Build GRCh38
rs908980569 Chr.13: 24933175-24933175 on Build GRCh38
rs1002360187 Chr.13: 24933478-24933478 on Build GRCh38
rs59347578 Chr.13: 24933214-24933214 on Build GRCh38
rs757455058 Chr.13: 24933069-24933069 on Build GRCh38
rs35642281 Chr.13: 24933284-24933285 on Build GRCh38
rs377730738 Chr.13: 24933519-24933519 on Build GRCh38
rs879735916 Chr.13: 24933385-24933385 on Build GRCh38
rs768520771 Chr.13: 24933133-24933133 on Build GRCh38
rs745916453 Chr.13: 24933123-24933123 on Build GRCh38
rs567311135 Chr.13: 24933062-24933062 on Build GRCh38
rs114460876 Chr.13: 24933439-24933439 on Build GRCh38
rs1026250335 Chr.13: 24933333-24933337 on Build GRCh38
rs909088331 Chr.13: 24933348-24933348 on Build GRCh38
rs914165575 Chr.13: 24933442-24933442 on Build GRCh38
rs1022405617 Chr.13: 24933072-24933072 on Build GRCh38
rs1055241770 Chr.13: 24933485-24933485 on Build GRCh38
rs770679970 Chr.13: 24933506-24933506 on Build GRCh38
rs564842950 Chr.13: 24933269-24933269 on Build GRCh38
rs569535010 Chr.13: 24933419-24933419 on Build GRCh38
rs1026700967 Chr.13: 24933521-24933521 on Build GRCh38
rs950665427 Chr.13: 24933344-24933344 on Build GRCh38
rs922996320 Chr.13: 24933378-24933378 on Build GRCh38
rs547688181 Chr.13: 24933061-24933061 on Build GRCh38
rs919158238 Chr.13: 24933139-24933139 on Build GRCh38
rs115158150 Chr.13: 24933296-24933296 on Build GRCh38
rs192564893 Chr.13: 24933177-24933177 on Build GRCh38
rs984566718 Chr.13: 24933168-24933168 on Build GRCh38
rs189632123 Chr.13: 24933486-24933486 on Build GRCh38
rs7995729 Chr.13: 24933351-24933351 on Build GRCh38
rs1002328998 Chr.13: 24933281-24933281 on Build GRCh38
rs7997570 Chr.13: 24933216-24933216 on Build GRCh38
rs8002258 Chr.13: 24933201-24933201 on Build GRCh38
rs904056236 Chr.13: 24933459-24933459 on Build GRCh38
rs527294051 Chr.13: 24933278-24933278 on Build GRCh38
rs576185342 Chr.13: 24933254-24933254 on Build GRCh38
rs933020200 Chr.13: 24933329-24933339 on Build GRCh38
rs558499505 Chr.13: 24933178-24933178 on Build GRCh38
rs137981065 Chr.13: 24933105-24933105 on Build GRCh38
rs112685128 Chr.13: 24933058-24933058 on Build GRCh38
rs929180970 Chr.13: 24933147-24933147 on Build GRCh38
rs1044000202 Chr.13: 24933456-24933456 on Build GRCh38
rs553433584 Chr.13: 24933515-24933515 on Build GRCh38
rs545148960 Chr.13: 24933266-24933266 on Build GRCh38
rs968093842 Chr.13: 24933079-24933079 on Build GRCh38
rs372207739 Chr.13: 24933117-24933117 on Build GRCh38
rs184946880 Chr.13: 24933226-24933226 on Build GRCh38
rs142064886 Chr.13: 24933219-24933219 on Build GRCh38

View Full Product Details