Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 40912474-40913023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUGT1P3
Entrez Gene ID
283507
Gene Name
SGT1 homolog, MIS12 kinetochore complex assembly cochaperone pseudogene 3
Gene Aliases
SUGT1L1
UniGene
Hs.442781
Gene Chromosome Location
Chr.13: 40911887-40921774 on Build GRCh38
Transcripts
NR_003365.2 NR_038258.1 NR_038259.1

Gene Symbol
TPTE2P5
Entrez Gene ID
100616668
Gene Name
transmembrane phosphoinositide 3-phosphatase and tensin homolog 2 pseudogene 5
Gene Aliases
-
UniGene
Hs.660718
Gene Chromosome Location
Chr.13: 40796985-40921750 on Build GRCh38
Transcripts
NR_003365.2 NR_038258.1 NR_038259.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7326679 c.97-3_97-2insT p.? Chr.13: 40912725-40912726 on Build GRCh38
5362024 c.97-3delT p.? Chr.13: 40912726-40912726 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752547514 Chr.13: 40912596-40912596 on Build GRCh38
rs780792197 Chr.13: 40912652-40912652 on Build GRCh38
rs777808638 Chr.13: 40912600-40912600 on Build GRCh38
rs575279380 Chr.13: 40912647-40912647 on Build GRCh38
rs540490094 Chr.13: 40912719-40912719 on Build GRCh38
rs78692708 Chr.13: 40912894-40912894 on Build GRCh38
rs944325802 Chr.13: 40912621-40912621 on Build GRCh38
rs773560208 Chr.13: 40912570-40912570 on Build GRCh38
rs573355351 Chr.13: 40912725-40912726 on Build GRCh38
rs183227568 Chr.13: 40912585-40912585 on Build GRCh38
rs537839168 Chr.13: 40912511-40912511 on Build GRCh38
rs558286155 Chr.13: 40912606-40912606 on Build GRCh38
rs568087479 Chr.13: 40912904-40912904 on Build GRCh38
rs1023630467 Chr.13: 40912792-40912792 on Build GRCh38
rs186578885 Chr.13: 40912685-40912685 on Build GRCh38
rs749370400 Chr.13: 40912615-40912615 on Build GRCh38
rs139824649 Chr.13: 40912513-40912513 on Build GRCh38
rs745376758 Chr.13: 40912666-40912666 on Build GRCh38
rs752320572 Chr.13: 40912751-40912751 on Build GRCh38
rs7989416 Chr.13: 40912850-40912850 on Build GRCh38
rs397963878 Chr.13: 40912907-40912908 on Build GRCh38
rs777562661 Chr.13: 40912514-40912514 on Build GRCh38
rs768188149 Chr.13: 40912561-40912561 on Build GRCh38
rs113004200 Chr.13: 40912730-40912730 on Build GRCh38
rs773575043 Chr.13: 40912705-40912705 on Build GRCh38
rs866300711 Chr.13: 40912949-40912949 on Build GRCh38
rs60699354 Chr.13: 40912826-40912826 on Build GRCh38
rs779999818 Chr.13: 40912612-40912612 on Build GRCh38
rs76185273 Chr.13: 40912893-40912893 on Build GRCh38
rs769054496 Chr.13: 40912632-40912632 on Build GRCh38
rs190079569 Chr.13: 40912556-40912556 on Build GRCh38
rs778416224 Chr.13: 40912893-40912894 on Build GRCh38
rs568109019 Chr.13: 40912523-40912523 on Build GRCh38
rs748148168 Chr.13: 40912926-40912926 on Build GRCh38
rs765826796 Chr.13: 40912575-40912575 on Build GRCh38
rs568903934 Chr.13: 40912503-40912503 on Build GRCh38
rs768929846 Chr.13: 40912533-40912533 on Build GRCh38
rs1013534511 Chr.13: 40912762-40912762 on Build GRCh38
rs912822487 Chr.13: 40912618-40912618 on Build GRCh38
rs761203855 Chr.13: 40912572-40912572 on Build GRCh38
rs771405001 Chr.13: 40912716-40912716 on Build GRCh38
rs772352918 Chr.13: 40912690-40912690 on Build GRCh38
rs191419969 Chr.13: 40912686-40912686 on Build GRCh38
rs944275783 Chr.13: 40912942-40912942 on Build GRCh38
rs552930202 Chr.13: 40912804-40912804 on Build GRCh38
rs982904272 Chr.13: 40912902-40912902 on Build GRCh38
rs59732089 Chr.13: 40912894-40912895 on Build GRCh38
rs192607788 Chr.13: 40912847-40912847 on Build GRCh38
rs764865028 Chr.13: 40912590-40912590 on Build GRCh38
rs533752967 Chr.13: 40912525-40912525 on Build GRCh38
rs1040014096 Chr.13: 40912634-40912634 on Build GRCh38
rs759034796 Chr.13: 40912726-40912726 on Build GRCh38
rs761100554 Chr.13: 40912712-40912712 on Build GRCh38
rs750924409 Chr.13: 40912615-40912624 on Build GRCh38
rs758170734 Chr.13: 40912599-40912599 on Build GRCh38
rs35843233 Chr.13: 40912607-40912608 on Build GRCh38
rs757157925 Chr.13: 40912609-40912609 on Build GRCh38
rs183737163 Chr.13: 40912717-40912717 on Build GRCh38
rs746012993 Chr.13: 40912518-40912518 on Build GRCh38
rs34071777 Chr.13: 40912764-40912764 on Build GRCh38
rs61189737 Chr.13: 40912934-40912934 on Build GRCh38
rs556019848 Chr.13: 40912588-40912588 on Build GRCh38
rs757322724 Chr.13: 40912646-40912646 on Build GRCh38
rs749482410 Chr.13: 40912527-40912527 on Build GRCh38
rs577078622 Chr.13: 40912557-40912557 on Build GRCh38
rs758473144 Chr.13: 40912726-40912726 on Build GRCh38

View Full Product Details