Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 22682009-22681449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WHAMMP3
Entrez Gene ID
339005
Gene Name
WAS protein homolog associated with actin, golgi membranes and microtubules pseudogene 3
Gene Aliases
WHAMML1, WHDC1L1
UniGene
Hs.212670
Gene Chromosome Location
Chr.15: 22664739-22685368 on Build GRCh38
Transcripts
NR_003521.1

View Full Product Details