Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 77916508-77917052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC645752
Entrez Gene ID
645752
Gene Name
golgin A6 family member A pseudogene
Gene Aliases
-
UniGene
Hs.546408
Gene Chromosome Location
Chr.15: 77914217-77926846 on Build GRCh38
Transcripts
NR_027024.1

SNP Detail

SNP ID Chromosome Location
rs550626024 Chr.15: 77916929-77916929 on Build GRCh38
rs948601990 Chr.15: 77916850-77916850 on Build GRCh38
rs764026248 Chr.15: 77916599-77916599 on Build GRCh38
rs376290007 Chr.15: 77916774-77916774 on Build GRCh38
rs767345155 Chr.15: 77916629-77916629 on Build GRCh38
rs761729064 Chr.15: 77916620-77916620 on Build GRCh38
rs746151049 Chr.15: 77916559-77916559 on Build GRCh38
rs759274787 Chr.15: 77916590-77916590 on Build GRCh38
rs760113708 Chr.15: 77916574-77916576 on Build GRCh38
rs1030300087 Chr.15: 77916945-77916945 on Build GRCh38
rs753963276 Chr.15: 77916635-77916635 on Build GRCh38
rs562056746 Chr.15: 77916899-77916899 on Build GRCh38
rs546867622 Chr.15: 77917008-77917008 on Build GRCh38
rs955108613 Chr.15: 77916739-77916739 on Build GRCh38
rs772632321 Chr.15: 77916586-77916586 on Build GRCh38
rs56290535 Chr.15: 77916574-77916574 on Build GRCh38
rs748723209 Chr.15: 77916985-77916985 on Build GRCh38
rs775993651 Chr.15: 77916547-77916549 on Build GRCh38
rs202106786 Chr.15: 77916638-77916638 on Build GRCh38
rs941736736 Chr.15: 77916546-77916546 on Build GRCh38
rs184660322 Chr.15: 77916873-77916873 on Build GRCh38
rs748527136 Chr.15: 77916580-77916580 on Build GRCh38
rs535231126 Chr.15: 77917006-77917006 on Build GRCh38
rs528406680 Chr.15: 77916534-77916534 on Build GRCh38
rs762217063 Chr.15: 77916532-77916532 on Build GRCh38
rs758751690 Chr.15: 77916734-77916735 on Build GRCh38
rs7161792 Chr.15: 77916789-77916789 on Build GRCh38
rs1044325081 Chr.15: 77916862-77916862 on Build GRCh38
rs200501542 Chr.15: 77916656-77916656 on Build GRCh38
rs1008007188 Chr.15: 77916966-77916966 on Build GRCh38
rs1052971309 Chr.15: 77916904-77916904 on Build GRCh38
rs528563814 Chr.15: 77916801-77916801 on Build GRCh38
rs149416952 Chr.15: 77916738-77916738 on Build GRCh38
rs373808485 Chr.15: 77916713-77916713 on Build GRCh38
rs973581984 Chr.15: 77916773-77916773 on Build GRCh38
rs929085707 Chr.15: 77916813-77916813 on Build GRCh38
rs752117696 Chr.15: 77916540-77916540 on Build GRCh38
rs780965277 Chr.15: 77916554-77916554 on Build GRCh38
rs780219060 Chr.15: 77916565-77916565 on Build GRCh38
rs758393574 Chr.15: 77916541-77916541 on Build GRCh38
rs775089553 Chr.15: 77916595-77916595 on Build GRCh38
rs904356576 Chr.15: 77916660-77916660 on Build GRCh38
rs772675666 Chr.15: 77916988-77916988 on Build GRCh38
rs528882104 Chr.15: 77916588-77916588 on Build GRCh38
rs573013792 Chr.15: 77916854-77916854 on Build GRCh38
rs756263782 Chr.15: 77916560-77916560 on Build GRCh38
rs935845075 Chr.15: 77916900-77916900 on Build GRCh38
rs1001320495 Chr.15: 77916667-77916667 on Build GRCh38
rs756983006 Chr.15: 77916553-77916553 on Build GRCh38
rs751371503 Chr.15: 77916549-77916549 on Build GRCh38
rs1018155737 Chr.15: 77916750-77916750 on Build GRCh38
rs192671766 Chr.15: 77916674-77916674 on Build GRCh38
rs553633208 Chr.15: 77916778-77916778 on Build GRCh38
rs539520919 Chr.15: 77916645-77916645 on Build GRCh38
rs529200969 Chr.15: 77916912-77916912 on Build GRCh38
rs762665192 Chr.15: 77917035-77917035 on Build GRCh38
rs956812684 Chr.15: 77916718-77916718 on Build GRCh38
rs963572023 Chr.15: 77916761-77916761 on Build GRCh38
rs751567245 Chr.15: 77916608-77916608 on Build GRCh38
rs919044112 Chr.15: 77916557-77916557 on Build GRCh38
rs575361782 Chr.15: 77916843-77916843 on Build GRCh38
rs749285000 Chr.15: 77916571-77916571 on Build GRCh38
rs780088428 Chr.15: 77916634-77916634 on Build GRCh38
rs1044599033 Chr.15: 77916657-77916657 on Build GRCh38
rs4473151 Chr.15: 77916847-77916847 on Build GRCh38
rs566398985 Chr.15: 77916990-77916990 on Build GRCh38
rs551524104 Chr.15: 77916970-77916970 on Build GRCh38
rs562455559 Chr.15: 77916938-77916938 on Build GRCh38
rs747289805 Chr.15: 77916999-77916999 on Build GRCh38
rs766141001 Chr.15: 77916533-77916533 on Build GRCh38
rs866435972 Chr.15: 77916886-77916886 on Build GRCh38
rs564387301 Chr.15: 77916849-77916849 on Build GRCh38
rs7182575 Chr.15: 77916538-77916538 on Build GRCh38
rs1009560045 Chr.15: 77916731-77916731 on Build GRCh38
rs201365798 Chr.15: 77916637-77916637 on Build GRCh38
rs550588407 Chr.15: 77916630-77916630 on Build GRCh38
rs762314447 Chr.15: 77916598-77916598 on Build GRCh38
rs369955605 Chr.15: 77916701-77916701 on Build GRCh38
rs556931897 Chr.15: 77916824-77916824 on Build GRCh38
rs774939377 Chr.15: 77916576-77916576 on Build GRCh38
rs756387714 Chr.15: 77916633-77916633 on Build GRCh38
rs779130018 Chr.15: 77916977-77916977 on Build GRCh38
rs973528134 Chr.15: 77917013-77917013 on Build GRCh38
rs188082555 Chr.15: 77916642-77916642 on Build GRCh38
rs7162136 Chr.15: 77916965-77916965 on Build GRCh38
rs763890980 Chr.15: 77916545-77916545 on Build GRCh38
rs992424650 Chr.15: 77916823-77916823 on Build GRCh38
rs534878874 Chr.15: 77916744-77916744 on Build GRCh38

View Full Product Details