Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 27425089-27425629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBC1D3P5
Entrez Gene ID
440419
Gene Name
TBC1 domain family member 3 pseudogene 5
Gene Aliases
-
UniGene
Hs.385755
Gene Chromosome Location
Chr.17: 27418005-27431623 on Build GRCh38
Transcripts
NR_033892.1

SNP Detail

SNP ID Chromosome Location
rs562557955 Chr.17: 27425285-27425285 on Build GRCh38
rs535595869 Chr.17: 27425435-27425435 on Build GRCh38
rs935023090 Chr.17: 27425376-27425376 on Build GRCh38
rs560329158 Chr.17: 27425543-27425543 on Build GRCh38
rs542162504 Chr.17: 27425207-27425207 on Build GRCh38
rs188203014 Chr.17: 27425138-27425138 on Build GRCh38
rs544354081 Chr.17: 27425515-27425515 on Build GRCh38
rs564835371 Chr.17: 27425480-27425480 on Build GRCh38
rs531924103 Chr.17: 27425582-27425582 on Build GRCh38
rs1027617096 Chr.17: 27425237-27425237 on Build GRCh38
rs544762525 Chr.17: 27425182-27425182 on Build GRCh38
rs1004864123 Chr.17: 27425482-27425482 on Build GRCh38
rs571208008 Chr.17: 27425359-27425359 on Build GRCh38
rs901806959 Chr.17: 27425605-27425605 on Build GRCh38
rs181164845 Chr.17: 27425253-27425253 on Build GRCh38
rs564872150 Chr.17: 27425249-27425249 on Build GRCh38
rs550318813 Chr.17: 27425585-27425585 on Build GRCh38
rs556488072 Chr.17: 27425427-27425427 on Build GRCh38
rs59131437 Chr.17: 27425508-27425508 on Build GRCh38
rs546637902 Chr.17: 27425587-27425587 on Build GRCh38
rs8065612 Chr.17: 27425315-27425315 on Build GRCh38
rs547960773 Chr.17: 27425541-27425541 on Build GRCh38
rs1043429513 Chr.17: 27425131-27425131 on Build GRCh38
rs1014446857 Chr.17: 27425212-27425212 on Build GRCh38
rs574837356 Chr.17: 27425163-27425163 on Build GRCh38
rs560701531 Chr.17: 27425440-27425440 on Build GRCh38
rs976532723 Chr.17: 27425495-27425495 on Build GRCh38
rs574838359 Chr.17: 27425429-27425429 on Build GRCh38
rs140950670 Chr.17: 27425254-27425254 on Build GRCh38
rs578079453 Chr.17: 27425143-27425143 on Build GRCh38
rs1013721926 Chr.17: 27425391-27425391 on Build GRCh38
rs560299077 Chr.17: 27425216-27425216 on Build GRCh38
rs970346574 Chr.17: 27425227-27425227 on Build GRCh38
rs529926562 Chr.17: 27425540-27425540 on Build GRCh38
rs138095050 Chr.17: 27425573-27425573 on Build GRCh38
rs949168348 Chr.17: 27425558-27425558 on Build GRCh38
rs771674600 Chr.17: 27425452-27425452 on Build GRCh38
rs386796302 Chr.17: 27425315-27425317 on Build GRCh38
rs921732334 Chr.17: 27425304-27425304 on Build GRCh38
rs781712715 Chr.17: 27425181-27425181 on Build GRCh38
rs953226354 Chr.17: 27425521-27425521 on Build GRCh38
rs966509815 Chr.17: 27425272-27425272 on Build GRCh38
rs552203362 Chr.17: 27425562-27425562 on Build GRCh38
rs540117804 Chr.17: 27425242-27425242 on Build GRCh38
rs552893383 Chr.17: 27425348-27425348 on Build GRCh38
rs8079476 Chr.17: 27425307-27425307 on Build GRCh38
rs752472801 Chr.17: 27425493-27425493 on Build GRCh38
rs568461572 Chr.17: 27425162-27425162 on Build GRCh38
rs529666014 Chr.17: 27425455-27425455 on Build GRCh38
rs761844748 Chr.17: 27425536-27425536 on Build GRCh38
rs538480812 Chr.17: 27425158-27425158 on Build GRCh38
rs562557394 Chr.17: 27425528-27425528 on Build GRCh38
rs1014945655 Chr.17: 27425595-27425595 on Build GRCh38
rs572775446 Chr.17: 27425229-27425229 on Build GRCh38
rs966168054 Chr.17: 27425494-27425494 on Build GRCh38
rs545836199 Chr.17: 27425461-27425461 on Build GRCh38
rs115464913 Chr.17: 27425438-27425438 on Build GRCh38
rs768628136 Chr.17: 27425484-27425484 on Build GRCh38
rs8079483 Chr.17: 27425317-27425317 on Build GRCh38
rs538393501 Chr.17: 27425385-27425385 on Build GRCh38
rs527274236 Chr.17: 27425560-27425560 on Build GRCh38
rs896263505 Chr.17: 27425388-27425388 on Build GRCh38
rs533805731 Chr.17: 27425332-27425332 on Build GRCh38

View Full Product Details