Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 39066522-39067071 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100131347
Entrez Gene ID
100131347
Gene Name
RAD52 motif containing 1 pseudogene
Gene Aliases
-
UniGene
Hs.676510
Gene Chromosome Location
Chr.17: 39057019-39081451 on Build GRCh38
Transcripts
NR_036551.1 NM_020405.4

Gene Symbol
PLXDC1
Entrez Gene ID
57125
Gene Name
plexin domain containing 1
Gene Aliases
TEM3, TEM7
UniGene
Hs.125036
Gene Chromosome Location
Chr.17: 39063303-39151649 on Build GRCh38
Transcripts
NR_036551.1 NM_020405.4

SNP Detail

SNP ID Chromosome Location
rs78446316 Chr.17: 39067007-39067007 on Build GRCh38
rs953779618 Chr.17: 39066695-39066695 on Build GRCh38
rs191715142 Chr.17: 39067024-39067024 on Build GRCh38
rs1031152479 Chr.17: 39066577-39066577 on Build GRCh38
rs985322222 Chr.17: 39066884-39066884 on Build GRCh38
rs896970329 Chr.17: 39066961-39066961 on Build GRCh38
rs546471458 Chr.17: 39066801-39066801 on Build GRCh38
rs1012300449 Chr.17: 39066863-39066863 on Build GRCh38
rs1056709293 Chr.17: 39067025-39067025 on Build GRCh38
rs1031204917 Chr.17: 39066879-39066879 on Build GRCh38
rs909517100 Chr.17: 39066719-39066719 on Build GRCh38
rs959014505 Chr.17: 39066591-39066591 on Build GRCh38
rs749895429 Chr.17: 39066582-39066588 on Build GRCh38
rs753617267 Chr.17: 39067005-39067005 on Build GRCh38
rs999704641 Chr.17: 39066576-39066576 on Build GRCh38
rs939175268 Chr.17: 39066815-39066815 on Build GRCh38
rs550870573 Chr.17: 39066826-39066826 on Build GRCh38
rs998095352 Chr.17: 39066935-39066935 on Build GRCh38
rs74701941 Chr.17: 39066592-39066592 on Build GRCh38
rs966275244 Chr.17: 39066894-39066894 on Build GRCh38
rs144672613 Chr.17: 39066696-39066696 on Build GRCh38
rs909570971 Chr.17: 39066965-39066965 on Build GRCh38
rs1012442014 Chr.17: 39066608-39066608 on Build GRCh38
rs35067606 Chr.17: 39066581-39066581 on Build GRCh38
rs879657502 Chr.17: 39066590-39066591 on Build GRCh38
rs1029127079 Chr.17: 39066940-39066940 on Build GRCh38
rs149168757 Chr.17: 39066583-39066583 on Build GRCh38
rs962360966 Chr.17: 39066989-39066989 on Build GRCh38
rs569132155 Chr.17: 39066849-39066849 on Build GRCh38
rs985182697 Chr.17: 39066955-39066955 on Build GRCh38
rs145550227 Chr.17: 39066582-39066582 on Build GRCh38
rs551956660 Chr.17: 39066555-39066555 on Build GRCh38
rs557933017 Chr.17: 39066927-39066927 on Build GRCh38
rs924201747 Chr.17: 39066741-39066741 on Build GRCh38
rs535397166 Chr.17: 39066798-39066798 on Build GRCh38
rs939277733 Chr.17: 39066991-39066991 on Build GRCh38
rs941273990 Chr.17: 39066733-39066733 on Build GRCh38
rs867798641 Chr.17: 39066918-39066918 on Build GRCh38
rs895224893 Chr.17: 39066859-39066859 on Build GRCh38
rs999760634 Chr.17: 39066877-39066877 on Build GRCh38
rs1011876681 Chr.17: 39067011-39067011 on Build GRCh38
rs7221482 Chr.17: 39066873-39066873 on Build GRCh38
rs941203746 Chr.17: 39066954-39066954 on Build GRCh38
rs1036934377 Chr.17: 39066734-39066734 on Build GRCh38
rs372388989 Chr.17: 39067032-39067032 on Build GRCh38
rs953940322 Chr.17: 39066943-39066943 on Build GRCh38
rs918499841 Chr.17: 39067018-39067018 on Build GRCh38
rs903840306 Chr.17: 39066874-39066874 on Build GRCh38

View Full Product Details