Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 62273779-62274337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBC1D3P2
Entrez Gene ID
440452
Gene Name
TBC1 domain family member 3 pseudogene 2
Gene Aliases
-
UniGene
Hs.448884
Gene Chromosome Location
Chr.17: 62264705-62275655 on Build GRCh38
Transcripts
NR_027486.1

SNP Detail

SNP ID Chromosome Location
rs763349420 Chr.17: 62273875-62273875 on Build GRCh38
rs759578376 Chr.17: 62273799-62273799 on Build GRCh38
rs117392685 Chr.17: 62273975-62273975 on Build GRCh38
rs201712090 Chr.17: 62274119-62274119 on Build GRCh38
rs574298094 Chr.17: 62274233-62274233 on Build GRCh38
rs765512660 Chr.17: 62274090-62274090 on Build GRCh38
rs1050463044 Chr.17: 62273987-62273987 on Build GRCh38
rs753282652 Chr.17: 62274156-62274156 on Build GRCh38
rs372433930 Chr.17: 62273842-62273842 on Build GRCh38
rs765556517 Chr.17: 62274128-62274128 on Build GRCh38
rs769231425 Chr.17: 62274121-62274121 on Build GRCh38
rs755946186 Chr.17: 62274153-62274153 on Build GRCh38
rs545534024 Chr.17: 62274278-62274278 on Build GRCh38
rs551031190 Chr.17: 62274191-62274191 on Build GRCh38
rs151095051 Chr.17: 62274110-62274110 on Build GRCh38
rs752705237 Chr.17: 62273912-62273912 on Build GRCh38
rs369998274 Chr.17: 62273817-62273817 on Build GRCh38
rs553432497 Chr.17: 62274258-62274258 on Build GRCh38
rs748842424 Chr.17: 62274106-62274106 on Build GRCh38
rs774809341 Chr.17: 62273881-62273881 on Build GRCh38
rs747644247 Chr.17: 62274049-62274049 on Build GRCh38
rs764122206 Chr.17: 62274073-62274073 on Build GRCh38
rs529186161 Chr.17: 62274162-62274162 on Build GRCh38
rs143214345 Chr.17: 62273948-62273948 on Build GRCh38
rs751336311 Chr.17: 62274092-62274092 on Build GRCh38
rs557333903 Chr.17: 62274021-62274021 on Build GRCh38
rs573987066 Chr.17: 62274034-62274034 on Build GRCh38
rs756792769 Chr.17: 62274107-62274107 on Build GRCh38
rs550062082 Chr.17: 62274118-62274118 on Build GRCh38
rs201780334 Chr.17: 62273836-62273836 on Build GRCh38
rs777097249 Chr.17: 62274105-62274105 on Build GRCh38
rs781283141 Chr.17: 62274044-62274044 on Build GRCh38
rs772795489 Chr.17: 62273874-62273874 on Build GRCh38
rs754056324 Chr.17: 62273998-62273998 on Build GRCh38
rs761962135 Chr.17: 62274125-62274125 on Build GRCh38
rs903170356 Chr.17: 62274237-62274237 on Build GRCh38
rs764094461 Chr.17: 62274154-62274154 on Build GRCh38
rs944782722 Chr.17: 62274183-62274183 on Build GRCh38
rs376224220 Chr.17: 62273970-62273978 on Build GRCh38
rs768440781 Chr.17: 62273818-62273820 on Build GRCh38
rs139807984 Chr.17: 62273818-62273818 on Build GRCh38
rs754181706 Chr.17: 62273829-62273830 on Build GRCh38
rs771236838 Chr.17: 62273878-62273878 on Build GRCh38
rs912073293 Chr.17: 62274157-62274157 on Build GRCh38
rs4968502 Chr.17: 62274096-62274096 on Build GRCh38
rs1038379634 Chr.17: 62273954-62273954 on Build GRCh38
rs754890668 Chr.17: 62274095-62274095 on Build GRCh38
rs748849522 Chr.17: 62274061-62274061 on Build GRCh38
rs573247163 Chr.17: 62274277-62274277 on Build GRCh38
rs752558677 Chr.17: 62274152-62274152 on Build GRCh38
rs779284545 Chr.17: 62273884-62273884 on Build GRCh38
rs534438802 Chr.17: 62274188-62274188 on Build GRCh38
rs767545316 Chr.17: 62273801-62273801 on Build GRCh38
rs548611862 Chr.17: 62274171-62274171 on Build GRCh38
rs759370429 Chr.17: 62274142-62274142 on Build GRCh38
rs532949087 Chr.17: 62274109-62274109 on Build GRCh38
rs200110677 Chr.17: 62274072-62274072 on Build GRCh38
rs565194557 Chr.17: 62274293-62274293 on Build GRCh38
rs754907181 Chr.17: 62274043-62274043 on Build GRCh38
rs767378407 Chr.17: 62274151-62274151 on Build GRCh38
rs545497228 Chr.17: 62274074-62274074 on Build GRCh38
rs756105721 Chr.17: 62274100-62274100 on Build GRCh38
rs774639058 Chr.17: 62274065-62274065 on Build GRCh38
rs559778933 Chr.17: 62274070-62274070 on Build GRCh38
rs751426189 Chr.17: 62274035-62274035 on Build GRCh38
rs940925926 Chr.17: 62273953-62273953 on Build GRCh38
rs978427261 Chr.17: 62273819-62273819 on Build GRCh38
rs578092863 Chr.17: 62273976-62273976 on Build GRCh38
rs756270141 Chr.17: 62273816-62273816 on Build GRCh38
rs768024141 Chr.17: 62273896-62273896 on Build GRCh38
rs746984066 Chr.17: 62273837-62273837 on Build GRCh38
rs759987265 Chr.17: 62273889-62273889 on Build GRCh38
rs769232029 Chr.17: 62274050-62274050 on Build GRCh38
rs534405478 Chr.17: 62273927-62273927 on Build GRCh38
rs761895310 Chr.17: 62274088-62274088 on Build GRCh38
rs3928651 Chr.17: 62274242-62274242 on Build GRCh38
rs537207203 Chr.17: 62274213-62274213 on Build GRCh38
rs758187908 Chr.17: 62273826-62273826 on Build GRCh38
rs752796913 Chr.17: 62273811-62273811 on Build GRCh38
rs777448679 Chr.17: 62274057-62274057 on Build GRCh38
rs760812949 Chr.17: 62273924-62273924 on Build GRCh38
rs542996623 Chr.17: 62274069-62274069 on Build GRCh38
rs769425325 Chr.17: 62273850-62273850 on Build GRCh38
rs770638453 Chr.17: 62274064-62274064 on Build GRCh38
rs750230212 Chr.17: 62273821-62273821 on Build GRCh38
rs773614622 Chr.17: 62274140-62274140 on Build GRCh38
rs746027807 Chr.17: 62274125-62274125 on Build GRCh38
rs752719861 Chr.17: 62274097-62274097 on Build GRCh38
rs757571908 Chr.17: 62273848-62273849 on Build GRCh38
rs571823946 Chr.17: 62274212-62274212 on Build GRCh38
rs747435288 Chr.17: 62274120-62274120 on Build GRCh38
rs547093140 Chr.17: 62274179-62274179 on Build GRCh38
rs563393138 Chr.17: 62274123-62274123 on Build GRCh38
rs146681039 Chr.17: 62273986-62273986 on Build GRCh38
rs3928650 Chr.17: 62274185-62274185 on Build GRCh38
rs557637737 Chr.17: 62274230-62274230 on Build GRCh38
rs768749774 Chr.17: 62273838-62273838 on Build GRCh38

View Full Product Details