Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75588320-75588901 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYO15B
Entrez Gene ID
80022
Gene Name
myosin XVB
Gene Aliases
MYO15BP
UniGene
Hs.390817
Gene Chromosome Location
Chr.17: 75587545-75626850 on Build GRCh38
Transcripts
NM_001309242.1 XM_017025120.1 XM_017025121.1 XM_017025122.1 XM_017025123.1 XM_017025124.1 XM_017025125.1 XM_017025126.1 XM_017025127.1 XM_017025128.1 XM_017025141.1 XM_017025142.1 XM_017025144.1

SNP Detail

SNP ID Chromosome Location
rs1033629074 Chr.17: 75588833-75588833 on Build GRCh38
rs577325841 Chr.17: 75588876-75588876 on Build GRCh38
rs372582469 Chr.17: 75588389-75588389 on Build GRCh38
rs878971662 Chr.17: 75588613-75588613 on Build GRCh38
rs917779696 Chr.17: 75588495-75588495 on Build GRCh38
rs959165378 Chr.17: 75588492-75588492 on Build GRCh38
rs879312949 Chr.17: 75588782-75588782 on Build GRCh38
rs1009585536 Chr.17: 75588552-75588552 on Build GRCh38
rs902055941 Chr.17: 75588704-75588704 on Build GRCh38
rs1026308926 Chr.17: 75588415-75588415 on Build GRCh38
rs926666883 Chr.17: 75588600-75588600 on Build GRCh38
rs913749921 Chr.17: 75588411-75588411 on Build GRCh38
rs574463855 Chr.17: 75588721-75588721 on Build GRCh38
rs374149176 Chr.17: 75588805-75588805 on Build GRCh38
rs1033771800 Chr.17: 75588484-75588484 on Build GRCh38
rs867738252 Chr.17: 75588840-75588840 on Build GRCh38
rs996478337 Chr.17: 75588479-75588479 on Build GRCh38
rs988078169 Chr.17: 75588419-75588419 on Build GRCh38
rs889368193 Chr.17: 75588507-75588507 on Build GRCh38
rs1041004984 Chr.17: 75588738-75588738 on Build GRCh38
rs1040583073 Chr.17: 75588393-75588393 on Build GRCh38
rs960595829 Chr.17: 75588606-75588606 on Build GRCh38
rs62089221 Chr.17: 75588744-75588744 on Build GRCh38
rs962494840 Chr.17: 75588564-75588564 on Build GRCh38
rs898080156 Chr.17: 75588741-75588741 on Build GRCh38
rs913555765 Chr.17: 75588467-75588467 on Build GRCh38
rs887862274 Chr.17: 75588749-75588749 on Build GRCh38
rs978447169 Chr.17: 75588529-75588529 on Build GRCh38
rs62089220 Chr.17: 75588588-75588588 on Build GRCh38
rs868529967 Chr.17: 75588710-75588710 on Build GRCh38
rs114717599 Chr.17: 75588771-75588771 on Build GRCh38
rs913836758 Chr.17: 75588637-75588637 on Build GRCh38
rs925016381 Chr.17: 75588713-75588713 on Build GRCh38
rs543640348 Chr.17: 75588406-75588408 on Build GRCh38
rs926509745 Chr.17: 75588395-75588395 on Build GRCh38
rs943732609 Chr.17: 75588735-75588735 on Build GRCh38
rs200226778 Chr.17: 75588737-75588738 on Build GRCh38
rs945161550 Chr.17: 75588697-75588697 on Build GRCh38
rs903605145 Chr.17: 75588472-75588472 on Build GRCh38
rs943816176 Chr.17: 75588671-75588671 on Build GRCh38
rs554278280 Chr.17: 75588715-75588715 on Build GRCh38
rs1028114850 Chr.17: 75588574-75588574 on Build GRCh38
rs568542110 Chr.17: 75588352-75588352 on Build GRCh38
rs1056299003 Chr.17: 75588405-75588405 on Build GRCh38
rs945240786 Chr.17: 75588457-75588457 on Build GRCh38
rs1013706047 Chr.17: 75588778-75588778 on Build GRCh38
rs952024317 Chr.17: 75588387-75588387 on Build GRCh38
rs62089219 Chr.17: 75588584-75588584 on Build GRCh38
rs972587812 Chr.17: 75588353-75588353 on Build GRCh38
rs1007687726 Chr.17: 75588391-75588391 on Build GRCh38
rs993698830 Chr.17: 75588568-75588568 on Build GRCh38
rs866796042 Chr.17: 75588619-75588619 on Build GRCh38
rs890823023 Chr.17: 75588731-75588731 on Build GRCh38
rs992011088 Chr.17: 75588494-75588494 on Build GRCh38
rs1869575 Chr.17: 75588702-75588702 on Build GRCh38
rs939307595 Chr.17: 75588399-75588399 on Build GRCh38
rs879215502 Chr.17: 75588744-75588745 on Build GRCh38
rs994039250 Chr.17: 75588743-75588743 on Build GRCh38
rs1043842293 Chr.17: 75588470-75588470 on Build GRCh38
rs538567128 Chr.17: 75588500-75588500 on Build GRCh38
rs76124218 Chr.17: 75588360-75588360 on Build GRCh38
rs558421429 Chr.17: 75588561-75588561 on Build GRCh38
rs74841633 Chr.17: 75588497-75588497 on Build GRCh38
rs754949877 Chr.17: 75588712-75588712 on Build GRCh38

View Full Product Details