Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 120467-121024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ROCK1P1
Entrez Gene ID
727758
Gene Name
Rho associated coiled-coil containing protein kinase 1 pseudogene 1
Gene Aliases
-
UniGene
Hs.585843
Gene Chromosome Location
Chr.18: 109065-122219 on Build GRCh38
Transcripts
NR_033770.1

SNP Detail

SNP ID Chromosome Location
rs1029516923 Chr.18: 120902-120902 on Build GRCh38
rs993935423 Chr.18: 120555-120555 on Build GRCh38
rs916767189 Chr.18: 120960-120960 on Build GRCh38
rs906168972 Chr.18: 120544-120544 on Build GRCh38
rs535830018 Chr.18: 120994-120994 on Build GRCh38
rs1005788237 Chr.18: 120611-120611 on Build GRCh38
rs14025 Chr.18: 120722-120722 on Build GRCh38
rs2847088 Chr.18: 120879-120879 on Build GRCh38
rs895378941 Chr.18: 120499-120499 on Build GRCh38
rs936887906 Chr.18: 120755-120755 on Build GRCh38
rs2847089 Chr.18: 120714-120714 on Build GRCh38
rs948297893 Chr.18: 120799-120799 on Build GRCh38
rs796525317 Chr.18: 120689-120689 on Build GRCh38
rs888188264 Chr.18: 120868-120869 on Build GRCh38
rs978489794 Chr.18: 120938-120938 on Build GRCh38
rs566730755 Chr.18: 120882-120882 on Build GRCh38
rs534542771 Chr.18: 120899-120899 on Build GRCh38
rs538451002 Chr.18: 120984-120984 on Build GRCh38
rs1025796141 Chr.18: 120600-120600 on Build GRCh38
rs11284195 Chr.18: 120684-120684 on Build GRCh38
rs954688042 Chr.18: 120646-120646 on Build GRCh38
rs775498983 Chr.18: 120795-120795 on Build GRCh38
rs531430198 Chr.18: 120861-120861 on Build GRCh38
rs574546148 Chr.18: 120573-120573 on Build GRCh38
rs559001766 Chr.18: 120958-120958 on Build GRCh38
rs954739755 Chr.18: 120916-120916 on Build GRCh38
rs929694699 Chr.18: 120989-120989 on Build GRCh38
rs976606723 Chr.18: 120639-120639 on Build GRCh38
rs187716944 Chr.18: 120665-120665 on Build GRCh38
rs924089993 Chr.18: 120941-120941 on Build GRCh38
rs906221183 Chr.18: 120831-120831 on Build GRCh38
rs1046707729 Chr.18: 120968-120968 on Build GRCh38
rs116450891 Chr.18: 120881-120881 on Build GRCh38
rs751768719 Chr.18: 120528-120532 on Build GRCh38
rs1054360724 Chr.18: 120774-120774 on Build GRCh38
rs928192067 Chr.18: 120983-120983 on Build GRCh38
rs1018535161 Chr.18: 120876-120876 on Build GRCh38
rs1046658952 Chr.18: 120812-120812 on Build GRCh38
rs562575241 Chr.18: 120501-120501 on Build GRCh38
rs564373517 Chr.18: 120847-120847 on Build GRCh38
rs536334535 Chr.18: 120880-120880 on Build GRCh38
rs8096198 Chr.18: 120645-120645 on Build GRCh38
rs916715850 Chr.18: 120793-120793 on Build GRCh38
rs12967551 Chr.18: 120684-120684 on Build GRCh38
rs552175273 Chr.18: 120846-120846 on Build GRCh38
rs963934488 Chr.18: 120631-120633 on Build GRCh38
rs1045149 Chr.18: 121003-121003 on Build GRCh38
rs577437992 Chr.18: 120966-120966 on Build GRCh38
rs879204599 Chr.18: 120705-120705 on Build GRCh38
rs1018228780 Chr.18: 120622-120622 on Build GRCh38
rs569016965 Chr.18: 120875-120875 on Build GRCh38
rs924008918 Chr.18: 120731-120731 on Build GRCh38
rs771703074 Chr.18: 120574-120574 on Build GRCh38
rs1005264170 Chr.18: 120869-120869 on Build GRCh38
rs183439928 Chr.18: 120606-120606 on Build GRCh38
rs143005099 Chr.18: 120862-120862 on Build GRCh38
rs866348655 Chr.18: 120517-120517 on Build GRCh38

View Full Product Details