Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49975314-49975864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

SNP Detail

SNP ID Chromosome Location
rs190608789 Chr.19: 49975509-49975509 on Build GRCh38
rs767160132 Chr.19: 49975567-49975567 on Build GRCh38
rs12609492 Chr.19: 49975384-49975384 on Build GRCh38
rs1052127989 Chr.19: 49975433-49975433 on Build GRCh38
rs567955523 Chr.19: 49975520-49975520 on Build GRCh38
rs566066434 Chr.19: 49975781-49975781 on Build GRCh38
rs780271912 Chr.19: 49975822-49975822 on Build GRCh38
rs138974185 Chr.19: 49975771-49975771 on Build GRCh38
rs562427319 Chr.19: 49975584-49975584 on Build GRCh38
rs1004986725 Chr.19: 49975605-49975625 on Build GRCh38
rs943947021 Chr.19: 49975832-49975832 on Build GRCh38
rs12609509 Chr.19: 49975527-49975527 on Build GRCh38
rs12611411 Chr.19: 49975675-49975675 on Build GRCh38
rs943978259 Chr.19: 49975546-49975546 on Build GRCh38
rs755219137 Chr.19: 49975835-49975835 on Build GRCh38
rs776008262 Chr.19: 49975505-49975505 on Build GRCh38
rs12984584 Chr.19: 49975821-49975821 on Build GRCh38
rs759221921 Chr.19: 49975510-49975510 on Build GRCh38
rs564644642 Chr.19: 49975669-49975669 on Build GRCh38
rs999275562 Chr.19: 49975592-49975592 on Build GRCh38
rs761042574 Chr.19: 49975580-49975580 on Build GRCh38
rs985239319 Chr.19: 49975810-49975810 on Build GRCh38
rs1023184208 Chr.19: 49975422-49975422 on Build GRCh38
rs370640451 Chr.19: 49975349-49975349 on Build GRCh38
rs547431752 Chr.19: 49975778-49975778 on Build GRCh38
rs12609811 Chr.19: 49975705-49975705 on Build GRCh38
rs535456897 Chr.19: 49975402-49975402 on Build GRCh38
rs778612523 Chr.19: 49975491-49975494 on Build GRCh38
rs961709040 Chr.19: 49975684-49975684 on Build GRCh38
rs969178598 Chr.19: 49975600-49975600 on Build GRCh38
rs931412862 Chr.19: 49975775-49975775 on Build GRCh38
rs775028880 Chr.19: 49975571-49975571 on Build GRCh38
rs554182573 Chr.19: 49975417-49975417 on Build GRCh38
rs539930239 Chr.19: 49975507-49975507 on Build GRCh38
rs1017173137 Chr.19: 49975599-49975599 on Build GRCh38
rs1041395416 Chr.19: 49975836-49975836 on Build GRCh38
rs960629213 Chr.19: 49975656-49975656 on Build GRCh38
rs951528270 Chr.19: 49975594-49975594 on Build GRCh38
rs545919586 Chr.19: 49975640-49975640 on Build GRCh38
rs368620184 Chr.19: 49975702-49975702 on Build GRCh38
rs764853717 Chr.19: 49975519-49975519 on Build GRCh38
rs187937972 Chr.19: 49975348-49975348 on Build GRCh38
rs910304373 Chr.19: 49975792-49975792 on Build GRCh38
rs990398743 Chr.19: 49975415-49975415 on Build GRCh38
rs780118537 Chr.19: 49975816-49975816 on Build GRCh38
rs73576692 Chr.19: 49975608-49975608 on Build GRCh38
rs572436467 Chr.19: 49975486-49975486 on Build GRCh38
rs569068878 Chr.19: 49975609-49975609 on Build GRCh38
rs746114583 Chr.19: 49975402-49975403 on Build GRCh38
rs1043448234 Chr.19: 49975785-49975785 on Build GRCh38
rs919964484 Chr.19: 49975720-49975720 on Build GRCh38
rs940100524 Chr.19: 49975499-49975499 on Build GRCh38
rs928643962 Chr.19: 49975484-49975484 on Build GRCh38
rs79125155 Chr.19: 49975595-49975595 on Build GRCh38
rs867148510 Chr.19: 49975823-49975823 on Build GRCh38
rs541458538 Chr.19: 49975601-49975601 on Build GRCh38
rs769495365 Chr.19: 49975424-49975424 on Build GRCh38
rs981412210 Chr.19: 49975480-49975480 on Build GRCh38
rs904721745 Chr.19: 49975517-49975517 on Build GRCh38
rs571287294 Chr.19: 49975514-49975514 on Build GRCh38
rs773221885 Chr.19: 49975458-49975458 on Build GRCh38
rs913615564 Chr.19: 49975436-49975436 on Build GRCh38
rs1019254815 Chr.19: 49975382-49975382 on Build GRCh38

View Full Product Details