Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 56376760-56377308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF542P
Entrez Gene ID
147947
Gene Name
zinc finger protein 542, pseudogene
Gene Aliases
ZNF542
UniGene
Hs.467326
Gene Chromosome Location
Chr.19: 56368099-56379828 on Build GRCh38
Transcripts
NR_003127.2 NR_024055.2 NR_024056.2 NR_024057.2 NR_033418.1

SNP Detail

SNP ID Chromosome Location
rs977121060 Chr.19: 56377166-56377166 on Build GRCh38
rs548272683 Chr.19: 56377253-56377253 on Build GRCh38
rs953316505 Chr.19: 56376931-56376931 on Build GRCh38
rs992958793 Chr.19: 56377191-56377191 on Build GRCh38
rs752130978 Chr.19: 56377076-56377076 on Build GRCh38
rs770358524 Chr.19: 56376953-56376953 on Build GRCh38
rs553394689 Chr.19: 56376978-56376978 on Build GRCh38
rs371596354 Chr.19: 56377266-56377266 on Build GRCh38
rs565324335 Chr.19: 56377055-56377055 on Build GRCh38
rs758374424 Chr.19: 56377180-56377180 on Build GRCh38
rs751270871 Chr.19: 56377137-56377137 on Build GRCh38
rs751892242 Chr.19: 56376990-56376990 on Build GRCh38
rs151225539 Chr.19: 56376983-56376983 on Build GRCh38
rs915645527 Chr.19: 56377217-56377217 on Build GRCh38
rs116797792 Chr.19: 56376929-56376929 on Build GRCh38
rs546707959 Chr.19: 56377115-56377116 on Build GRCh38
rs1012517043 Chr.19: 56377069-56377069 on Build GRCh38
rs199975898 Chr.19: 56376848-56376849 on Build GRCh38
rs938394715 Chr.19: 56377027-56377027 on Build GRCh38
rs989338217 Chr.19: 56377030-56377030 on Build GRCh38
rs767799961 Chr.19: 56377160-56377160 on Build GRCh38
rs2304126 Chr.19: 56377157-56377157 on Build GRCh38
rs999179794 Chr.19: 56377213-56377213 on Build GRCh38
rs186663695 Chr.19: 56377112-56377112 on Build GRCh38
rs761903763 Chr.19: 56377158-56377158 on Build GRCh38
rs751530808 Chr.19: 56376800-56376800 on Build GRCh38
rs1016392873 Chr.19: 56376901-56376902 on Build GRCh38
rs781364065 Chr.19: 56377088-56377088 on Build GRCh38
rs753515776 Chr.19: 56377172-56377172 on Build GRCh38
rs1005763125 Chr.19: 56376898-56376898 on Build GRCh38
rs12185527 Chr.19: 56377186-56377186 on Build GRCh38
rs2304125 Chr.19: 56377085-56377085 on Build GRCh38
rs897913445 Chr.19: 56377102-56377102 on Build GRCh38
rs1030179873 Chr.19: 56377119-56377119 on Build GRCh38
rs768911205 Chr.19: 56377153-56377153 on Build GRCh38
rs2304124 Chr.19: 56377081-56377081 on Build GRCh38
rs975950461 Chr.19: 56376927-56376927 on Build GRCh38
rs1015050346 Chr.19: 56376896-56376896 on Build GRCh38
rs536464742 Chr.19: 56376954-56376954 on Build GRCh38
rs1023659800 Chr.19: 56376835-56376835 on Build GRCh38
rs1015306276 Chr.19: 56377090-56377090 on Build GRCh38
rs923897823 Chr.19: 56376977-56376977 on Build GRCh38
rs374673565 Chr.19: 56376856-56376856 on Build GRCh38
rs754820405 Chr.19: 56377048-56377048 on Build GRCh38
rs759375286 Chr.19: 56377174-56377174 on Build GRCh38
rs1006208005 Chr.19: 56376853-56376853 on Build GRCh38
rs367644769 Chr.19: 56377284-56377285 on Build GRCh38
rs541712881 Chr.19: 56377018-56377018 on Build GRCh38
rs373652021 Chr.19: 56377209-56377209 on Build GRCh38
rs559008583 Chr.19: 56377144-56377144 on Build GRCh38
rs888598096 Chr.19: 56376897-56376897 on Build GRCh38
rs191113370 Chr.19: 56377125-56377125 on Build GRCh38
rs977130290 Chr.19: 56377057-56377057 on Build GRCh38
rs777389199 Chr.19: 56377079-56377079 on Build GRCh38
rs1012464436 Chr.19: 56376830-56376830 on Build GRCh38
rs33949495 Chr.19: 56377281-56377281 on Build GRCh38
rs953245131 Chr.19: 56377132-56377132 on Build GRCh38
rs574139249 Chr.19: 56376948-56376948 on Build GRCh38
rs562264261 Chr.19: 56376962-56376962 on Build GRCh38
rs753660826 Chr.19: 56377077-56377077 on Build GRCh38
rs952054412 Chr.19: 56376845-56376845 on Build GRCh38
rs4801719 Chr.19: 56377183-56377183 on Build GRCh38
rs760500782 Chr.19: 56376994-56376994 on Build GRCh38
rs564791573 Chr.19: 56377041-56377041 on Build GRCh38
rs922584713 Chr.19: 56377060-56377060 on Build GRCh38
rs959153786 Chr.19: 56376924-56376924 on Build GRCh38
rs780730166 Chr.19: 56377070-56377070 on Build GRCh38
rs776207327 Chr.19: 56377114-56377114 on Build GRCh38
rs193113066 Chr.19: 56376920-56376920 on Build GRCh38
rs78332918 Chr.19: 56376873-56376873 on Build GRCh38
rs140409785 Chr.19: 56377139-56377139 on Build GRCh38
rs571588747 Chr.19: 56376872-56376872 on Build GRCh38
rs183939266 Chr.19: 56377008-56377008 on Build GRCh38
rs377203920 Chr.19: 56377113-56377113 on Build GRCh38
rs746259049 Chr.19: 56377095-56377095 on Build GRCh38

View Full Product Details