Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 87055746-87056267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC285074
Entrez Gene ID
285074
Gene Name
anaphase promoting complex subunit 1 pseudogene
Gene Aliases
-
UniGene
Hs.424184
Gene Chromosome Location
Chr.2: 87030675-87076413 on Build GRCh38
Transcripts
NR_026846.1

SNP Detail

SNP ID Chromosome Location
rs201342610 Chr.2: 87056041-87056043 on Build GRCh38
rs907150160 Chr.2: 87056101-87056101 on Build GRCh38
rs75961009 Chr.2: 87055770-87055770 on Build GRCh38
rs1011332166 Chr.2: 87056188-87056188 on Build GRCh38
rs1046996056 Chr.2: 87056018-87056018 on Build GRCh38
rs867473917 Chr.2: 87056244-87056244 on Build GRCh38
rs538325576 Chr.2: 87055971-87055971 on Build GRCh38
rs570906843 Chr.2: 87055916-87055916 on Build GRCh38
rs942816649 Chr.2: 87055919-87055919 on Build GRCh38
rs938468689 Chr.2: 87056117-87056117 on Build GRCh38
rs987014984 Chr.2: 87055892-87055892 on Build GRCh38
rs894178706 Chr.2: 87056150-87056150 on Build GRCh38
rs776957772 Chr.2: 87056152-87056152 on Build GRCh38
rs902850971 Chr.2: 87056230-87056230 on Build GRCh38
rs568192518 Chr.2: 87055959-87055959 on Build GRCh38
rs1055651614 Chr.2: 87056123-87056123 on Build GRCh38
rs929999966 Chr.2: 87055980-87055980 on Build GRCh38
rs919931532 Chr.2: 87055978-87055978 on Build GRCh38
rs552718230 Chr.2: 87055855-87055855 on Build GRCh38
rs1043230965 Chr.2: 87056203-87056203 on Build GRCh38
rs911267065 Chr.2: 87055903-87055903 on Build GRCh38

View Full Product Details