Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 91775912-91776358 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGT8P
Entrez Gene ID
645367
Gene Name
gamma-glutamyltransferase 8 pseudogene
Gene Aliases
GGT1P1
UniGene
Hs.650223
Gene Chromosome Location
Chr.2: 91775342-91782127 on Build GRCh38
Transcripts
NR_003503.1

SNP Detail

SNP ID Chromosome Location
rs545603671 Chr.2: 91776099-91776099 on Build GRCh38
rs535325482 Chr.2: 91776057-91776057 on Build GRCh38
rs202152243 Chr.2: 91776004-91776004 on Build GRCh38
rs71425417 Chr.2: 91776159-91776159 on Build GRCh38
rs189211741 Chr.2: 91776096-91776096 on Build GRCh38
rs980227317 Chr.2: 91776003-91776003 on Build GRCh38
rs1020515057 Chr.2: 91776248-91776248 on Build GRCh38
rs967062258 Chr.2: 91776255-91776255 on Build GRCh38
rs1001660507 Chr.2: 91776243-91776243 on Build GRCh38
rs201505853 Chr.2: 91776285-91776285 on Build GRCh38
rs907294463 Chr.2: 91776220-91776220 on Build GRCh38
rs971561810 Chr.2: 91775994-91775994 on Build GRCh38
rs771645906 Chr.2: 91776048-91776048 on Build GRCh38
rs1034896210 Chr.2: 91776244-91776244 on Build GRCh38
rs113372566 Chr.2: 91776287-91776287 on Build GRCh38
rs747965370 Chr.2: 91776312-91776313 on Build GRCh38
rs994182261 Chr.2: 91775977-91775977 on Build GRCh38
rs4547565 Chr.2: 91776221-91776221 on Build GRCh38
rs989842039 Chr.2: 91776024-91776024 on Build GRCh38
rs975855004 Chr.2: 91776068-91776068 on Build GRCh38
rs4552222 Chr.2: 91776194-91776194 on Build GRCh38
rs931683087 Chr.2: 91776271-91776271 on Build GRCh38
rs572921939 Chr.2: 91775978-91775978 on Build GRCh38
rs1017307355 Chr.2: 91775955-91775955 on Build GRCh38
rs532174300 Chr.2: 91776315-91776315 on Build GRCh38
rs909012231 Chr.2: 91776290-91776290 on Build GRCh38
rs113798130 Chr.2: 91776070-91776070 on Build GRCh38
rs912862346 Chr.2: 91776039-91776039 on Build GRCh38
rs941910113 Chr.2: 91776113-91776113 on Build GRCh38
rs1024445990 Chr.2: 91775980-91775980 on Build GRCh38
rs569083836 Chr.2: 91776065-91776065 on Build GRCh38
rs4547564 Chr.2: 91776224-91776224 on Build GRCh38
rs187412858 Chr.2: 91776222-91776222 on Build GRCh38
rs4547566 Chr.2: 91776210-91776210 on Build GRCh38
rs200296792 Chr.2: 91776005-91776005 on Build GRCh38
rs528502768 Chr.2: 91776253-91776253 on Build GRCh38
rs879085815 Chr.2: 91776168-91776168 on Build GRCh38
rs945799029 Chr.2: 91776059-91776059 on Build GRCh38
rs985991803 Chr.2: 91776288-91776288 on Build GRCh38
rs201095403 Chr.2: 91776006-91776006 on Build GRCh38
rs181738693 Chr.2: 91776150-91776150 on Build GRCh38
rs958537658 Chr.2: 91776139-91776139 on Build GRCh38
rs575340535 Chr.2: 91776149-91776149 on Build GRCh38
rs1000434980 Chr.2: 91776029-91776029 on Build GRCh38
rs990783590 Chr.2: 91776246-91776246 on Build GRCh38
rs565208266 Chr.2: 91776270-91776270 on Build GRCh38
rs957656635 Chr.2: 91776245-91776245 on Build GRCh38
rs767656542 Chr.2: 91775946-91775948 on Build GRCh38
rs4496366 Chr.2: 91776018-91776018 on Build GRCh38
rs563594631 Chr.2: 91776135-91776135 on Build GRCh38
rs868859192 Chr.2: 91776040-91776040 on Build GRCh38
rs558415161 Chr.2: 91775971-91775971 on Build GRCh38
rs554608054 Chr.2: 91775943-91775943 on Build GRCh38
rs1034148058 Chr.2: 91776117-91776117 on Build GRCh38
rs546558725 Chr.2: 91776266-91776266 on Build GRCh38
rs1024372536 Chr.2: 91776219-91776219 on Build GRCh38
rs957817721 Chr.2: 91776016-91776016 on Build GRCh38
rs897244976 Chr.2: 91775974-91775974 on Build GRCh38

View Full Product Details