Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219266418-219266962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TUBA4B
Entrez Gene ID
80086
Gene Name
tubulin alpha 4b
Gene Aliases
TUBA4
UniGene
Hs.664469
Gene Chromosome Location
Chr.2: 219253243-219272188 on Build GRCh38
Transcripts
NR_003063.1

SNP Detail

SNP ID Chromosome Location
rs934762660 Chr.2: 219266635-219266635 on Build GRCh38
rs182768813 Chr.2: 219266745-219266745 on Build GRCh38
rs1021961774 Chr.2: 219266828-219266828 on Build GRCh38
rs561260307 Chr.2: 219266685-219266685 on Build GRCh38
rs187885083 Chr.2: 219266892-219266892 on Build GRCh38
rs771568312 Chr.2: 219266800-219266800 on Build GRCh38
rs1001899535 Chr.2: 219266762-219266762 on Build GRCh38
rs913923558 Chr.2: 219266581-219266581 on Build GRCh38
rs1003308476 Chr.2: 219266791-219266791 on Build GRCh38
rs1007535285 Chr.2: 219266495-219266495 on Build GRCh38
rs959050732 Chr.2: 219266590-219266590 on Build GRCh38
rs796843940 Chr.2: 219266541-219266543 on Build GRCh38
rs899655672 Chr.2: 219266644-219266644 on Build GRCh38
rs966740363 Chr.2: 219266923-219266923 on Build GRCh38
rs1003228287 Chr.2: 219266579-219266579 on Build GRCh38
rs547731179 Chr.2: 219266678-219266678 on Build GRCh38
rs943459427 Chr.2: 219266938-219266938 on Build GRCh38
rs537649950 Chr.2: 219266856-219266856 on Build GRCh38
rs943431125 Chr.2: 219266610-219266610 on Build GRCh38
rs990461807 Chr.2: 219266593-219266593 on Build GRCh38
rs368660073 Chr.2: 219266451-219266451 on Build GRCh38
rs1039592197 Chr.2: 219266638-219266638 on Build GRCh38
rs1004251323 Chr.2: 219266726-219266726 on Build GRCh38
rs950508456 Chr.2: 219266569-219266569 on Build GRCh38
rs1012360208 Chr.2: 219266818-219266818 on Build GRCh38
rs527889280 Chr.2: 219266622-219266622 on Build GRCh38
rs774962573 Chr.2: 219266594-219266594 on Build GRCh38
rs906328577 Chr.2: 219266757-219266758 on Build GRCh38
rs911954931 Chr.2: 219266679-219266679 on Build GRCh38
rs899040234 Chr.2: 219266549-219266549 on Build GRCh38
rs890446984 Chr.2: 219266477-219266477 on Build GRCh38
rs545524283 Chr.2: 219266535-219266535 on Build GRCh38
rs886259013 Chr.2: 219266759-219266759 on Build GRCh38
rs1051790667 Chr.2: 219266476-219266476 on Build GRCh38
rs931269865 Chr.2: 219266659-219266659 on Build GRCh38
rs748850728 Chr.2: 219266559-219266559 on Build GRCh38
rs569076673 Chr.2: 219266598-219266598 on Build GRCh38
rs1038998695 Chr.2: 219266697-219266697 on Build GRCh38
rs924756468 Chr.2: 219266631-219266631 on Build GRCh38
rs1051860866 Chr.2: 219266672-219266672 on Build GRCh38
rs576594193 Chr.2: 219266462-219266462 on Build GRCh38
rs899111515 Chr.2: 219266730-219266730 on Build GRCh38
rs995086071 Chr.2: 219266553-219266553 on Build GRCh38
rs987596352 Chr.2: 219266893-219266893 on Build GRCh38
rs969402251 Chr.2: 219266832-219266832 on Build GRCh38
rs911985512 Chr.2: 219266931-219266931 on Build GRCh38
rs192657625 Chr.2: 219266586-219266586 on Build GRCh38
rs980864562 Chr.2: 219266534-219266534 on Build GRCh38
rs979381291 Chr.2: 219266605-219266605 on Build GRCh38
rs1039380101 Chr.2: 219266548-219266548 on Build GRCh38
rs1033804154 Chr.2: 219266560-219266560 on Build GRCh38
rs979026133 Chr.2: 219266847-219266847 on Build GRCh38
rs894864917 Chr.2: 219266810-219266810 on Build GRCh38
rs958314833 Chr.2: 219266565-219266565 on Build GRCh38
rs1048044604 Chr.2: 219266746-219266746 on Build GRCh38
rs752489073 Chr.2: 219266592-219266592 on Build GRCh38
rs1034736062 Chr.2: 219266799-219266799 on Build GRCh38

View Full Product Details