Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 58232676-58233225 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP4R1L
Entrez Gene ID
55370
Gene Name
protein phosphatase 4 regulatory subunit 1 like (pseudogene)
Gene Aliases
C20orf192, PPP4R1P1, PRO1085
UniGene
Hs.473179
Gene Chromosome Location
Chr.20: 58232777-58309439 on Build GRCh38
Transcripts
NR_003505.3

SNP Detail

SNP ID Chromosome Location
rs942490262 Chr.20: 58233145-58233145 on Build GRCh38
rs1052463801 Chr.20: 58232893-58232893 on Build GRCh38
rs936888368 Chr.20: 58232881-58232881 on Build GRCh38
rs955002091 Chr.20: 58233080-58233080 on Build GRCh38
rs898399952 Chr.20: 58232890-58232890 on Build GRCh38
rs761720634 Chr.20: 58232821-58232821 on Build GRCh38
rs968928904 Chr.20: 58233051-58233051 on Build GRCh38
rs748381639 Chr.20: 58232910-58232910 on Build GRCh38
rs1052197404 Chr.20: 58233156-58233156 on Build GRCh38
rs530661005 Chr.20: 58232730-58232730 on Build GRCh38
rs2274932 Chr.20: 58232736-58232736 on Build GRCh38
rs914107907 Chr.20: 58233131-58233131 on Build GRCh38
rs775551439 Chr.20: 58232930-58232930 on Build GRCh38
rs771198018 Chr.20: 58232790-58232790 on Build GRCh38
rs773160064 Chr.20: 58232991-58232995 on Build GRCh38
rs200608823 Chr.20: 58232737-58232737 on Build GRCh38
rs1025956675 Chr.20: 58233020-58233020 on Build GRCh38
rs182894152 Chr.20: 58232865-58232865 on Build GRCh38
rs6026139 Chr.20: 58233053-58233053 on Build GRCh38
rs768922265 Chr.20: 58233046-58233046 on Build GRCh38
rs917707109 Chr.20: 58233049-58233049 on Build GRCh38
rs925427368 Chr.20: 58233094-58233094 on Build GRCh38
rs3818744 Chr.20: 58232913-58232913 on Build GRCh38
rs535907359 Chr.20: 58232861-58232861 on Build GRCh38
rs991939016 Chr.20: 58232811-58232811 on Build GRCh38
rs889192944 Chr.20: 58232937-58232937 on Build GRCh38
rs969540959 Chr.20: 58233068-58233068 on Build GRCh38
rs372428961 Chr.20: 58232733-58232733 on Build GRCh38
rs367783800 Chr.20: 58232870-58232870 on Build GRCh38
rs568466251 Chr.20: 58232837-58232837 on Build GRCh38
rs1035798089 Chr.20: 58232707-58232707 on Build GRCh38
rs557214647 Chr.20: 58232869-58232869 on Build GRCh38
rs765644842 Chr.20: 58232780-58232780 on Build GRCh38
rs79426246 Chr.20: 58232836-58232836 on Build GRCh38
rs977540828 Chr.20: 58232832-58232832 on Build GRCh38
rs1047026320 Chr.20: 58232912-58232912 on Build GRCh38
rs977612538 Chr.20: 58233054-58233054 on Build GRCh38
rs866776185 Chr.20: 58233151-58233151 on Build GRCh38
rs747142124 Chr.20: 58232882-58232882 on Build GRCh38
rs962632227 Chr.20: 58232987-58232987 on Build GRCh38
rs758130540 Chr.20: 58232720-58232720 on Build GRCh38
rs1006720639 Chr.20: 58232938-58232938 on Build GRCh38
rs762961191 Chr.20: 58233038-58233038 on Build GRCh38
rs781743062 Chr.20: 58232840-58232840 on Build GRCh38
rs114004804 Chr.20: 58232833-58232833 on Build GRCh38
rs980959798 Chr.20: 58233075-58233075 on Build GRCh38
rs201085063 Chr.20: 58232799-58232799 on Build GRCh38
rs539671729 Chr.20: 58233153-58233153 on Build GRCh38
rs936792977 Chr.20: 58233116-58233116 on Build GRCh38
rs774428745 Chr.20: 58233048-58233048 on Build GRCh38
rs914817972 Chr.20: 58232756-58232756 on Build GRCh38
rs917616639 Chr.20: 58232819-58232819 on Build GRCh38
rs892479511 Chr.20: 58232906-58232906 on Build GRCh38
rs1018047008 Chr.20: 58232946-58232946 on Build GRCh38
rs1052690817 Chr.20: 58233117-58233117 on Build GRCh38
rs995397835 Chr.20: 58233016-58233016 on Build GRCh38

View Full Product Details