Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 45072933-45073481 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC105372836
Entrez Gene ID
105372836
Gene Name
uncharacterized LOC105372836
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.21: 45055869-45073903 on Build GRCh38
Transcripts
XM_011529812.2 NR_027292.2

Gene Symbol
SSR4P1
Entrez Gene ID
728039
Gene Name
signal sequence receptor subunit 4 pseudogene 1
Gene Aliases
C21orf122, PRED57, PRED90
UniGene
Hs.309203
Gene Chromosome Location
Chr.21: 45070955-45073211 on Build GRCh38
Transcripts
XM_011529812.2 NR_027292.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7309961 c.16G>T p.G6C Chr.21: 45072997-45072997 on Build GRCh38
6502291 c.74G>T p.S25I Chr.21: 45072939-45072939 on Build GRCh38
4387537 c.21C>A p.F7L Chr.21: 45072992-45072992 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs950248290 Chr.21: 45073052-45073052 on Build GRCh38
rs1028947009 Chr.21: 45073237-45073237 on Build GRCh38
rs904071236 Chr.21: 45073247-45073247 on Build GRCh38
rs749391843 Chr.21: 45072976-45072976 on Build GRCh38
rs534101654 Chr.21: 45073250-45073250 on Build GRCh38
rs996412306 Chr.21: 45073236-45073236 on Build GRCh38
rs28760601 Chr.21: 45073208-45073208 on Build GRCh38
rs543566160 Chr.21: 45073102-45073102 on Build GRCh38
rs967729150 Chr.21: 45073002-45073002 on Build GRCh38
rs1039827858 Chr.21: 45073170-45073170 on Build GRCh38
rs996243479 Chr.21: 45073407-45073407 on Build GRCh38
rs934072281 Chr.21: 45073404-45073404 on Build GRCh38
rs980524955 Chr.21: 45073047-45073047 on Build GRCh38
rs769740781 Chr.21: 45073256-45073256 on Build GRCh38
rs903671510 Chr.21: 45073422-45073422 on Build GRCh38
rs956394256 Chr.21: 45073450-45073450 on Build GRCh38
rs921244761 Chr.21: 45073171-45073171 on Build GRCh38
rs1056591075 Chr.21: 45073029-45073029 on Build GRCh38
rs530071304 Chr.21: 45073205-45073205 on Build GRCh38
rs527576199 Chr.21: 45073377-45073377 on Build GRCh38
rs759690324 Chr.21: 45073460-45073460 on Build GRCh38
rs909042959 Chr.21: 45073375-45073375 on Build GRCh38
rs768711115 Chr.21: 45073245-45073245 on Build GRCh38
rs984581882 Chr.21: 45073108-45073108 on Build GRCh38
rs112113119 Chr.21: 45073161-45073161 on Build GRCh38
rs930900996 Chr.21: 45073349-45073349 on Build GRCh38
rs145121069 Chr.21: 45073003-45073003 on Build GRCh38
rs1044674999 Chr.21: 45073069-45073069 on Build GRCh38
rs1040503431 Chr.21: 45073393-45073393 on Build GRCh38
rs991948966 Chr.21: 45073263-45073263 on Build GRCh38
rs1005274593 Chr.21: 45073390-45073390 on Build GRCh38
rs923576432 Chr.21: 45073004-45073004 on Build GRCh38
rs915814 Chr.21: 45073088-45073088 on Build GRCh38
rs1043867905 Chr.21: 45073244-45073244 on Build GRCh38
rs956343755 Chr.21: 45073253-45073253 on Build GRCh38
rs1049097826 Chr.21: 45073110-45073110 on Build GRCh38
rs886416947 Chr.21: 45073127-45073127 on Build GRCh38
rs967740468 Chr.21: 45073039-45073039 on Build GRCh38
rs933979991 Chr.21: 45073204-45073204 on Build GRCh38
rs960428682 Chr.21: 45073081-45073081 on Build GRCh38
rs935082996 Chr.21: 45073019-45073019 on Build GRCh38
rs1044949950 Chr.21: 45073307-45073307 on Build GRCh38
rs974716921 Chr.21: 45072955-45072955 on Build GRCh38
rs757880051 Chr.21: 45073160-45073160 on Build GRCh38
rs927410441 Chr.21: 45073335-45073335 on Build GRCh38
rs781337980 Chr.21: 45073251-45073251 on Build GRCh38
rs979280295 Chr.21: 45073249-45073249 on Build GRCh38
rs780227363 Chr.21: 45072968-45072968 on Build GRCh38
rs1009668427 Chr.21: 45073015-45073015 on Build GRCh38
rs1025985327 Chr.21: 45073299-45073299 on Build GRCh38
rs1033762709 Chr.21: 45073070-45073070 on Build GRCh38
rs1051071248 Chr.21: 45073429-45073429 on Build GRCh38
rs964562406 Chr.21: 45073387-45073387 on Build GRCh38
rs917435070 Chr.21: 45073031-45073031 on Build GRCh38
rs913940914 Chr.21: 45073445-45073445 on Build GRCh38
rs1049313874 Chr.21: 45073372-45073372 on Build GRCh38
rs142287788 Chr.21: 45073378-45073378 on Build GRCh38
rs57550246 Chr.21: 45073242-45073242 on Build GRCh38
rs745378326 Chr.21: 45073382-45073382 on Build GRCh38
rs375030923 Chr.21: 45073371-45073371 on Build GRCh38
rs1029246955 Chr.21: 45073411-45073411 on Build GRCh38
rs892734457 Chr.21: 45073011-45073011 on Build GRCh38
rs116214745 Chr.21: 45073292-45073292 on Build GRCh38
rs919151321 Chr.21: 45073097-45073097 on Build GRCh38
rs2838770 Chr.21: 45072982-45072982 on Build GRCh38
rs866182237 Chr.21: 45073396-45073396 on Build GRCh38
rs1030861115 Chr.21: 45073448-45073448 on Build GRCh38
rs950285398 Chr.21: 45073295-45073295 on Build GRCh38
rs1000748770 Chr.21: 45073425-45073425 on Build GRCh38
rs370232464 Chr.21: 45073355-45073355 on Build GRCh38

View Full Product Details