Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 21010283-21010790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
P2RX6
Entrez Gene ID
9127
Gene Name
purinergic receptor P2X 6
Gene Aliases
P2RXL1, P2X6, P2XM
UniGene
Hs.113275
Gene Chromosome Location
Chr.22: 21009699-21028013 on Build GRCh38
Transcripts
XM_017029075.1 NR_027051.1 NR_003608.1

Gene Symbol
THAP7-AS1
Entrez Gene ID
439931
Gene Name
THAP7 antisense RNA 1
Gene Aliases
-
UniGene
Hs.517430
Gene Chromosome Location
Chr.22: 21001922-21010374 on Build GRCh38
Transcripts
XM_017029075.1 NR_027051.1 NR_003608.1

Gene Symbol
TUBA3FP
Entrez Gene ID
113691
Gene Name
tubulin alpha 3f pseudogene
Gene Aliases
-
UniGene
Hs.585006
Gene Chromosome Location
Chr.22: 21008203-21014287 on Build GRCh38
Transcripts
XM_017029075.1 NR_027051.1 NR_003608.1

SNP Detail

SNP ID Chromosome Location
rs766445808 Chr.22: 21010388-21010388 on Build GRCh38
rs984828837 Chr.22: 21010391-21010392 on Build GRCh38
rs998160869 Chr.22: 21010589-21010589 on Build GRCh38
rs748708988 Chr.22: 21010640-21010640 on Build GRCh38
rs972136183 Chr.22: 21010664-21010664 on Build GRCh38
rs2516545 Chr.22: 21010578-21010578 on Build GRCh38
rs981083397 Chr.22: 21010710-21010710 on Build GRCh38
rs902020980 Chr.22: 21010579-21010579 on Build GRCh38
rs953497472 Chr.22: 21010634-21010634 on Build GRCh38
rs917766267 Chr.22: 21010439-21010439 on Build GRCh38
rs555898407 Chr.22: 21010518-21010518 on Build GRCh38
rs918049755 Chr.22: 21010310-21010310 on Build GRCh38
rs578005701 Chr.22: 21010673-21010673 on Build GRCh38
rs949298904 Chr.22: 21010468-21010468 on Build GRCh38
rs541272198 Chr.22: 21010628-21010628 on Build GRCh38
rs1029277780 Chr.22: 21010593-21010593 on Build GRCh38
rs962069856 Chr.22: 21010657-21010657 on Build GRCh38
rs546172096 Chr.22: 21010735-21010735 on Build GRCh38
rs940526391 Chr.22: 21010406-21010406 on Build GRCh38
rs868449836 Chr.22: 21010755-21010755 on Build GRCh38
rs904838851 Chr.22: 21010504-21010504 on Build GRCh38
rs80026591 Chr.22: 21010432-21010432 on Build GRCh38
rs949271256 Chr.22: 21010706-21010706 on Build GRCh38
rs1000564618 Chr.22: 21010512-21010512 on Build GRCh38
rs558074076 Chr.22: 21010668-21010668 on Build GRCh38
rs547663438 Chr.22: 21010382-21010382 on Build GRCh38
rs5997148 Chr.22: 21010343-21010343 on Build GRCh38
rs940005278 Chr.22: 21010340-21010340 on Build GRCh38
rs1016221914 Chr.22: 21010643-21010643 on Build GRCh38
rs569688548 Chr.22: 21010537-21010537 on Build GRCh38
rs909337778 Chr.22: 21010405-21010405 on Build GRCh38
rs182391782 Chr.22: 21010311-21010311 on Build GRCh38
rs976366596 Chr.22: 21010330-21010330 on Build GRCh38
rs141964913 Chr.22: 21010561-21010561 on Build GRCh38
rs765021892 Chr.22: 21010568-21010568 on Build GRCh38
rs917719575 Chr.22: 21010676-21010676 on Build GRCh38
rs377332062 Chr.22: 21010539-21010539 on Build GRCh38
rs527915193 Chr.22: 21010348-21010348 on Build GRCh38
rs1045006573 Chr.22: 21010502-21010502 on Build GRCh38
rs79055771 Chr.22: 21010307-21010307 on Build GRCh38
rs395938 Chr.22: 21010560-21010560 on Build GRCh38
rs539039910 Chr.22: 21010484-21010484 on Build GRCh38

View Full Product Details