Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 23689940-23690502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GUSBP11
Entrez Gene ID
91316
Gene Name
glucuronidase, beta pseudogene 11
Gene Aliases
-
UniGene
Hs.148656
Gene Chromosome Location
Chr.22: 23638487-23717423 on Build GRCh38
Transcripts
NR_024448.2

SNP Detail

SNP ID Chromosome Location
rs768394075 Chr.22: 23690221-23690221 on Build GRCh38
rs917791968 Chr.22: 23690254-23690254 on Build GRCh38
rs1036741816 Chr.22: 23690026-23690026 on Build GRCh38
rs180781136 Chr.22: 23689971-23689971 on Build GRCh38
rs1035258113 Chr.22: 23690431-23690431 on Build GRCh38
rs746136090 Chr.22: 23690200-23690200 on Build GRCh38
rs944767494 Chr.22: 23690105-23690105 on Build GRCh38
rs946426528 Chr.22: 23690016-23690016 on Build GRCh38
rs536355085 Chr.22: 23690111-23690111 on Build GRCh38
rs1044009934 Chr.22: 23690020-23690020 on Build GRCh38
rs1050919586 Chr.22: 23690082-23690082 on Build GRCh38
rs992046480 Chr.22: 23690240-23690240 on Build GRCh38
rs986751068 Chr.22: 23690333-23690333 on Build GRCh38
rs935261966 Chr.22: 23690329-23690329 on Build GRCh38
rs961303262 Chr.22: 23690451-23690451 on Build GRCh38
rs1024859983 Chr.22: 23690421-23690421 on Build GRCh38
rs560220685 Chr.22: 23690369-23690369 on Build GRCh38
rs761673546 Chr.22: 23690281-23690281 on Build GRCh38
rs943650322 Chr.22: 23690296-23690296 on Build GRCh38
rs937955426 Chr.22: 23689982-23689982 on Build GRCh38
rs781243044 Chr.22: 23690177-23690177 on Build GRCh38
rs550781944 Chr.22: 23690083-23690083 on Build GRCh38
rs540050344 Chr.22: 23690366-23690366 on Build GRCh38
rs1013515384 Chr.22: 23690408-23690408 on Build GRCh38
rs376328794 Chr.22: 23690176-23690176 on Build GRCh38
rs182605431 Chr.22: 23690340-23690340 on Build GRCh38
rs774147129 Chr.22: 23690258-23690258 on Build GRCh38
rs956604456 Chr.22: 23690475-23690475 on Build GRCh38
rs896634869 Chr.22: 23690439-23690439 on Build GRCh38
rs775418186 Chr.22: 23690394-23690394 on Build GRCh38
rs994866520 Chr.22: 23690163-23690163 on Build GRCh38
rs1009809568 Chr.22: 23690358-23690358 on Build GRCh38
rs550502721 Chr.22: 23690292-23690292 on Build GRCh38
rs531844659 Chr.22: 23690471-23690471 on Build GRCh38
rs979524791 Chr.22: 23690209-23690209 on Build GRCh38
rs773259530 Chr.22: 23690395-23690395 on Build GRCh38
rs917785304 Chr.22: 23689990-23689990 on Build GRCh38
rs939825542 Chr.22: 23690012-23690012 on Build GRCh38
rs189971089 Chr.22: 23690288-23690288 on Build GRCh38
rs554807093 Chr.22: 23690339-23690339 on Build GRCh38
rs185201326 Chr.22: 23690210-23690210 on Build GRCh38
rs545798109 Chr.22: 23690440-23690440 on Build GRCh38
rs1042974123 Chr.22: 23690361-23690361 on Build GRCh38
rs1009716319 Chr.22: 23690137-23690137 on Build GRCh38
rs765953163 Chr.22: 23690463-23690463 on Build GRCh38
rs898213949 Chr.22: 23690069-23690069 on Build GRCh38
rs1030660761 Chr.22: 23690473-23690473 on Build GRCh38
rs557426120 Chr.22: 23690069-23690071 on Build GRCh38
rs771096277 Chr.22: 23690143-23690143 on Build GRCh38
rs990563170 Chr.22: 23689995-23689995 on Build GRCh38
rs886702634 Chr.22: 23690199-23690199 on Build GRCh38
rs567471491 Chr.22: 23690104-23690104 on Build GRCh38
rs544491485 Chr.22: 23690335-23690335 on Build GRCh38
rs919755389 Chr.22: 23690279-23690279 on Build GRCh38
rs931078115 Chr.22: 23690280-23690280 on Build GRCh38
rs968567599 Chr.22: 23690152-23690152 on Build GRCh38
rs141047078 Chr.22: 23690422-23690422 on Build GRCh38
rs1028294661 Chr.22: 23690174-23690174 on Build GRCh38
rs17003376 Chr.22: 23690142-23690142 on Build GRCh38
rs531074007 Chr.22: 23690065-23690065 on Build GRCh38
rs889440400 Chr.22: 23690090-23690090 on Build GRCh38
rs979127470 Chr.22: 23690306-23690306 on Build GRCh38
rs566851815 Chr.22: 23690194-23690194 on Build GRCh38
rs1007853711 Chr.22: 23690098-23690098 on Build GRCh38
rs532091615 Chr.22: 23690380-23690380 on Build GRCh38
rs117163650 Chr.22: 23690277-23690277 on Build GRCh38
rs979813084 Chr.22: 23689966-23689966 on Build GRCh38
rs1023768920 Chr.22: 23690457-23690457 on Build GRCh38
rs147714995 Chr.22: 23689986-23689986 on Build GRCh38
rs1039114437 Chr.22: 23690359-23690359 on Build GRCh38
rs902018996 Chr.22: 23690129-23690129 on Build GRCh38
rs960889167 Chr.22: 23690224-23690224 on Build GRCh38
rs1034138827 Chr.22: 23690220-23690220 on Build GRCh38
rs931005061 Chr.22: 23690074-23690074 on Build GRCh38
rs923813953 Chr.22: 23690307-23690307 on Build GRCh38

View Full Product Details