Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24257764-24258310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POM121L9P
Entrez Gene ID
29774
Gene Name
POM121 transmembrane nucleoporin like 9, pseudogene
Gene Aliases
DKFZP434P211, UNQ2565
UniGene
Hs.534980
Gene Chromosome Location
Chr.22: 24251621-24265524 on Build GRCh38
Transcripts
NR_003714.1

SNP Detail

SNP ID Chromosome Location
rs2095939 Chr.22: 24258258-24258258 on Build GRCh38
rs1016386953 Chr.22: 24258042-24258042 on Build GRCh38
rs963240618 Chr.22: 24257848-24257848 on Build GRCh38
rs868241036 Chr.22: 24258008-24258008 on Build GRCh38
rs546878179 Chr.22: 24257980-24257980 on Build GRCh38
rs989225881 Chr.22: 24257911-24257911 on Build GRCh38
rs397843153 Chr.22: 24258268-24258268 on Build GRCh38
rs79996970 Chr.22: 24258092-24258092 on Build GRCh38
rs982478200 Chr.22: 24258170-24258170 on Build GRCh38
rs552408331 Chr.22: 24257953-24257953 on Build GRCh38
rs867184698 Chr.22: 24258100-24258100 on Build GRCh38
rs1033551853 Chr.22: 24258240-24258240 on Build GRCh38
rs563612816 Chr.22: 24257889-24257889 on Build GRCh38
rs3869878 Chr.22: 24257828-24257828 on Build GRCh38
rs778801684 Chr.22: 24257809-24257812 on Build GRCh38
rs531070611 Chr.22: 24257890-24257890 on Build GRCh38
rs4050455 Chr.22: 24258226-24258226 on Build GRCh38
rs3868228 Chr.22: 24257790-24257790 on Build GRCh38
rs574877483 Chr.22: 24257858-24257858 on Build GRCh38
rs542255414 Chr.22: 24257863-24257863 on Build GRCh38
rs3869877 Chr.22: 24257795-24257795 on Build GRCh38
rs970676550 Chr.22: 24258157-24258157 on Build GRCh38
rs914981766 Chr.22: 24257923-24257923 on Build GRCh38
rs3869880 Chr.22: 24257909-24257909 on Build GRCh38
rs4050026 Chr.22: 24257993-24257993 on Build GRCh38
rs972292887 Chr.22: 24257858-24257858 on Build GRCh38
rs868111240 Chr.22: 24258001-24258001 on Build GRCh38
rs569564836 Chr.22: 24258064-24258064 on Build GRCh38
rs200611128 Chr.22: 24257960-24257960 on Build GRCh38
rs371869831 Chr.22: 24257841-24257841 on Build GRCh38
rs947604193 Chr.22: 24257938-24257938 on Build GRCh38
rs4050453 Chr.22: 24258228-24258228 on Build GRCh38
rs997545329 Chr.22: 24257801-24257801 on Build GRCh38
rs889159242 Chr.22: 24258017-24258017 on Build GRCh38
rs889213210 Chr.22: 24257837-24257837 on Build GRCh38
rs3984038 Chr.22: 24258176-24258176 on Build GRCh38
rs376518267 Chr.22: 24258180-24258180 on Build GRCh38
rs369387134 Chr.22: 24258269-24258269 on Build GRCh38
rs4050454 Chr.22: 24258227-24258227 on Build GRCh38
rs796959080 Chr.22: 24257811-24257811 on Build GRCh38
rs535846507 Chr.22: 24258010-24258010 on Build GRCh38
rs899226818 Chr.22: 24258048-24258048 on Build GRCh38
rs1007557865 Chr.22: 24258018-24258018 on Build GRCh38
rs62232019 Chr.22: 24258036-24258036 on Build GRCh38
rs62232020 Chr.22: 24258047-24258047 on Build GRCh38
rs879995265 Chr.22: 24258009-24258009 on Build GRCh38
rs878891196 Chr.22: 24257875-24257875 on Build GRCh38
rs1051772192 Chr.22: 24258015-24258015 on Build GRCh38
rs926254790 Chr.22: 24257884-24257884 on Build GRCh38
rs3875969 Chr.22: 24257917-24257917 on Build GRCh38
rs989090166 Chr.22: 24258271-24258271 on Build GRCh38
rs937678584 Chr.22: 24257902-24257902 on Build GRCh38
rs3984021 Chr.22: 24258283-24258283 on Build GRCh38
rs959062177 Chr.22: 24258261-24258261 on Build GRCh38
rs568458890 Chr.22: 24257992-24257992 on Build GRCh38
rs558687513 Chr.22: 24258127-24258127 on Build GRCh38
rs755952411 Chr.22: 24257927-24257927 on Build GRCh38
rs564313666 Chr.22: 24257956-24257956 on Build GRCh38
rs1016354349 Chr.22: 24257846-24257846 on Build GRCh38
rs5751830 Chr.22: 24258082-24258082 on Build GRCh38
rs62232022 Chr.22: 24258181-24258181 on Build GRCh38
rs528668865 Chr.22: 24257961-24257961 on Build GRCh38
rs548202304 Chr.22: 24258057-24258057 on Build GRCh38
rs915011088 Chr.22: 24258287-24258287 on Build GRCh38
rs4050035 Chr.22: 24258146-24258146 on Build GRCh38
rs3984016 Chr.22: 24258259-24258259 on Build GRCh38
rs62232021 Chr.22: 24258091-24258091 on Build GRCh38
rs3984012 Chr.22: 24258201-24258201 on Build GRCh38
rs878975697 Chr.22: 24258103-24258103 on Build GRCh38
rs971025471 Chr.22: 24257874-24257874 on Build GRCh38

View Full Product Details