Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 21406096-21406649 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VENTXP7
Entrez Gene ID
391518
Gene Name
VENT homeobox pseudogene 7
Gene Aliases
HPX42, VENTX1
UniGene
Hs.543226
Gene Chromosome Location
Chr.3: 21405726-21406708 on Build GRCh38
Transcripts
NR_002311.1

SNP Detail

SNP ID Chromosome Location
rs963986643 Chr.3: 21406318-21406318 on Build GRCh38
rs533332772 Chr.3: 21406295-21406295 on Build GRCh38
rs1054042257 Chr.3: 21406606-21406606 on Build GRCh38
rs372420655 Chr.3: 21406473-21406473 on Build GRCh38
rs546852717 Chr.3: 21406417-21406417 on Build GRCh38
rs755381238 Chr.3: 21406576-21406576 on Build GRCh38
rs959131220 Chr.3: 21406233-21406233 on Build GRCh38
rs770478083 Chr.3: 21406471-21406471 on Build GRCh38
rs943431933 Chr.3: 21406349-21406349 on Build GRCh38
rs913104018 Chr.3: 21406161-21406161 on Build GRCh38
rs557396339 Chr.3: 21406176-21406176 on Build GRCh38
rs1000833792 Chr.3: 21406254-21406254 on Build GRCh38
rs377531093 Chr.3: 21406498-21406498 on Build GRCh38
rs780280828 Chr.3: 21406560-21406560 on Build GRCh38
rs911156758 Chr.3: 21406314-21406314 on Build GRCh38
rs893200116 Chr.3: 21406163-21406163 on Build GRCh38
rs779101639 Chr.3: 21406187-21406187 on Build GRCh38
rs180877648 Chr.3: 21406390-21406390 on Build GRCh38
rs950284939 Chr.3: 21406247-21406247 on Build GRCh38
rs949165573 Chr.3: 21406574-21406574 on Build GRCh38
rs767203005 Chr.3: 21406557-21406557 on Build GRCh38
rs190591886 Chr.3: 21406141-21406141 on Build GRCh38
rs924841515 Chr.3: 21406305-21406305 on Build GRCh38
rs142715866 Chr.3: 21406155-21406155 on Build GRCh38
rs567647995 Chr.3: 21406402-21406402 on Build GRCh38
rs777157931 Chr.3: 21406440-21406440 on Build GRCh38
rs1032372821 Chr.3: 21406262-21406262 on Build GRCh38
rs943361828 Chr.3: 21406556-21406556 on Build GRCh38
rs1009687914 Chr.3: 21406288-21406288 on Build GRCh38
rs371788955 Chr.3: 21406528-21406529 on Build GRCh38
rs141708999 Chr.3: 21406389-21406389 on Build GRCh38
rs563002072 Chr.3: 21406618-21406618 on Build GRCh38
rs1015904379 Chr.3: 21406417-21406417 on Build GRCh38
rs800605 Chr.3: 21406404-21406404 on Build GRCh38
rs1052226487 Chr.3: 21406627-21406627 on Build GRCh38
rs1046817269 Chr.3: 21406379-21406379 on Build GRCh38
rs936885164 Chr.3: 21406585-21406585 on Build GRCh38
rs918038408 Chr.3: 21406388-21406388 on Build GRCh38
rs138058311 Chr.3: 21406243-21406243 on Build GRCh38
rs527719987 Chr.3: 21406413-21406414 on Build GRCh38
rs376847119 Chr.3: 21406558-21406558 on Build GRCh38
rs753877585 Chr.3: 21406460-21406460 on Build GRCh38
rs892475664 Chr.3: 21406455-21406455 on Build GRCh38
rs892436925 Chr.3: 21406607-21406607 on Build GRCh38
rs1047670260 Chr.3: 21406225-21406225 on Build GRCh38
rs1030435202 Chr.3: 21406461-21406461 on Build GRCh38
rs566795354 Chr.3: 21406523-21406524 on Build GRCh38
rs430387 Chr.3: 21406307-21406307 on Build GRCh38
rs440905 Chr.3: 21406342-21406342 on Build GRCh38
rs1047532061 Chr.3: 21406569-21406569 on Build GRCh38
rs781691080 Chr.3: 21406171-21406171 on Build GRCh38
rs956987937 Chr.3: 21406268-21406268 on Build GRCh38
rs1001096556 Chr.3: 21406433-21406433 on Build GRCh38
rs112237068 Chr.3: 21406409-21406410 on Build GRCh38
rs974131212 Chr.3: 21406331-21406331 on Build GRCh38
rs1039281052 Chr.3: 21406384-21406384 on Build GRCh38
rs963955899 Chr.3: 21406517-21406517 on Build GRCh38
rs748577881 Chr.3: 21406184-21406184 on Build GRCh38
rs544213941 Chr.3: 21406579-21406579 on Build GRCh38
rs150567334 Chr.3: 21406544-21406544 on Build GRCh38
rs776644 Chr.3: 21406465-21406465 on Build GRCh38
rs781520824 Chr.3: 21406624-21406624 on Build GRCh38
rs770530195 Chr.3: 21406222-21406222 on Build GRCh38
rs72625843 Chr.3: 21406543-21406543 on Build GRCh38
rs981587486 Chr.3: 21406510-21406511 on Build GRCh38
rs539986301 Chr.3: 21406216-21406216 on Build GRCh38
rs550573657 Chr.3: 21406414-21406414 on Build GRCh38
rs561339050 Chr.3: 21406230-21406230 on Build GRCh38
rs926833555 Chr.3: 21406577-21406577 on Build GRCh38
rs757440919 Chr.3: 21406168-21406169 on Build GRCh38
rs1038099592 Chr.3: 21406199-21406199 on Build GRCh38
rs934869131 Chr.3: 21406525-21406525 on Build GRCh38
rs929705000 Chr.3: 21406373-21406373 on Build GRCh38
rs989038364 Chr.3: 21406147-21406147 on Build GRCh38
rs563220495 Chr.3: 21406293-21406293 on Build GRCh38
rs539211823 Chr.3: 21406470-21406470 on Build GRCh38
rs1020503312 Chr.3: 21406291-21406291 on Build GRCh38
rs147375591 Chr.3: 21406181-21406181 on Build GRCh38
rs531213425 Chr.3: 21406567-21406567 on Build GRCh38
rs1045291835 Chr.3: 21406237-21406237 on Build GRCh38
rs11718548 Chr.3: 21406232-21406232 on Build GRCh38
rs923345677 Chr.3: 21406566-21406566 on Build GRCh38
rs986724652 Chr.3: 21406174-21406174 on Build GRCh38
rs11129005 Chr.3: 21406505-21406505 on Build GRCh38
rs572744388 Chr.3: 21406518-21406518 on Build GRCh38
rs555281269 Chr.3: 21406403-21406412 on Build GRCh38
rs867636356 Chr.3: 21406228-21406228 on Build GRCh38

View Full Product Details